RNA Stabilization Solution is an aqueous, non-toxic tissue storage reagent that protects RNA within intact, unfrozen samples. RNAlater® was designed to eliminate the need to immediately process tissue samples or to freeze samples in liquid nitrogen for later processing. Recently, Dr. Christian Koppelstaetter and colleagues (Medical University Innsbruck, Austria and Medical University of Silesia, Poland) completed a study using quantitative PCR (qPCR) to look at telomeric DNA. They found that while tissues processed immediately or treated with RNAlater produced similar results, formaldehyde-fixed tissue generated inconsistent measurements of telomere length [1].
Ordering Information
Catalog # Name Size Price (USD) Qty
AM7020 RNAlater™ Stabilization Solution, 100 mL Each
206.65
Online Exclusive
215.00
Save 8.35 (4%)
AM7022 RNAlater™ Stabilization Solution, 50 x 1.5 mL Each
351.65
Online Exclusive
362.00
Save 10.35 (3%)
AM7021 RNAlater™ Stabilization Solution, 500 mL Each
585.65
Online Exclusive
602.00
Save 16.35 (3%)
AM7024 RNAlater™ Stabilization Solution, 1 x 250 mL Each
458.65
Online Exclusive
481.00
Save 22.35 (5%)
AM7023 RNAlater™ Stabilization Solution, 20 x 5 mL Each
388.65
Online Exclusive
407.00
Save 18.35 (5%)
Research on Aging and Telomere Length
Telomeres are the non-coding, repetitive DNA sequences at the ends of chromosomes that are required for chromosome positioning in the nucleus and for DNA protection from recombination, degradation, and replicative damage. In cellular senescence research, the length of the telomeres can be used as a marker for biological age, where older cells have shorter telomeres. With the hope that various tissue archives could be used in aging studies, Dr. Koppelstaetter and colleagues sought to assess whether different types of sample preservation methods would still allow accurate determination of telomere length by a commonly accepted technique used with small sample sizes (qPCR: ratio of the amount of amplicon from telomere sequences to that of a single copy gene).
RNAlater, but Not Formaldehyde, Stabilizes DNA in Archived Samples
Normal renal cortical tissue was obtained from a kidney removed due to carcinoma. DNA immediately isolated from one piece served as the reference sample. Triplicate samples, preserved in either RNA later or 4% formaldehyde, were stored in darkness at room temperature for one of six different time periods (range: 3–168 hr). DNA was isolated from the samples and qPCR assays were run with telomere specific primers, as well as primers specific for three single copy genes (36B4, GAPDH, b-actin).
At all time points, RNA later-treated samples gave results essentially identical to the reference sample for relative telomere length. In contrast, formaldehyde fixation caused an apparent time-dependent increase in relative telomere length, which was mainly due to inconsistencies in the amplification of the single copy gene PCR products. Formaldehyde, which is known to denature double-stranded DNA at AT-rich regions, destabilized genomic DNA in the samples, so that amplification of the longer PCR products assessed in this study had the most variability. In contrast, RNA later has been proven to stabilize RNA and is now shown to preserve DNA within tissues. For additional details, see Koppelstaetter, et al. [1].
References
  1. Koppelstaetter C, Jennings P, Hochegger K, Perco P, Ischia R, Karkoszka H, Mayer G (2005) Effect of tissue fixatives on telomere length determination by quantitative PCR, Mech Ageing Dev 126(12): 1331–1333. Epub 2005 Sep 22.