Search
TOPO TA subcloning kits utilize our popular pCR 2.1-TOPO TA vector for the direct ligation of Taq-amplified PCR products in a 5-minute, benchtop reaction yielding up to 95% recombinants.
The pCR 2.1-TOPO TA vector features:
Each TOPO TA Cloning Kit contains linearized and topoisomerase I-activated pCR 2.1-TOPO Vector, Salt Solution, dNTPs, Control Template and Primers, M13 Forward and Reverse Primers, One Shot Chemically Competent or Electrocomp E. coli, S.O.C. Medium, and a supercoiled control plasmid. TheTOPO TA Cloning Kit for Subcloning without competent cells allows the flexibility of using your own strain for increased flexibility so competent cells and S.O.C. Medium are not provided.
In addition, TOPO TA Cloning Kits are available in combo kits combined with a PureLink Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO-cloned inserts for downstream analysis.
Thermostable polymerases containing extensive 3´ to 5´ exonuclease activity, such as Platinum SuperFi, do not leave 3´ A-overhangs. PCR products generated with Taq polymerase have a high efficiency of cloning in the TOPO TA subcloning kits because the 3´ A-overhangs are not removed. However, if you use a proofreading polymerase or wish to clone blunt-ended fragments, you can add 3´ A-overhangs by incubating with Taq at the end of your cycling program.
Alternatively, you may want to try Zero Blunt TOPO PCR Cloning Kits. These kits offer fast and efficient cloning of blunt-end PCR products generated using thermostable, proofreading polymerases. They also utilize the ccdB gene for positive selection.
| Cat. No. | Product description | Competent cells supplied | Number of reactions | |
|---|---|---|---|---|
| 451641 | TOPO TA Cloning Kit for Subcloning | None | 10 | Order now |
| K4500J10 | TOPO TA Cloning Kit for Subcloning | TOP10 (genotype) | 10 | Order now |
| 450641 | TOPO TA Cloning Kit for Subcloning, without chemically competent cells | None | 25 | Order now |
| K450001 | TOPO TA Cloning Kit for Subcloning, with One Shot TOP10 Chemically Competent E. coli | TOP10 (genotype) | 25 | Order now |
| K452001 | TOPO TA Cloning Kit with One Shot MAX Efficiency DH5α T1 Phage-Resistant Chemically Competent E. coli | MAX Efficiency DH5α-T1R (genotype) | 25 | Order now |
| K455001 | TOPO TA Cloning Kit for Subcloning, with One Shot TOP10F' Chemically Competent E. coli | TOP10F' (genotype) | 25 | Order now |
| K456001 | TOPO TA Cloning Kit for Subcloning, with One Shot TOP10 Electrocomp E. coli | Electrocompetent TOP10 (genotype) | 25 | Order now |
| K451020 | TOPO TA Cloning Kit for Subcloning, with One Shot Mach1 Phage-Resistant Chemically Competent E. coli | Mach1 T1R (genotype) | 25 | Order now |
| K450002 | TOPO TA Cloning Kit with One Shot TOP10 Chemically Competent E. coli and PureLink Quick Plasmid Miniprep Kit | TOP10 (genotype) | 25 | Order now |
| K451022 | TOPO TA Cloning Kit for Subcloning, with One Shot Mach1 Phage-Resistant Chemically Competent E. coli and PureLink Quick Plasmid Miniprep Kit | Mach1 T1R (genotype) | 25 | Order now |
| K450040 | TOPO TA Cloning Kit for Subcloning, with One Shot TOP10 Chemically Competent E. coli | TOP10 (genotype) | 50 | Order now |
| K452040 | TOPO TA Cloning Kit with One Shot MAX Efficiency DH5α-T1 Phage-Resistant Chemically Competent E. coli | MAX Efficiency DH5α-T1R (genotype) | 50 | Order now |
| K455040 | TOPO TA Cloning Kit for Subcloning, with One Shot TOP10F' Chemically Competent E. coli | TOP10F' (genotype) | 50 | Order Now |
| K456040 | TOPO TA Cloning Kit for Subcloning, with One Shot TOP10 Electrocomp E. coli | Electrocompetent TOP10 (genotype) | 50 | Order now |
Cloning Support Center
Find tips, troubleshooting help, and resources for your cloning applications
Cloning Education
Learn about PCR cloning strategies and subcloning approaches
PCR Education
Review the principles of PCR, components, cycling parameters, methods, and applications
For Research Use Only. Not for use in diagnostic procedures.