Blasticidin
Table 1 - Suggested Blasticidin Concentrations
Cell Line | Concentration [ug/ml] | Reference |
---|---|---|
HT1080 |
1-5 10 20 |
J Biol Chem 283:11556–11564 (2008) J Virol 82:7325–7335 (2008) Mol Biol Cell 19:8–16 (2008) |
HeLa |
10 10 20 |
J Gen Virol 89:2611–2621 (2008) Mol Biol Cell 19:8–16 (2008). J Immunol 181:22–26 (2008) |
HEK293 |
5 10 |
J Biol Chem 283:27534–27546 (2008) J Virol 82:1665–1678 (2008) |
Hep G2 |
4 5 |
J Biol Chem 283:16320–16331 (2008) Blood 113: 1786–1793 (2009) |
MCF-7 |
2 5 |
Mol Cancer Res 6:555–567 (2008) Mol Endocrinol 22: 361–379 (2008) |
A549 (lung cancer) |
10 |
Cancer Res 68:5040–5048 (2008) |