Invitrogen Lipofectamine 3000 Transfection Reagent is a highly efficient, cost-effective tool for lentiviral production (Figure 1). This versatile reagent enables high viral titers even with genes that are large or difficult to package. Here, we demonstrate the effectiveness of Lipofectamine 3000 reagent for the production of high-quality, high-titer lentivirus while minimizing time and reagent usage. A workflow is provided in Figure 2, followed by a more detailed protocol.
Lentiviral production involves the generation of lentiviruses, which are a type of retrovirus used extensively for gene delivery in various research and therapeutic applications due to their ability to integrate into the host genome and provide long-term expression of transgenes. Cells commonly used for lentivirus production are human embryonic kidney (HEK) 293T cells, which are highly transfectable and support high levels of viral protein expression. A transfection reagent is a chemical or biological agent used to introduce nucleic acids, such as DNA or RNA, into cells to facilitate gene expression or knockdown. The best transfection reagent depends on the specific application and cell type, but an excellent option is Lipofectamine 3000 Reagent.
Advantages of lentiviral production using Lipofectamine 3000 reagent:
- Superior performance—higher unconcentrated viral titers than leading reagents (Figure 3).
- Larger gene sizes—high viral titers even with genes that are large or difficult to package.
- Simpler—no additional reagents required for boosting lentiviral production (e.g., chloroquine phosphate, sodium butyrate, caffeine).
- Faster—time to virus collection is 24 hours less than other reagents, and reverse transfection time is almost 2 days less than other reagents with comparable viral titers.
- Easier—eliminates the need for coating of plates or flasks.
Figure 1. Lentiviral production using Lipofectamine 3000 reagent. A lentiviral construct containing the gene of interest along with lentiviral packaging mix is cotransfected into 293T or 293FT cells using Lipofectamine 3000 reagent. Following incubation of cells, supernatant containing lentivirus is harvested and cellular debris is removed by centrifugation. Depending on the lentiviral construct, viral titer is determined either by blasticidin selection or analyzing the percentage of GFP-positive cells.
Figure 2. Lentiviral production workflow using Lipofectamine 3000 reagent.
Figure 3. Lipofectamine 3000 reagent performs significantly better than leading competitor transfection reagents for lentiviral production. (A) Titers obtained using Lipofectamine 3000 reagent compared to commonly used reagents in 293FT cells using a 3 kb insert gene. (B) Summary of fold increases in titer levels using two different cell lines and insert sizes. (C) Serial dilution and Blasticidin selection of concentrated virus demonstrates the high titer levels achieved using Lipofectamine 3000 reagent.
Detailed protocol for lentiviral production using Lipofectamine 3000 reagent
NOTE: All virus preparation and work should be in accordance with your institution’s guidelines for Biosafety Level 2 (BL-2) protocols.
Materials
| Reagent | Cat. No. |
|---|
| DMEM, high glucose, GlutaMAX Supplement, pyruvate | 10569010 |
| Fetal Bovine Serum, certified, US origin | 16000044 |
| Geneticin Selective Antibiotic (50 mg/mL) | 10131035 |
| Opti-MEM I Reduced Serum Medium, GlutaMAX Supplement | 51985034 |
| Opti-MEM I Reduced Serum Medium | 31985062 |
| Sodium Pyruvate (100 mM) | 11360070 |
| ViraPower Lentiviral Packaging Mix | K497500 |
| Lipofectamine 3000 Transfection Reagent | L3000015 |
| Vivid Colors pLenti6.3/V5-GW/EmGFP Expression Control Vector | V37006 |
Media preparation
Complete culturing medium
500 mL Gibco DMEM, high glucose, GlutaMAX Supplement, pyruvate with 10% Gibco FBS
To prepare complete culturing medium for 293T and 293FT cells, aseptically mix the following components:
- 450 mL DMEM, high glucose, GlutaMAX Supplement, pyruvate
- 50 mL Fetal Bovine Serum, certified, US origin
- If using 293FT cells, additionally include 500 µg/mL Geneticin antibiotic
Lentivirus packaging medium
500 mL Gibco Opti-MEM I Reduced Serum Medium, GlutaMAX Supplement with 1 mM Gibco Sodium Pyruvate and 5% Gibco FBS
To prepare lentivirus packaging medium, aseptically mix the following components:
- 474 mL of Opti-MEM I Reduced Serum Medium, GlutaMAX Supplement
- 25 mL Fetal Bovine Serum, certified, US origin
- 1 mL Sodium Pyruvate (100 mM)
Forward transfection (6-well plate)
Instructions below are tailored to the 6-well plate format. For scaling up transfections, see Table 1.
Day 1 (afternoon): seeding of cells
- Seed 293T or 293FT cells onto 6-well culture plates at 70–80% confluence. For a 6-well culture plate, plate cells at ~1.2 x 106 cells per well in 2 mL of lentivirus packaging medium.
- Incubate cells overnight at 37°C, 5% CO2.
Day 2 (morning): transfection
At time of transfection, cell density should be 95–99% confluent.
- Bring Opti-MEM I Reduced Serum Medium to room temperature and prepare Tube A and Tube B as described in the following table:
| Component | Volume |
|---|
| Tube A | |
| Opti-MEM I Reduced-Serum Medium | 250 µL |
| Lipofecatmine 3000 Transfection Reagent | 7 µL |
| Tube B | |
| Opti-MEM I Reduced-Serum Medium | 250 µL |
| P3000 Enhancer reagent | 6 µL |
| ViraPower Lentiviral Packaging Mix (1 μg/µL) | 2.25 µL |
| pLenti expression vector | 0.75 µL |
NOTE: Lipofectamine 3000 reagent diluted in Opti-MEM medium should be used within 15 minutes of dilution. Longer times can result in a loss of transfection efficiency.
- To prepare lipid-DNA complexes, transfer contents of Tube A to Tube B and mix well.
- Incubate the complexes for 10–20 minutes at room temperature.
- Prior to adding complex, remove 1 mL of medium from each well, leaving a total of 1 mL in each well.
- Add 500 μL of lipid–DNA complex to each well, taking care to dispense liquid against the well wall to avoid disrupting cells. Gently agitate plate to evenly distribute.
- Incubate plate for 6 hours at 37°C, 5% CO2.
- At 6 hours post-transfection, change plate medium from each well. Carefully remove medium that contains lipid-DNA complexes from each well and treat with 10% bleach prior to disposal. Replace with 2 mL of pre-warmed lentivirus packaging medium.
NOTE: Because removed medium contains a small amount of lentivirus, it should be appropriately treated with bleach and discarded as per institutional guidelines.
- Return plate to incubator, and incubate overnight at 37°C, 5% CO2.
Day 3 (morning): harvest of first batch of virus
- At 24 hours post-transfection, collect the 2 mL cell supernatant from each well and store in 15 mL conical tubes at 4°C.
- Replace the collected medium with 2 mL of pre-warmed lentivirus packaging medium.
- Incubate the plate overnight at 37°C, 5% CO2.
Day 4 (mid-afternoon): harvest of second batch of virus
- Approximately 52 hours post-transfection, collect 2 mL of cell supernatant from each well and combine with the first collection for a total of 4 mL of collected supernatant.
NOTE: Prior to disposal, treat all cell culture vessels and tips with 10% bleach.
- Centrifuge the 4 mL of collected supernatant at 2,000 rpm for 10 minutes at room temperature to remove cellular debris. Collect and transfer supernatant, discarding cell pellet.
- Filter the clarified lentiviral supernatant using a 0.45 μm pore filter to remove any remaining cellular debris.
- Aliquot virus into cryovials and store at –80°C. Virus can also be concentrated prior to freezing. Limit freezing and thawing of virus aliquots to maintain titer.
Table 1. Scale-up of viral production using forward transfection.
| Cell culture vessel | Multiplication factor* | Volume of packaging medium for seeding (mL) | 293T or 293FT cell numbers per well† | DNA cotransfection | Volume of lentivirus packaging medium for replacement (mL) | Total volume of harvested lentivirus (mL) |
|---|
| Volume of Opti-MEM I medium for complexing | Lentivirus packaging (DNA) | P3000 reagent (µL) | Lipofectamine 3000 reagent (µL) |
|---|
| ViraPower packaging mix (μg) | pLenti-based vector (μg) |
|---|
| 6-well | 1 | 2 | 1.2 x 106 | 2 x 250 µL | 2.25 | 0.75 | 6 | 7 | 2 | 2 x 2 |
| 60 mm | 2.2 | 4 | 2.6 x 106 | 2 x 500 µL | 5.0 | 1.7 | 13 | 15 | 4 | 2 x 4 |
| 10 cm | 5.8 | 12 | 7.0 x 106 | 2 x 1.5 mL | 13.0 | 4.3 | 35 | 41 | 12 | 2 x 12 |
| T75 | 7.9 | 16 | 9.5 x 106 | 2 x 2 mL | 17.8 | 5.9 | 47 | 55 | 16 | 2 x 16 |
| T175 | 18.4 | 37 | 22.1 x 106 | 2 x 4.6 mL | 41.4 | 13.8 | 111 | 129 | 37 | 2 x 37 |
* Multiplication factor is based on 6-well plate growth area. † Confluency required the day before transfection is ~75–80% and ~95–99% the day of transfection. |
Reverse transfection (6-well plate)
Day 1 (morning): complex formation and seeding of cells
- Prepare cell suspension in pre-warmed lentivirus packaging medium at the appropriate cell concentration (see Table 2).
- Bring Opti-MEM I Reduced Serum Medium to room temperature and prepare Tube A and Tube B as described in the following table:
| Component | Volume |
|---|
| Tube A | |
| Opti-MEM I Reduced-Serum Medium | 250 µL |
| Lipofecatmine 3000 Transfection Reagent | 7 µL |
| Tube B | |
| Opti-MEM I Reduced-Serum Medium | 250 µL |
| P3000 Enhancer reagent | 6 µL |
| ViraPower Lentiviral Packaging Mix (1 μg/µL) | 2.25 µL |
| pLenti expression vector | 0.75 µL |
- Add contents of Tube A to Tube B, and incubate for 10–20 minutes at room temperature.
- Add 500 µL of lipid–DNA complex directly to the plate well.
- Seed 293T or 293FT cells onto 6-well culture plates at ~3.6 x 106 cells per well in 1 mL of lentivirus packaging medium.
NOTE: For reverse transfection, seed cells at 3x higher density than for forward transfection.
- Incubate plate for 6 hours at 37°C, 5% CO2.
Day 1 (afternoon): media change
- At 6 hours post-transfection, change plate medium from each well. Carefully remove complex-containing medium from each well and treat with 10% bleach prior to disposal. Replace with 2 mL of pre-warmed packaging medium.
- Return plate to incubator, and incubate overnight at 37°C, 5% CO2.
Day 2 (morning): harvest of first batch of virus
- At 24 hours post-transfection, collect the 2mL cell supernatant from each well and store in 15 mL conical tubes at 4°C.
- Replace the collected medium with 2 mL of pre-warmed packaging medium.
- Incubate plate overnight at 37°C, 5% CO2.
Day 3 (mid-afternoon): harvest of second batch of virus
- Approximately 52 hours post-transfection, collect 2 mL of cell supernatant from each well and combine with first collection for a total of 4 mL of collected supernatant.
- Centrifuge the 4 mL of collected supernatant at 2,000 rpm for 10 minutes at room temperature to remove cellular debris. Collect and transfer supernatant, discarding cell pellet.
- Filter the clarified lentiviral supernatant using a 0.45 μm pore filter to remove any remaining cellular debris.
- Aliquot virus into cryovials and store at –80°C. Virus can also be concentrated prior to freezing. Limit freezing and thawing of virus aliquots to maintain titer. Titers achieved using forward and reverse transfection are compared in Figure 4.
Table 2. Scale-up of viral production using reverse transfection.
| Cell culture vessel | Multiplication factor* | Volume of packaging medium for seeding (mL) | 293T or 293FT cell numbers per well | DNA cotransfection | Volume of lentivirus packaging medium for replacement (mL) | Total volume of harvested lentivirus (mL) |
|---|
| Volume of Opti-MEM I medium for complexing | Lentivirus packaging (DNA) | P3000 reagent (µL) | Lipofectamine 3000 reagent (µL) |
|---|
| ViraPower packaging mix (μg) | pLenti-based vector (μg) |
|---|
| 6-well | 1 | 1 | 3.6 x 106 | 2 x 250 µL | 2.25 | 0.75 | 6 | 7 | 2 | 2 x 2 |
| 60 mm | 2.2 | 2 | 7.92 x 106 | 2 x 500 µL | 5.0 | 1.7 | 13 | 15 | 4 | 2 x 4 |
| 10 cm | 5.8 | 6 | 21 x 106 | 2 x 1.5 mL | 13.0 | 4.3 | 35 | 41 | 12 | 2 x 12 |
| T75 | 7.9 | 8 | 28.4 x 106 | 2 x 2 mL | 17.8 | 5.9 | 47 | 55 | 16 | 2 x 16 |
| T175 | 18.4 | 18 | 66.2 x 106 | 2 x 4.6 mL | 41.4 | 13.8 | 111 | 129 | 37 | 2 x 37 |
* Multiplication factor is based on 6-well plate growth area.
|
Figure 4. Comparative titers achieved using forward vs. reverse transfection. Titers were compared in 293FT and 293T cells using forward and reverse transfection protocols and 2 different gene sizes. Titer levels were comparable between the two methods.
Measurement of lentiviral titer by GFP selection or Blasticidin selection
Important tips:
Lentivirus must be thawed and remain on ice throughout use. Following thaw, mix virus by gently tapping tube or inverting slowly—do not vortex.
Handling of lentivirus must be performed as per institutional guidelines. All materials should be treated with a 10% bleach solution prior to disposal.
Titering by GFP selection
Materials
- Lentivirus
- HT1080 cell line
- Culture medium:
- Gibco DMEM, high glucose
- GlutaMAX Supplement, pyruvate with 10% FBS
- Polybrene™ reagent at 10 mg/mL
- Corning™ 96-well round-bottom plate for dilutions
Day 1 (morning)
This protocol is tailored for a 96-well plate format for high-throughput flow cytometery analysis.
- Seed a 96-well plate with HT1080 cells at a density of 7,000 cells/well in 100 μL of culture medium.
- Store in an incubator for 4–5 hours until transduction.
- Prior to transduction, freshly prepare virus dilutions as follows:
- Combine 15 mL of fresh culture medium and 12 μL of 10 mg/mL Polybrene reagent (final concentration of 8 mg/mL). Vortex well to combine.
- Per vial sample, add 135 µL of medium prepared in the previous step to 16 wells of a 96-well round-bottom plate in a 4-well by 4-well pattern (see illustration above).
NOTE: n = 4 is recommended for titering.
- Add 15 μL of lentiviral supernatant to each well in row 1 (total volume 150 μL; 1:10 dilution).
- Mix well using a pipette.
- Perform a serial dilution of row 1 (use a multichannel pipette, if available):
- Transfer 15 μL from row 1 to row 2, mix well (1:100)
- Transfer 15 μL from row 2 to row 3, mix well (1:1,000)
- Transfer 15 μL from row 3 to row 4, mix well (1:10,000)
- To transduce cells, remove plating medium from HT1080 cells and transfer 100 µL of the prepared dilutions to each corresponding well (use a multichannel pipette, if available).
- Centrifuge the plate at 2,000 rpm for 30 minutes at room temperature.
- Incubate the cell plate overnight.
Day 2
- Remove medium containing viral supernatant, and replace with fresh HT1080 culture medium (without Polybrene reagent).
- Incubate cells for an additional 3 days. Analyze the percentage of GFP-positive cells (flow cytometry analysis is recommended).
Calculation of titer
To calculate the titer in transducing units (TU) per mL, determine the appropriate dilution factor to use based on the percentage of GFP-positive cells. The desired transduction range is 1–20%.
Use the following formula:
Titer = (F × C/V) × D
where:
F = frequency of GFP-positive cells (percent GFP-positive cells/100)
C = cell number per well at the time of transduction (7,000 cells)
V = volume of inoculum in mL (0.1 mL)
D = lentivirus dilution factor
Example (based on 96-well protocol):
The lentivirus dilution of 104 is chosen for the calculation because the GFP-positive cell value falls within the desired 1–20% range:
F = 4.4/100
C = 7,000 (cell number at time of transduction)
V = 0.1 (100 µL of medium)
D = 104
Calculation as follows:
Titer = (0.044 x 7,000/0.1) x 104 = 3.08 x 107 TU/mL
Titering by Blasticidin selection
Materials
- Lentivirus
- HT1080 cell line
- Culture medium:
- Gibco DMEM, high glucose
- GlutaMAX Supplement, pyruvate with 10% FBS
- Polybrene reagent at 10 mg/mL
- Crystal violet
- Selection medium:
Day 1 (morning)
This protocol is tailored for a 24-well plate format.
- Seed 24-well plate with HT1080 cells at a density of 42,000 cells per well (30–50% confluent) in 500 µL of culture medium.
- Store in incubator for 4–5 hours until transduction.
NOTE: Duplicate titering is recommended.
- Prior to transduction, freshly prepare virus dilutions as follows:
- Combine 15 mL of fresh culture medium with 12 μL of 10 mg/mL Polybrene reagent (final concentration of 8 mg/mL). Vortex well to combine.
- Prepare 5 aliquots of 135 μL of medium from step a, and label tubes as 1–5.
- Add 15 μL of lentivirus to tube 1 and pipette to mix well.
Perform a serial dilution:
i. Transfer 15 µL from tube 1 to tube 2, mix well (1:100)
ii. Transfer 15 µL from tube 2 to tube 3, mix well (1:1,000)
iii. Transfer 15 µL from tube 3 to tube 4, mix well (1:10,000)
iv. Transfer 15 µL from tube 4 to tube 5, mix well (1:100,000)
- At 4–5 hours post-seeding, aspirate medium from 24-well plate and replace with 450 μL of medium prepared in step a. Number wells 1–5 and a control.
- Transduce HT1080 cells by transferring 50 μL of dilution 1 to well 1 and mix well by pipetting.
NOTE: This step further dilutes your virus by 10-fold, thus well 1 now contains a 10
2 dilution.
- Repeat Step 5 for remaining prepared dilutions 2–5, transferring dilutions to corresponding numbered wells.
- For the control, add 50 μL of culture medium with Polybrene reagent (from step a) to control well.
- Gently swirl the plate to mix.
- Spin plate at 2,000 rpm at room temperature for 30 minutes.
- Incubate cell plate overnight.
Day 2
- Change medium in each well and replace with 500 μL of culture medium (without polybrene).
- Incubate cells for an additional 24 hours.
Blasticidin selection
- 48 hours post-transduction, remove medium and replace with selection medium (culture medium with Blasticidin S HCl at 10 μg/mL final concentration)
- Change and replace the selection medium every 2 days for 10 days.
NOTE: The control well should have no surviving cells prior to staining.
Day 12
- Remove medium and wash each well with 1 mL of PBS.
- Prepare a working solution of 1% crystal violet in a 10% solution of ethanol in water.
- Stain wells by adding 250 μL diluted crystal violet.
- Incubate plate for 20 minutes at room temperature.
- Remove and collect staining solution (staining solution can be reused, or appropriately discarded).
- Wash plate with water several times to reduce background.
Calculation of titer
To calculate the viral titer, count the number of colonies stained with crystal violet per well. Use the following formula to calculate the titer (TU/mL) of the viral stock:
TU/mL = (No. of discrete colonies x dilution factor)/volume of inoculum
Example:
Count of 35 plaques stained in the 1:100,000 dilution well.
Volume of diluted virus: 0.5 mL
Titer = (35 x 105)/0.5 = 7 x 106 TU/mL
Protocols and Citations Library
Search our extensive collection of FAQs to find the transfection answers you need. Input key terms to discover a collection of carefully curated and regularly updated answers.
Search the transfection FAQs database
Cell Culture & Transfection Application Notes