Search
Biotin quantitation kits are used for determining biotinylation levels and efficiency by either colorimetric or fluorescence detection methods. These kits contain the simple and accurate HABA dye used to measure the amount of biotin attached to proteins or other biomolecules. Individual fluorescent quantitation reagents are also available for detecting and quantitating biotin-binding sites. Now you can determine the molar ratio of biotin to a labeled antibody molecule in an easy, efficient manner.
Following biotin labeling, the HABA calculator can be used to determine the average number of biotin groups per protein molecule.
| Product | Features | Detection method | SKU |
|---|---|---|---|
| Fluorescence Biotin Quantitation Kit | Fluorescent assay reagent and HABA dye for rapid and sensitive determination of biotinylation levels in labeled antibodies and other proteins with minimal sample quantities | Fluorescent | 46610 |
| Pierce Biotin Quantitation Kit | A simple, colorimetric method for determining biotinylation efficiency | Colorimetric | 28005 |
| HABA | HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins | Colorimetric | 28010 |
| FluoReporter Biotin Quantitation Assay Kit | Fluorometric assay for accurately determining the number of biotin labels on a protein with higher sensitivity than the HABA biotin binding assay | Fluorescent | F30751 |
| Product | Features | Detection method | SKU |
|---|---|---|---|
| Fluorescence Biotin Quantitation Kit | Fluorescent assay reagent and HABA dye for rapid and sensitive determination of biotinylation levels in labeled antibodies and other proteins with minimal sample quantities | Fluorescent | 46610 |
| Pierce Biotin Quantitation Kit | A simple, colorimetric method for determining biotinylation efficiency | Colorimetric | 28005 |
| HABA | HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins | Colorimetric | 28010 |
| FluoReporter Biotin Quantitation Assay Kit | Fluorometric assay for accurately determining the number of biotin labels on a protein with higher sensitivity than the HABA biotin binding assay | Fluorescent | F30751 |
Everything you need to biotinylate, purify, or detect proteins
For Research Use Only. Not for use in diagnostic procedures.