Search
Immunology is considered a capstone area of biology, pulling from many fields such as molecular and cell biology, biochemistry, development, physiology, medicine, and pathology. There are many rich sub-disciplines within immunology, from the basic characterization of immune cells and the molecules they use to fight infections, to clinical immunology dealing with autoimmune diseases. An emerging area of medical research is immunotherapy, where immune system components are used to treat disease. The treatment of cancer using immunological approaches is known as immuno-oncology.
Signaling pathways are central to immunology. Some of the most famous signaling pathways are active in cells of the immune system, such as T-cell receptor, interferon-gamma, tumor necrosis factor, and complement signaling pathways. Measuring the expression of signaling pathway constituents is a key method for characterizing the response of immune cells to experimental treatments. Applied Biosystems TaqMan Gene Expression Assays enable researchers to reliably detect and quantitate the expression levels of common immunology biomarkers in addition to novel biomarkers identified in high-throughput transcriptomic studies that require verification.
Download a list of genes and corresponding assay IDs for common immunology biomarker targets for your research.
Search for a pre-designed TaqMan Assay for your immunology target using the tool below.
TaqMan Arrays are pre-spotted 96-well plates or 384-well microfluidic array cards, saving you the time of pipetting many assays into an empty plate. Just add your cDNA and master mix and you’re ready to go.
Custom TaqMan Arrays
These made-to-order cards and plates allow you to choose exactly which targets you want to spot on your panel - select from any of our >2.8 million predesigned TaqMan Gene Expression assays.
Configure Custom TaqMan Array Plates (96-well standard 0.2mL and fast 0.1mL)
Configure Custom TaqMan Array Cards (384-well)
Contact our Specialty TaqMan Plating team to inquire about Custom TaqMan 384-well plates.
Inventoried TaqMan Arrays
We also offer a variety of fixed-content TaqMan Array products focused on immune profiling and pathways. These products are readily available, with a minimum order quantity of 1 for plates and 4 for cards. The 96-well plates are available in both Standard 0.2-mL and Fast 0.1-mL formats.
| Description | Standard 0.2 mL | Fast 0.1 mL |
| Human Chemokines | 4414084 | 4418729 |
| Human Cholera Infection | 4414175 | 4418820 |
| Human CMV & MAPK Pathways | 4414169 | 4418814 |
| Human Complement Pathway | 4414120 | 4418765 |
| Human Cytokine Network | 4414124 | 4418769 |
| Human IGF1R Signaling | 4414141 | 4418786 |
| Human IL-1 Pathway | 4414142 | 4418787 |
| Human IL-10 Pathway | 4414143 | 4418788 |
| Human IL-2 Gene Expression in Activated and Quiescent T-Cells | 4414144 | 4418789 |
| Human IL-2 Pathway | 4414145 | 4418790 |
| Human IL-3 Pathway | 4414147 | 4418792 |
| Human IL-4 Pathway | 4414148 | 4418793 |
| Human IL-6 Pathway | 4414149 | 4418794 |
| Human IL-9 Pathway | 4414150 | 4418795 |
| Human ILK Signaling | 4414151 | 4418796 |
| Human Immune Response | 4414073 | 4418718 |
| Human Interferon Pathway | 4414154 | 4418799 |
| Human JAK-STAT Pathway | 4414156 | 4418801 |
| Human NFkB Pathway | 4414095 | 4418740 |
| Human Phagocytosis of Microbes | 4414178 | 4418823 |
| Human PI3K Signaling in B-lymphocytes | 4414173 | 4418818 |
| Human T-Cell Receptor and CD3 Complex | 4414186 | 4418831 |
| Human TGFB Pathway | 4414097 | 4418742 |
| Human TNF Superfamily Pathway | 4414189 | 4418834 |
| Human Toll Comparative Pathway | 4414192 | 4418837 |
| Human Toll-Like Receptors Pathway | 4414193 | 4418838 |
| Mouse Immune Response | 4414079 | 4418724 |
We also offer a variety of made-to-order, flexible content TaqMan Array products focused on immunological processes and pathways. Flexible panels start with curated gene content, with the ability to modify the configuration as needed. Swap out or remove assays and reconfigure the layout to fit your experiment design. They are available in 96-well array plates (minimum order of 6 plates), 384-well array cards (minimum order of 10 cards), and OpenArray format. Following is a selection of our more popular made-to-order arrays.
Learn more about Made-to-order Flexible Content panels ›
| Antibacterial Response | Human | Mouse | Rat |
| Autoimmune Diseases | Human | Mouse | Rat |
| Bacterial Infections | Human | Mouse | Rat |
| Cancer Inflammation & Immunity Crosstalk | Human | Mouse | Rat |
| Chemokines & Receptors | Human | Mouse | Rat |
| Cytokines | Human | Mouse | Rat |
| Cytokines & Chemokines | Human | Mouse | Rat |
| Cytokines lncRNA | Human | ||
| Dendritic & Antigen Presenting Cells | Human | Mouse | Rat |
| DNA Virus Infections | Human | Mouse | Rat |
| Flavivirus Infections | Human | Mouse | Rat |
| Gram-negative Bacterial Infections | Human | Mouse | Rat |
| Gram-positive Bacterial Infections | Human | Mouse | Rat |
| Helicobacter Infections | Rat | ||
| HIV Infections | Human | Mouse | Rat |
| Human HIV Infection and Host Response | Human | ||
| Immunotoxicity | Human | Mouse | Rat |
| Inflammatory Cytokines & Receptors | Human | Mouse | Rat |
| Inflammatory Response and Autoimmunity | Human | Mouse | Rat |
| Innate & Adaptive Immune Response lncRNA | Human | ||
| Innate and Adaptive Immune Responses | Human | Mouse | Rat |
| Lentivirus Infections | Human | Mouse | Rat |
| T Helper Cell Differentiation | Human | Mouse | Rat |
| TaqMan Immune Response | Human | ||
| T-cell and B-cell Activation | Human | Mouse | Rat |
| T-cell Anergy and Immune Tolerance | Human | Mouse | Rat |
| TGF-Beta Signaling Targets | Human | Mouse | Rat |
| Th17 for Autoimmunity & Inflammation | Human | Mouse | Rat |
| Th1-Th2-Th3 | Human | Mouse | Rat |
| TNF Signaling Pathway | Human | Mouse | Rat |
| Toll-Like Receptor Signaling Pathway | Human | Mouse | Rat |
| Tumor Virus Infections | Human | Mouse | Rat |
For Research Use Only. Not for use in diagnostic procedures.