Search
FastDigest Restriction EnzymesBrowse selection tool Download flyer |
Discover the efficiency and convenience of Thermo Scientific FastDigest restriction enzymes, designed to simplify your molecular biology workflows. FastDigest restriction enzymes help enable rapid DNA digestion in just 5–15 minutes, using a universal buffer system that helps ensure compatibility with all enzymes in the range. This helps streamline your processes, allowing for simultaneous digestion with multiple enzymes without the need for buffer changes. Explore our comprehensive selection of FastDigest restriction enzymes to help achieve accurate and reliable results in your cloning, sequencing, and other DNA manipulation experiments.
| FastDigest restriction enzymes | Conventional restriction enzymes | |
|---|---|---|
| Buffer system | One universal buffer | Up to 20 buffers |
| Double/multiple restriction digestion | Unlimited—all enzymes are 100% active in one buffer | Limited by buffer compatibility |
| Reaction time | 5–15 minutes | 1 hour–overnight |
| Direct loading on gels | Yes | No |
| Compatibility in downstream applications | 100% | Partial |
| Star activity | No | Yes |
| Activity definition | 1 µL of FastDigest enzyme cleaves 1 µg of substrate DNA in 5 to 15 minutes in FastDigest buffer | 1 unit of enzyme hydrolyzes 1 µg of lambda DNA in 60 minutes in an optimal buffer for an enzyme |
Reaction enzyme buffers help provide optimal conditions necessary for enzyme activity, including pH, ionic strength, and cofactors. They are crucial for precise DNA manipulation. A universal buffer for restriction enzymes is a significant advancement, simplifying experimental setups and reducing the need for multiple, specific buffers, thus helping save time and reducing errors.
FastDigest reaction buffer is a specially formulated to support the activity of multiple different restriction enzymes, helping them to function efficiently in the same reaction mixture. The Thermo Scientific FastDigest system uses a universal buffer that is compatible with all FastDigest enzymes, which helps eliminate the need for buffer changes when using multiple enzymes, streamlining the digestion process and helping reduce the potential for experimental error.
The FastDigest Green Buffer and Thermo Scientific FastDigest Buffer are proprietary digestion buffers which help support 100% activity of all FastDigest restriction enzymes. This system allows for double and multiple digestions with any combination of enzymes (Figure 1). No sequential digestions or buffer changes are needed.
Because FastDigest buffers help support 100% activity of FastDigest restriction enzymes and do not require sequential digestions or buffer changes, protocol time is reduced.
Digestion times for reaction conditions are provided for all types of DNA templates (plasmid DNA, PCR product, genomic DNA) and no overnight digestions are required for any template. In addition, star activity is eliminated due to short reaction times. See an example of reaction times for a double digestion in Table 1.
| Conventional restriction enzymes Sequential digestion | FastDigest restriction enzymes One reaction mixture | ||
|---|---|---|---|
| Reaction setup for Apal | ~2 min | Reaction set up with FastDigest ApaI and XhoI | ~2 min |
| Incubation | 60 min | Incubation | 5 min |
| Reaction setup for Xhol | ~2 min | — | — |
| Incubation | 60 min | — | — |
| Total time | >2 hours | Total time | 7 min |
DNA/RNA modifying enzymes, such as ligases, phosphatases, kinases, and mesophilic DNA polymerases have up to 100% activity in FastDigest Buffer and FastDigest Green Buffer (Table 2). Therefore, enzymes used in downstream applications can be directly added to the FastDigest reaction mix. No buffer changes or purification steps are needed.
| DNA/RNA modifying enzyme | Activity in FastDigest Green Buffer/FastDigest Buffer | |
|---|---|---|
DNA Polymerase I, E. coli | 100% | Order now |
Klenow Fragment | 100% | Order now |
Klenow Fragment, exo- | 100% | Order now |
T4 DNA Polymerase | 100% | Order now |
T7 DNA Polymerase | 100% | Order now |
T4 DNA Ligase* | 75–100% | Order now |
FastAP Thermosensitive Alkaline Phosphatase | 100% | Order now |
T4 Polynucleotide Kinase | 100% | Order now |
*0.5 mM ATP is required for T4 DNA Ligase activity
FastDigest Green Buffer offers the same performance as the colorless FastDigest Buffer, but FastDigest Green Buffer is ready for direct loading of the reaction mixture into gels. The 10X FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading. In a 1% agarose gel, the blue dye migrates with 3–5 kb DNA fragments and has an excitation peak of 424 nm, while the yellow dye migrates faster than 10 bp DNA fragments and has an excitation peak of 615 nm (Figure 2).
FastDigest restriction enzymes in combination with FastDigest Green Buffer are designed to eliminate star activity due to short incubation time and optimal enzyme and glycerol concentrations.
Conventional restriction enzymes may display star or "relaxed" activity due to prolonged incubation times, high enzyme and/or glycerol concentration, high pH values, or low ionic strength. By addressing all these issues, FastDigest enzymes enable single, double and even triple digestion in five minutes without any signs of star activity.
For Research Use Only. Not for use in diagnostic procedures.