Search
Standard kits include competent cells in a bulk format. Standard kits are an economical choice and they are designed for scientists who perform more than one transformation at a time and for general use in a busy lab. Kits are available in 500 μL (5 × 100 μL), 1 mL (5 × 200 μL), and 2 mL (4 × 500 μL) quantities. These kits include SOC medium and pUC19 positive control.
Standard kits include competent cells in a bulk format. Standard kits are an economical choice and they are designed for scientists who perform more than one transformation at a time and for general use in a busy lab. Kits are available in 500 μl (5 × 100 μl), 1 ml (5 × 200 μl), and 2 ml (4 × 500 μl) quantities. These kits include SOC medium and pUC19 positive control.
Standard kits include competent cells in a bulk format. Standard kits are an economical choice and they are designed for scientists who perform more than one transformation at a time and for general use in a busy lab. Kits are available in 500 μl (5 × 100 μl), 1 ml (5 × 200 μl), and 2 ml (4 × 500 μl) quantities. These kits include SOC medium and pUC19 positive control.
In addition to chemically competent cells and electrocompetent cells, we offer a broad suite of products for the cloning workflow.
Explore types of bacterial growth media to use for cloning, plasmid DNA preparation, and protein expression.
Learn more about universal-fit electroporation cuvettes for use with competent cells.
Learn more about one-step PCR cloning that combines traditional restriction and ligation cloning.
Gibson Assembly cloning kits provide highly efficient, seamless cloning, enabling the assembly of multiple DNA fragments of varying lengths into any vector.
Have any questions on competent cells or transformation? Click on the resources listed below to access overviews, videos, genotype guides, and educational resources.
Review rich educational content on transformation workflow, troubleshooting, strategies, and selection guides.
Stay up-to-date on industry trends in molecular cloning with on-demand webinars.
Access white papers and technical and application notes on molecular cloning.
Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to molecular cloning.