Search
This resource is designed to help researchers enhance their genome editing techniques in various cell models that closely mimic physiological conditions. Find recommendations and detailed protocols designed to help optimize your transfection and genome editing processes. Whether you are working with CRISPR, TALENs, or other genome editing technologies, learn about achieving precise and efficient genetic modification to help improve your research in understanding gene functions and developing novel therapeutics.
Genome editing is an advanced technique that allows scientists to make targeted modifications to the DNA of living organisms. This technology is important because it enables researchers to investigate the functions of specific genes, understand the genetic basis of diseases, and develop new treatments. By accurately altering genetic sequences, genome editing can correct mutations that cause genetic disorders, create disease-resistant crops, and even potentially treat inherited diseases. Techniques such as CRISPR-Cas9 have significantly impacted the field by providing a highly efficient and accessible method for editing the genome, paving the way for notable advancements in medicine, agriculture, and biotechnology.
With increasing expansion into research areas of more biological relevance, existing molecular and cellular techniques need to be improved. Invitrogen Lipofectamine 3000 Transfection Reagent, a new reagent developed to improve delivery and enable use of new technologies, can be used in more relevant systems enabling faster and more reliable outcomes. The area of genome editing is rapidly growing and requires more advanced techniques to maximize its potential applications. Transcriptional activator-like effector nucleases (TALENs) and technology derived from clustered regularly interspaced short palindromic repeats (CRISPRs) allow precise cleavage of DNA at specific loci. However, the effectiveness of these tools is contingent upon the intrinsic properties of the locus of interest, efficient delivery, and the painstaking downstream processes of generating stable cell lines and knockout models to study the phenotypic effects of such genetic modifications. During the development of Lipofectamine 3000 reagent, we assessed transfection of HepG2 and U2OS cell lines with TALEN and CRISPR vectors designed using Invitrogen GeneArt Gene Synthesis services to target a specific locus. We observed improvements in transfection efficiency, mean fluorescence intensity, and genomic cleavage. These advancements in delivery help minimize painstaking downstream workflows, enable easier stem cell manipulation, and enhance site-specific insertion of transgenes into the cellular genome.
Invitrogen GeneArt Precision TALs and GeneArt CRISPR Nuclease Vectors were designed using the GeneArt web design tool (thermofisher.com/tals). The forward and reverse TALENs contain the FokI nuclease and target the AAVS1 safe harbor locus. The all-in-one CRISPR vector system contains a Cas9 nuclease expression cassette and a guide RNA cloning cassette that target the AAVS1 safe harbor locus, combined with a downstream orange fluorescent protein (OFP) reporter. Invitrogen pcDNA3.3 vector was also used as a negative control throughout the assay. The plasmids were transformed into competent E. coli cells. Clones were analyzed and sequenced for specificity and then purified using an Invitrogen PureLink HiPure Plasmid Filter Maxiprep Kit to ensure low endotoxin activity and high-quality DNA.
Cells were cultured using Gibco DMEM, high-glucose, with Gibco GlutaMAX Supplement and 10% fetal bovine serum for 4–5 passages after thawing; cells were dissociated using Gibco TrypLE Express Enzyme and seeded in a 12-well plate at 2 x 10e5 cells per well in 1 mL complete medium to ensure 70–90% confluence on the day of transfection.
Transfection with Lipofectamine 3000 reagent and Invitrogen Lipofectamine 2000 Transfection Reagent was compared. For transfection with Lipofectamine 3000 reagent (Figure 1), in separate tubes, 1.5 µL of Lipofectamine 3000 reagent and 1 µg of DNA were each diluted in 50 µL Gibco Opti-MEM Reduced-Serum Medium; then 2 µL P3000 reagent was added to the diluted DNA. The diluted DNA with P3000 reagent was added to the diluted Lipofectamine 3000 reagent and incubated at room temperature for 5 minutes. Then 100 µL of the resulting complex was added to cells in complete medium. The procedure was the same for Lipofectamine 2000 reagent, except that the amount of transfection reagent was increased to 3 µL and no additional reagent was added to the diluted DNA before adding it to the diluted Lipofectamine 2000 reagent. All downstream analysis was performed 72 hours post-transfection.
OFP expression from the CRISPR vector was determined by flow cytometry and microscopy. An Invitrogen EVOS FL Imaging System was used to acquire images with the RFP filter. Cells were then dissociated 72 hours posttransfection with TrypLE Express Enzyme and analyzed using a BD Accuri C6 Flow Cytometer with an FL-2 filter and blue laser.
Figure 1. Lipofectamine 3000 reagent transfection workflow.
The Invitrogen GeneArt Genomic Cleavage Detection Kit provides a reliable and rapid method for the detection of locus-specific cleavage (Figure 2). Cells were dissociated with TrypLE Express Enzyme, washed with Dulbecco’s phosphate-buffered saline, and pelleted. Cells were lysed with the Cell Lysis Buffer and Protein Degrader Mix from the Invitrogen GeneArt Genomic Cleavage Detection Kit. The DNA was extracted and then PCR-amplified with forward and reverse primers. Denaturing and reannealing reactions were then performed to randomly anneal the mutated and unmutated PCR fragments. Detection enzyme (1 µL) was added, the mix was incubated for 1 hour at 37°C, and then the entire mix was electrophoresed in an Invitrogen E-Gel EX 2% agarose gel to determine the percent genome modification. AlphaView™ Software was used to determine cleavage efficiency using the formula: cleavage efficiency = 1 – [(1 – fraction cleaved)1/2]
U2OS cells, derived from human bone osteosarcoma, and HepG2 cells, derived from a human hepatocellular carcinoma, were transfected with Lipofectamine 3000 reagent. Both cell lines showed improved transfection efficiency and protein expression compared to transfection mediated with Lipofectamine 2000 reagent. Transfection efficiency and protein expression were assessed using a CRISPR construct that contains the OFP reporter gene. U2OS cells transfected with Lipofectamine 3000 reagent had 2-fold improved transfection efficiency (data not shown) and 4-fold improved fluorescence intensity (Figure 3A). HepG2 cells showed 20-fold improvement in transfection efficiency (data not shown) and 80-fold higher fluorescence intensity (Figure 3B). Most importantly, we observed increased TALEN- and CRISPR-mediated cleavage for the AAVS1 target locus in both cell lines transfected with Lipofectamine 3000 reagent, demonstrating that increasing the transfection efficiency and, by implication, protein expression, will increase the cleavage rate of TALENs and CRISPR nucleases. U2OS cells transfected with Lipofectamine 3000 reagent showed 1.5-fold improved TALEN cleavage efficiency and slightly improved CRISPR cleavage (Figure 4A). HepG2 cells had 3-fold higher TALEN cleavage efficiency and 8-fold higher CRISPR cleavage (Figure 4B).
Figure 2. GeneArt Genomic Cleavage Detection Kit workflow.
Figure 3. Transfection efficiency and protein expression using a CRISPR vector. The vector contained an OFP reporter gene and was transfected with Lipofectamine 2000 or Lipofectamine 3000 reagent into (A) U2OS and (B) HepG2 cell lines. Bar graphs show reporter gene expression; images show fluorescence of corresponding cells expressing OFP.
Figure 4. Cleavage efficiency of GeneArt TALENs and CRISPR nucleases. The TALENs and CRISPR nucleases targeted the AAVS1 locus in (A) U2OS and (B) HepG2 cell lines. Cleavage was assayed using a GeneArt Genomic Cleavage Detection Kit.
Lipofectamine 3000 reagent was developed to improve transfection efficiency in difficult-to-transfect cell lines and to minimize painstaking downstream processes involved in genetic engineering protocols. In this study, we demonstrated that using a better transfection reagent such as Lipofectamine 3000 reagent will improve the cleavage efficiency with TALENs or CRISPR nucleases, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes. Clonal selection is one example of a protocol that would benefit from an improved transfection reagent. Therefore, this study confirms the hypothesis that improving transfection will improve cleavage and recombination efficiency when using genome editing tools such as GeneArt Precision TALs and CRISPR nucleases.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Cell Culture Selection Tools
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support?
Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.