Search Thermo Fisher Scientific
Functional analysis of regulatory RNAs like microRNA (miRNA) can help decipher complex cellular processes in development and disease etiology.
Our suite of synthetic miRNA mimics and inhibitors are:
We recommend mirVana mimics and inhibitors for anyone getting started with functional analysis of endogenous microRNA and looking for the latest advances in specificity and potency. Our Pre-miR and Anti-miR reagents remain relevant for ongoing studies that have benefited from these original product formats.
| miRNA mimics for gain-of-function studies | miRNA inhibitors for loss-of-function studies | |||
|---|---|---|---|---|
| mirVana Mimics RECOMMENDED | Pre-miR Mimics | mirVana Inhibitors RECOMMENDED | Anti-miR Inhibitors | |
| Content database coverage | miRBase v.22 | miRBase v.15 | miRBase v.22 | miRBase v.15 |
| Available formats | Individual tubes, custom plated collections, or whole genome library | Individual tubes | Individual tubes, custom plated collections, or whole genome library | Individual tubes |
| Product summary |
|
|
|
|
mirVana miRNA mimics are chemically modified double-stranded RNA molecules designed to mimic endogenous microRNAs (miRNAs), resulting in down-regulation of target mRNA translation due to mRNA sequestration or degradation. Like native miRNAs, these mimics have two strands—the mature, or guide strand, which is functional and used by the Argonaute (Ago) protein to target mRNAs, and the passenger strand (formerly called the star strand), which may or may not have native targets of its own.
The chemical modifications in mirVana miRNA mimics inactivate the passenger strand to help ensure that the guide strand (representing the desired mature miRNA) is taken up by Ago to produce the miRNA effect (Figure 1). mirVana miRNA mimics are designed to produce maximum and consistent effects at concentrations as low as 0.3 nM and are available individually or as libraries.
Figure 1. mirVana miRNA mimics demonstrate high specificity. HeLa cells were transfected with one of six different mirVana miRNA mimics at 3 nM concentration and a corresponding reporter plasmid. The fold change in reporter gene expression for each miRNA mimic strand was determined by measuring expression in the presence of mimic relative to that of a negative control (set at 1.0). Each plasmid has the reporter gene cloned both in forward orientation (Fwd) to measure activity of the miRNA mimic mature strand, and in reverse/complement orientation (Rev) to test activity of the passenger strand. For all six mimics, mature strand activity is high (reporter gene expression reduced 5- to 10-fold compared with negative control), and passenger strand activity is low or absent (similar to negative control).
mirVana miRNA inhibitors are chemically modified, single-stranded oligonucleotides with patented secondary structure designed to specifically bind to and inhibit endogenous miRNAs, resulting in up-regulation of target mRNA translation. Compared with competitors, these inhibitors have the highest-potency in vitro inhibition at the lowest miRNA inhibitor concentration (Figure 2). Like the mimics, the mirVana miRNA inhibitors are available individually or as libraries.
Figure 2. mirVana miRNA inhibitors are more potent than those from leading competitors. mirVana miRNA inhibitors and two other commercially available let-7 inhibitors were each transfected into HeLa cells at 10 nM concentration using Lipofectamine RNAiMAX reagent. Twenty-four hours later, HMGA2 mRNA levels (a natural target of the let-7 microRNA) were measured by qRT-PCR using TaqMan assays.
Figure 3. mirVana miRNA inhibitors effectively suppress miRNA in vivo. miR-122 or Negative Control #1 mirVana miRNA inhibitors were complexed with Invivofectamine 2.0 reagent and injected into the tail veins of Balb-C mice on three consecutive days at 7 mg per kg body weight. Twenty-four hours after the last injection, expression levels of four natural targets of miR-122 were measured in the liver using TaqMan assays. Significant up-regulation of all four mRNAs was detected in mice treated with miR-122 inhibitor, as compared with mice that received no treatment or the negative control. Results indicate that mirVana miRNA inhibitors were efficiently delivered to the liver with Invivofectamine 2.0, where they inactivated miR-122, leading to up-regulation of genes naturally suppressed by miR-122.
In addition to our pre-defined miRNA libraries, mirVana mimics and inhibitors are available as custom collections.
| Product line | Quantity per well |
|---|---|
| mirVana miRNA mimics–human, mouse, or rat | 0.25 or 1.0 nmol/well |
| mirVana miRNA inhibitors–human, mouse, or rat | 0.25 or 1.0 nmol/well |
mirVana miRNA mimics are chemically modified double-stranded RNA molecules designed to mimic endogenous microRNAs (miRNAs), resulting in down-regulation of target mRNA translation due to mRNA sequestration or degradation. Like native miRNAs, these mimics have two strands—the mature, or guide strand, which is functional and used by the Argonaute (Ago) protein to target mRNAs, and the passenger strand (formerly called the star strand), which may or may not have native targets of its own.
The chemical modifications in mirVana miRNA mimics inactivate the passenger strand to help ensure that the guide strand (representing the desired mature miRNA) is taken up by Ago to produce the miRNA effect (Figure 1). mirVana miRNA mimics are designed to produce maximum and consistent effects at concentrations as low as 0.3 nM and are available individually or as libraries.
Figure 1. mirVana miRNA mimics demonstrate high specificity. HeLa cells were transfected with one of six different mirVana miRNA mimics at 3 nM concentration and a corresponding reporter plasmid. The fold change in reporter gene expression for each miRNA mimic strand was determined by measuring expression in the presence of mimic relative to that of a negative control (set at 1.0). Each plasmid has the reporter gene cloned both in forward orientation (Fwd) to measure activity of the miRNA mimic mature strand, and in reverse/complement orientation (Rev) to test activity of the passenger strand. For all six mimics, mature strand activity is high (reporter gene expression reduced 5- to 10-fold compared with negative control), and passenger strand activity is low or absent (similar to negative control).
mirVana miRNA inhibitors are chemically modified, single-stranded oligonucleotides with patented secondary structure designed to specifically bind to and inhibit endogenous miRNAs, resulting in up-regulation of target mRNA translation. Compared with competitors, these inhibitors have the highest-potency in vitro inhibition at the lowest miRNA inhibitor concentration (Figure 2). Like the mimics, the mirVana miRNA inhibitors are available individually or as libraries.
Figure 2. mirVana miRNA inhibitors are more potent than those from leading competitors. mirVana miRNA inhibitors and two other commercially available let-7 inhibitors were each transfected into HeLa cells at 10 nM concentration using Lipofectamine RNAiMAX reagent. Twenty-four hours later, HMGA2 mRNA levels (a natural target of the let-7 microRNA) were measured by qRT-PCR using TaqMan assays.
Figure 3. mirVana miRNA inhibitors effectively suppress miRNA in vivo. miR-122 or Negative Control #1 mirVana miRNA inhibitors were complexed with Invivofectamine 2.0 reagent and injected into the tail veins of Balb-C mice on three consecutive days at 7 mg per kg body weight. Twenty-four hours after the last injection, expression levels of four natural targets of miR-122 were measured in the liver using TaqMan assays. Significant up-regulation of all four mRNAs was detected in mice treated with miR-122 inhibitor, as compared with mice that received no treatment or the negative control. Results indicate that mirVana miRNA inhibitors were efficiently delivered to the liver with Invivofectamine 2.0, where they inactivated miR-122, leading to up-regulation of genes naturally suppressed by miR-122.
In addition to our pre-defined miRNA libraries, mirVana mimics and inhibitors are available as custom collections.
| Product line | Quantity per well |
|---|---|
| mirVana miRNA mimics–human, mouse, or rat | 0.25 or 1.0 nmol/well |
| mirVana miRNA inhibitors–human, mouse, or rat | 0.25 or 1.0 nmol/well |
miRNAs require special tools for accurate and sensitive analysis. We offer a broad portfolio of products that provides complete solutions for either miRNA expression profiling or a targeted quantitation/target validation.
| miRNA custom synthesis and libraries | miRNA and small RNA extraction | miRNA analysis solutions | miRNA and non-coding RNA microarrays |
|---|---|---|---|
We've made it easier for you to quickly find and buy products for every step in your transfection workflow.
Ensure effective delivery, detection, and interpretation of your miRNA modulation experiments
Learn more about native regulatory RNAs
If you have any questions on miRNA products or ordering, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.