Hypersil GOLD HPLC and UHPLC Columns – Designed for optimum chromatography
Consistency you can rely on, results you can trust
Enjoy the range and capabilities of these state-of-the-art columns and gain confidence in the accuracy and quality of your analytical data. The Thermo Scientific Hypersil GOLD family of liquid chromatography (LC) columns is the culmination of 40 years of experience in the media and column product development and manufacturing.
Contact us
Peak symmetry
Hypersil GOLD columns produce highly symmetrical peaks, which facilitate the detection of low-level impurities that elute immediately after the main component. When there is even minor tailing, small impurity peaks can elute on the shoulder of the main peak, complicating the integration process.
Hypersil GOLD column phase properties
PhasePhase Properties
Hypersil GOLD Peptide HPLC Columns 
Consistent retention of all peptides, including   hydrophilic ones, and resolve deamidated species
Hypersil GOLD C18 Selectivity LC ColumnsExcellent peak shape for all analyte types.
Hypersil GOLD C8 LC ColumnsSimilar selectivity to C18 columns, but with reduced retention.
Hypersil GOLD C4 LC ColumnsSimilar selectivity to C18 and C8 columns, but with reduced retention.
Hypersil GOLD aQ LC ColumnsPolar endcapped C18 columns, stable in 100% aqueous mobile phase, providing enhanced retention and resolution of polar analytes.
Hypersil GOLD PFP LC ColumnsPerfluorinated phenyl columns offering alternative selectivity. The fluorine atoms around the phenyl ring enhance pi-pi interactions with aromatic molecules.
Hypersil GOLD Phenyl LC ColumnsExcellent retention and unique selectivity for aromatic analytes.
Hypersil GOLD Cyano LC ColumnsAlternative selectivity with lower hydrophobicity; can also be used for normal phase separations.
Hypersil GOLD Amino LC ColumnsCan be used in reversed phase, normal phase, ion exchange, and HILIC modes; particularly useful for separating carbohydrates.
Hypersil GOLD AX LC ColumnsSeparation of smaller proteins, peptides, anionic species, and polar molecules.
Hypersil GOLD SAX LC ColumnsA quaternary amino ion exchange ligand ideally suited to separating small polar organic analytes in aqueous mobile phases.
Hypersil GOLD Silica LC ColumnsSeparation of nonpolar and moderately polar organic compounds by normal phase chromatography.
Hypersil GOLD PEI HILIC LC ColumnsEnhanced retention of polar and hydrophilic analytes that are problematic using reversed phase columns.
Important product information
Resources
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.
The Thermo Scientific Orbitrap Exploris and Excedion series mass spectrometers are Orbitrap instruments with an atmospheric pressure ionization (API) source for liquid chromatography (LC) mass spectrometry (MS) high-throughput applications. The instruments are designed to be placed on a bench in the laboratory and maintain mass accuracy across chromatographic peaks, supporting confident identification and quantitation using high-resolution accurate-mass detection.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs