Tailored for challenging and critically important applications
The novel and unique chemistries of these Thermo Scientific Acclaim columns provide superior resolution with ease of use for key pharmaceutical and environmental applications.
Column | Applications |
---|---|
Acclaim Carbamate LC Columns | Achieve baseline separation of carbamate specified in U.S. EPA Method 531.2 with these high-efficiency columns. |
Acclaim Carbonyl C18 LC Columns | Use for superior resolution and high throughput separation of DNPH derivatives of aldehydes and ketones. |
Acclaim Explosives E2 LC Columns | Achieve baseline resolution of all 14 explosives listed in EPA Method. |
Acclaim Organic Acid LC Columns | Use these reversed phase silica columns for unparalleled performance in separating hydrophilic, aliphatic, and aromatic organic acids. |
Acclaim Surfactant LC Columns | Use these high efficiency columns to achieve outstanding separation of all classes of surfactants. |
Acclaim Surfactant Plus LC Columns | Choose this column for surfactant analysis using high sensitivity detection. |
Acclaim Trinity P1 LC Columns | Separate pharmaceutical drug substances and their counter ions simultaneously and with high efficiency. |
Acclaim Trinity P2 LC Columns | Separate pharmaceutical counter ions, including monovalent and divalent cations or anions. |
Acclaim Trinity Q1 LC | Separate the herbicides diquat and paraquat simultaneously. |
Hypersil Green PAH LC Columns | Use these columns for rapid analysis of polyaromatic hydrocarbons (PAHs). |
Acclaim AmG C18 LC Columns | Meet today’s demands for aminoglycoside separation with these columns which have unique tolerance to harsh conditions. |
- High-performance, silica-based C18 column specifically designed for ion pairing, reversed phase LC analysis
- Unique column chemistry has unusual tolerance toward low pH, high temperature, organic solvents, and aqueous mobile phases
- Detection by UV, CAD, or or ELSD
- Aminoglycoside
- Carbamate
- Explosives
- DNPH derivatives of aldehydes and ketones
- Hydrophilic, aliphatic, aromatic organic acids
- Surfactants
- Drugs and counter ions
- Herbicides diquat and paraquat
- Polyaromatic hydrocarbons (PAHs)
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Access the latest version of our Chromatography Consumables Catalog to ensure that you're optimizing your applications! Our enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Discover live and on-demand webinars that enable you to improve productivity, maintain compliance, and learn about the constantly evolving scientific landscape.