Search
Invitrogen Zero Blunt TOPO technology provides a highly efficient, 5-minute, one-step cloning strategy for the direct insertion of blunt-ended PCR products amplified with proofreading thermostable polymerases into a plasmid vector. Each kit has the Zero Blunt TOPO vector (see figure) containing the ccdB gene for positive selection, only permitting growth of plasmid vectors with recombinants so you can obtain up to 95% clones with correct insert.
The pCR Blunt II-TOPO vector is designed to clone blunt-ended PCR products generated by thermostable proofreading polymerases such as Platinum SuperFi DNA Polymerase.
Each Zero Blunt TOPO for subcloning Kit contains linearized and topoisomerase I-activated pCR Blunt II-TOPOVector, Salt Solution, dNTPs, Control Template and Primers, M13 Forward and Reverse Primers, One Shot Chemically Competent or Electrocomp E. coli, S.O.C. Medium, and a supercoiled control plasmid. The Zero Blunt TOPO PCR Cloning Kit for Subcloning, without competent cells allows the flexibility of using your own strain for increased flexibility so competent cells and S.O.C. Medium are not provided.
In addition, a Zero Blunt TOPO Cloning Kit is available combined with a Invitrogen PureLink Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO-cloned inserts for downstream analysis.
| Includes One Shot Competent E. coli | Advantage | Rxn number | Cat. No. | |
|---|---|---|---|---|
| None |
| 10 | 451245 | Order now |
| TOP10 (genotype) |
| 10 | K2800J10 | Order now |
| None |
| 25 | 450245 | Order now |
| TOP10 (genotype) |
| 25 | K280020 | Order now |
| MAX Efficiency DH5 αT1 Phage Resistant (genotype) |
| 25 | K282020 | Order now |
| TOP10 Invitrogen Electrocomp(genotype) |
| 25 | K286020 | Order now |
| Invitrogen Mach1 T1R (genotype) |
| 25 | K283020 | Order now |
| TOP10 (genotype) |
| 25 | K280002 | Order now |
| TOP10 (genotype) |
| 50 | K280040 | Order now |
| MAX Efficiency DH5 α T1 Phage Resistant (genotype) |
| 50 | K282040 | Order now |
| TOP10 Electrocomp(genotype) |
| 50 | K286040 | Order now |
For Research Use Only. Not for use in diagnostic procedures.