Search
Air-drying and Lyophilization for Assay |
Stabilizing diagnostic assays for reliable room-temperature storage and transport is critical for assay developers. Thermo Fisher Scientific offers the lyo-ready portfolio of glycerol-free enzymes and reagents optimized for lyophilization for excellent stability and cost-effective logistics. Additionally, our newest Invitrogen Dry-Ready Reagents simplify assay stabilization through a fast, affordable air-drying process that requires no specialized equipment, featuring a verified excipient mix to help ensure consistent results and accelerate assay development.
| Air-drying | Lyophilization | |
|---|---|---|
| Process: ~2 hours | Process: >24 hours | |
| Equipment: Heating oven | Equipment: Lyophilizator | |
| Product shipment and storage at room temperature | Product shipment and storage at room temperature | |
| Excipients and manual included | Lyophilization service provider recommendation |
The Invitrogen Dry-Ready RT-qPCR Kit offers an innovative solution for preparing hot air-dried RT-qPCR assays. Leveraging advanced Invitrogen reagents, this simple and rapid air-drying method outperforms traditional lyophilization in speed and ease, while enabling stable room-temperature storage and shipment of assays.
| Product | Components |
|---|---|
| Dry-Ready RT-qPCR Kit | Dry-Ready Platinum II DNA Polymerase |
| Dry-Ready SuperScript III Flash Reverse Transcriptase | |
| Dry-Ready RNaseOUT Recombinant Ribonuclease Inhibitor | |
| Dry-Ready dNTP Mix | |
| 10X Dry-Ready Reaction Buffer | |
| MgCl2 |
The Dry-Ready RT-LAMP Kit is a comprehensive solution for air-drying RT-LAMP assays. Optimized for a simple drying process that helps preserve assay functionality, it offers flexibility in initial sample volume and enhances sensitivity—making it an exceptional choice for robust stabilization in point-of-care applications.
| Product | Components |
|---|---|
| Dry-Ready RT-LAMP Kit | Dry-Ready SuperScript IV Reverse Transcriptase |
| Dry-Ready Bst DNA Polymerase | |
| Dry-Ready dNTP Mix | |
| 10X Dry-Ready RT-LAMP Reaction Buffer |
Stability testing of the Dry-Ready RT-qPCR Kit demonstrated that air-dried assays retained full activity after 10 weeks of storage at 50°C using the XENO RNA template. Across four input copy numbers (2,000; 10,000; 50,000; and 200,000), the air-dried assay (yellow) showed no loss of activity or sensitivity compared to freshly prepared assays (blue), confirming its robustness and reliability for long-term storage at elevated temperatures.
Figure 2. Stability of Dry-Ready RT-qPCR Kit after air drying.
The Dry-Ready Excipient Mix stabilizes air-dried assays containing Dry-Ready SuperScript III Reverse Transcriptase and Dry-Ready Platinum Taq DNA Polymerase for at least 10 weeks at 50°C, as demonstrated with the XENO RNA template. Tested at two input levels (2,000 and 200,000 copies), the air-dried assay (yellow) showed no loss of activity or sensitivity compared to freshly prepared assays (blue), confirming its stability and reliability for long-term storage at elevated temperatures.
Dry-Ready Excipient is a customized product for OEM use. For custom and commercial supply needs, please contact us at thermofisher.com/MDxContact.
Stability data for the Dry-Ready RT-LAMP Kit show that air-dried assays retained full activity after 14 weeks of storage at 37°C as demonstrated with human Adenovirus41 DNA template. The air-dried assays (red) demonstrated no loss of activity or sensitivity compared to freshly prepared assays (blue), confirming their stability and reliability for long-term storage at elevated temperatures.
Figure 4. Stability of Dry-Ready RT-LAMP Kit after air drying.
The Invitrogen Dry-Ready RT-qPCR Kit offers an innovative solution for preparing hot air-dried RT-qPCR assays. Leveraging advanced Invitrogen reagents, this simple and rapid air-drying method outperforms traditional lyophilization in speed and ease, while enabling stable room-temperature storage and shipment of assays.
| Product | Components |
|---|---|
| Dry-Ready RT-qPCR Kit | Dry-Ready Platinum II DNA Polymerase |
| Dry-Ready SuperScript III Flash Reverse Transcriptase | |
| Dry-Ready RNaseOUT Recombinant Ribonuclease Inhibitor | |
| Dry-Ready dNTP Mix | |
| 10X Dry-Ready Reaction Buffer | |
| MgCl2 |
The Dry-Ready RT-LAMP Kit is a comprehensive solution for air-drying RT-LAMP assays. Optimized for a simple drying process that helps preserve assay functionality, it offers flexibility in initial sample volume and enhances sensitivity—making it an exceptional choice for robust stabilization in point-of-care applications.
| Product | Components |
|---|---|
| Dry-Ready RT-LAMP Kit | Dry-Ready SuperScript IV Reverse Transcriptase |
| Dry-Ready Bst DNA Polymerase | |
| Dry-Ready dNTP Mix | |
| 10X Dry-Ready RT-LAMP Reaction Buffer |
Stability testing of the Dry-Ready RT-qPCR Kit demonstrated that air-dried assays retained full activity after 10 weeks of storage at 50°C using the XENO RNA template. Across four input copy numbers (2,000; 10,000; 50,000; and 200,000), the air-dried assay (yellow) showed no loss of activity or sensitivity compared to freshly prepared assays (blue), confirming its robustness and reliability for long-term storage at elevated temperatures.
Figure 2. Stability of Dry-Ready RT-qPCR Kit after air drying.
The Dry-Ready Excipient Mix stabilizes air-dried assays containing Dry-Ready SuperScript III Reverse Transcriptase and Dry-Ready Platinum Taq DNA Polymerase for at least 10 weeks at 50°C, as demonstrated with the XENO RNA template. Tested at two input levels (2,000 and 200,000 copies), the air-dried assay (yellow) showed no loss of activity or sensitivity compared to freshly prepared assays (blue), confirming its stability and reliability for long-term storage at elevated temperatures.
Dry-Ready Excipient is a customized product for OEM use. For custom and commercial supply needs, please contact us at thermofisher.com/MDxContact.
Stability data for the Dry-Ready RT-LAMP Kit show that air-dried assays retained full activity after 14 weeks of storage at 37°C as demonstrated with human Adenovirus41 DNA template. The air-dried assays (red) demonstrated no loss of activity or sensitivity compared to freshly prepared assays (blue), confirming their stability and reliability for long-term storage at elevated temperatures.
Figure 4. Stability of Dry-Ready RT-LAMP Kit after air drying.
Lyo-ready enzymes hold promise and potential for faster and accurate point of care diagnostics. According to an article published in Nature , these 'lyo-ready' enzymes can be used to build dried mixtures of PCR reagents that enable sample-to-answer assays. Thermo Fisher Scientific offers customized solutions for specific applications, including modified concentrations, volume, and packaging.
Explore lyo-ready enzymes for reverse transcription, isothermal amplification, PCR amplification, and other related technologies.
| Product | Applicable technology |
|---|---|
| Lyo-ready SuperScript III Flash RT | 1-step RT-qPCR |
| Invitrogen Lyo-ready SuperScript Reverse Transcriptase, 1-step RT-qPCR | 1-step RT-qPCR |
| Invitrogen SuperScript III RT | 1-step RT-qPCR |
| Invitrogen SuperScript IV RT | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis |
| Thermo Scientific Maxima H Minus RT | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis |
| Thermo Scientific Maxima RT | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis |
| Thermo Scientific M-MLV RT | 1-step RT-qPCR |
| Thermo Scientific RevertAid H Minus RT | 2-step RT-qPCR, 1-step RT-PCR, RT-LAMP, cDNA synthesis |
| Thermo Scientific RevertAid M-MLV RT | 1-step RT-qPCR |
| Product | Applicable technology |
|---|---|
| RPA proteins and reagents | RPA |
| Invitrogen Bst DNA Polymerase | LAMP, HAD, EXPAR, RCA, RPA |
| Thermo Scientific EquiPhi29 DNA Polymerase | MDA, WGA, RCA |
| Product | Applicable technology |
|---|---|
| Invitrogen Platinum II Taq DNA polymerase | 1-step RT-qPCR |
| Invitrogen Platinum Taq DNA Polymerase | 1-step RT-qPCR |
| Applied Biosystems AmpliTaq Gold DNA Polymerase | 1-step RT-qPCR |
| Invitrogen LibertyTaq DNA Polymerase | 1-step RT-qPCR |
| Thermo Scientific Phire Hot Start II DNA Polymerase | PCR |
| Product | Applicable technology |
|---|---|
| Invitrogen RNaseOUT RNase inhibitor | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis, NASBA, TMA |
| Thermo Scientific RiboLock RNase Inhibitor | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis, NASBA, TMA |
| Product | Applicable technology |
|---|---|
| Lyo-ready SuperScript III Flash RT | 1-step RT-qPCR |
| Invitrogen Lyo-ready SuperScript Reverse Transcriptase, 1-step RT-qPCR | 1-step RT-qPCR |
| Invitrogen SuperScript III RT | 1-step RT-qPCR |
| Invitrogen SuperScript IV RT | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis |
| Thermo Scientific Maxima H Minus RT | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis |
| Thermo Scientific Maxima RT | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis |
| Thermo Scientific M-MLV RT | 1-step RT-qPCR |
| Thermo Scientific RevertAid H Minus RT | 2-step RT-qPCR, 1-step RT-PCR, RT-LAMP, cDNA synthesis |
| Thermo Scientific RevertAid M-MLV RT | 1-step RT-qPCR |
| Product | Applicable technology |
|---|---|
| RPA proteins and reagents | RPA |
| Invitrogen Bst DNA Polymerase | LAMP, HAD, EXPAR, RCA, RPA |
| Thermo Scientific EquiPhi29 DNA Polymerase | MDA, WGA, RCA |
| Product | Applicable technology |
|---|---|
| Invitrogen Platinum II Taq DNA polymerase | 1-step RT-qPCR |
| Invitrogen Platinum Taq DNA Polymerase | 1-step RT-qPCR |
| Applied Biosystems AmpliTaq Gold DNA Polymerase | 1-step RT-qPCR |
| Invitrogen LibertyTaq DNA Polymerase | 1-step RT-qPCR |
| Thermo Scientific Phire Hot Start II DNA Polymerase | PCR |
| Product | Applicable technology |
|---|---|
| Invitrogen RNaseOUT RNase inhibitor | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis, NASBA, TMA |
| Thermo Scientific RiboLock RNase Inhibitor | 2-step RT-qPCR, 1-step RT-PCR, LAMP, cDNA synthesis, NASBA, TMA |
Check out our resources on molecular assay development.
Access technical and application notes on molecular cloning.
Stay up-to-date on industry trends in molecular diagnostics development with on-demand webinars from industry experts.
Review content on workflow, troubleshooting, strategies, and selection guides.
Get support for your molecular assay development. For information on a personalized IVD transition plan contact us at ivdrtransition@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.