Stack of TaqMan array cards
Experimental throughput in research laboratories continues to increase, and so does the need for simple and efficient ways to run tests. Researchers are often left to choose between cumbersome, time-consuming manual approaches and costly automation. The Applied Biosystems TaqMan array cards for gene expression provide quality performance of probe-based TaqMan assays in an easy and efficient mid- to high-throughput format. TaqMan array cards for gene expression provide an alternative that enables highly reproducible and sensitive results with higher throughput, without the expense of liquid-handling robotics or error potential of excessive pipetting.
Array card formats and applications
While used extensively in gene expression applications, TaqMan Array cards are also available for additional applications as shown here:
  Applications
Gene Expression
Advanced miRNA
miRNA
SNP genotyping
Copy number
Mutation detection
FormatsInventoried✓✓✓   
Flexible, made to order✓     
Custom✓✓✓   
Specialty   ✓✓✓
Predesigned TaqMan Array cards
Thermo Fisher Scientific’s preconfigured, off-the-shelf array cards containing a set of curated TaqMan assays relevant for specific pathways, biomarker sets, or disease states. Use the search tool below to find an ideal TaqMan Array card.
In the search bar, enter a target gene, pathway, or specific keyword. Initiate the search to view a list of relevant assays. Results can be refined by filtering based on criteria such as gene symbol, gene name, species, and the number of RefSeq transcripts recognized. This will help narrow down options to find the most suitable assay to meet experimental needs. Each search result provides detailed information, including assay compatibility and related publications, ensuring the best match for chosen gene expression studies.
Search all TaqMan gene expression array cards
Flexible TaqMan Array cards
Flexible TaqMan Array cards are customizable microfluidic cards designed for high-throughput gene expression analysis. They allow researchers to simultaneously analyze multiple genes with high precision and efficiency. These cards can be tailored to specific experimental needs, making them exceptional for a wide range of applications, including biomarker discovery and validation. Select a predesigned TaqMan Array card using the above tool and then modify the assay content to suit experimental needs or visit the flexible content webpage.
Learn more: Flexible TaqMan array
Custom TaqMan Array cards
Custom TaqMan Array cards are the custom version of our predesigned TaqMan array cards for relative quantification of RNA expression. Provide a target sequence of interest or submit custom primer/probe sequences. Use the custom TaqMan Array card online ordering tool found below to select assays, replicates, controls, and more.
Pre-loaded 384-well microfluidic cards with selected assays in each reaction well. Choose target assays from over thousands of inventoried TaqMan Gene Expression Assays. Minimum order quantity of ten cards.
Specialty TaqMan Array cards
The Specialty Plating Service can be utilized for TaqMan Array cards that require custom TaqMan Assays, a mix of predesigned and custom TaqMan Assays, or a layout that is not supported by the online configurator tool. This service supports all TaqMan Assay types, including gene expression, Advanced miRNA, classic chemistry miRNA, SNP genotyping, DME, copy number variation, and mutation detection.
Explore: Specialty TaqMan Array cards
Endogenous controls for TaqMan Array cards
All custom TaqMan array cards contain a manufacturing control position. This position is used during our manufacturing quality processes; however, this assay can also be used as an endogenous control. 18S (Hs99999901_s1) is a manufacturing control choice for all designs, while human GAPDH (Hs99999905_m1) can be chosen for designs containing human and/or microbe detection assays. Adding 1–2 more candidate endogenous control assays is recommended, as 18S and GAPDH are not suitable endogenous controls in all experiments.
Learn more: TaqMan endogenous control assays
Benefits of using TaqMan Array cards
Widely cited in publications, TaqMan Array cards are exceptional and cost effective for medium-to-high throughput validation and profiling studies, whether researching cancer, stem cells, inflammation, or infectious diseases. Use the cards to validate tens or hundreds of initial hits generated from microarrays or next generation sequencing or use them for screening biomarkers and toxicology panels or analyzing pathways, target classes, and complete disease sets. Use less master mix and help stretch limited samples by using 5X less sample volume; excellent for low-expressing genes or precious samples.
Convenient
Flexible
Consistent
Highly reproducible
Time to results
Load 384 wells in approximately 10 minutes as the card arrives preloaded with your specified assays. Simply add sample and master mix via 8 channels, not individual wells.Choose from flexible designs: predesigned, flexible, custom, and specialty. Profile related genes simultaneously. Analyze up to 500 cards in a single study using our free analysis tools.Produce consistent results with low variability across multiple users and laboratories.Uniform distribution of samples with minimal handling helps reduce cross-contamination and variability.Streamlined reaction setup saves time and reduces labor-intensive pipetting steps. 
Watch this short video to see how easy it is to load a TaqMan Array card, featuring Nathan Zuzow, PhD, Field Application Scientist at Thermo Fisher Scientific.
Learn more:  TaqMan Array cards user guide
Flexible vs custom content TaqMan Array cards
TaqMan Array card resources
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments