Search
Raw Materials for GMP mRNA Manufacturing |
Help accelerate mRNA therapeutics research and commercialization by sourcing the right raw materials. Thermo Fisher Scientific helps provide a wide range of products and services throughout every stage of your mRNA therapeutic development cycle such as research and discovery, pre-clinical development, clinical development, and commercial manufacturing.
Request Access to a DMF Letter of Authorization (LOA) or a Regulatory Support File (RSF)
Figure 1. mRNA manufacturing workflow
mRNA production involves DNA template preparation, in vitro transcription (IVT), mRNA modification, and DNA and protein removal steps. Our innovative solutions and custom services are designed to meet your specific quality, purity requirements, and scalable batch-to-batch consistency.
TheraPure SuperFi II DNA Polymerase is a hot-start, high-fidelity enzyme manufactured using animal-origin-free (AOF) and beta-lactam–free processes and materials. It is designed for PCR-based DNA template generation for mRNA production by in vitro transcription (IVT). Enzyme delivers exceptional accuracy, purity, and consistency—enabling mRNA therapeutic developers and manufacturers to rapidly generate high-quality DNA templates.
In vitro DNA templates can be produced using techniques such as rolling circle amplification (RCA) or PCR. RCA is a cell-free amplification method that is a promising alternative to traditional plasmid DNA and PCR amplification. RCA requires polymerases with strong displacement activity and dNTPs for strand synthesis; the reactions take place at constant temperature.
| Product | Standard | TheraPure | TheraPure GMP |
|---|---|---|---|
| SuperFi II DNA polymerase | Platinum SuperFi II DNA Polymerase | NEW TheraPure SuperFi II DNA Polymerase (2 U/μL) | - |
| Phi29 DNA polymerase | EP0091 | TheraPure phi29 DNA Polymerase (10 U/μL) | - |
| dNTPs | dNTP Set, 100 mM Solutions | - | TheraPure GMP dNTPs |
Plasmid DNA is linearized with restriction enzyme, and the linearized DNA is then used as a template for in vitro transcription to produce mRNA.
| Product | Standard | TheraPure | TheraPure GMP |
|---|---|---|---|
| Eam1104I restriction enzyme | Eam1104I (EarI) (10 U/μL) | TheraPure restriction enzymes | TheraPure GMP Eam1104I Restriction Enzyme, 20 U/μL |
During IVT reaction, the DNA template, RNA polymerase, NTPs, RNase inhibitor, pyrophosphatase, and IVT buffer are critical components for mRNA production.
| Product | Standard | TheraPure | TheraPure GMP |
|---|---|---|---|
| T7 RNA polymerase | T7 RNA Polymerase, HC (200 U/μL) | TheraPure RNA polymerases | TheraPure GMP T7 RNA Polymerase, 200 U/μL |
| SP6 RNA polymerase | SP6 RNA Polymerase, HC (≥100 U/μL) | - | - |
| Ribonuclease inhibitor | RiboLock RNase Inhibitor (40 U/μL) | TheraPure modifying enzymes | TheraPure GMP RNase Inhibitor, 40 U/μL |
| Pyrophosphatase, inorganic | Pyrophosphatase, inorganic (0.1 U/μL) | TheraPure modifying enzymes | TheraPure GMP Pyrophosphatase, inorganic, 0.1 U/μL |
| NTPs | NTP Set, Tris buffered NTP Set, 100 mM Solution | TheraPure nucleotides | TheraPure GMP nucleotides |
| N1-Methylpseudo-UTP | Contact us | - | TheraPure GMP N1-Methylpseudo-UTP, 100 mM sodium solution |
| IVT buffer | Buffers for RNA transcription and post-transcriptional modifications | - | Contact us |
Co-transcriptional or post-translational capping protects mRNA and promotes mRNA function. The poly-A tail addition stabilizes and prevents degradation of mRNA.
| Product | Standard | TheraPure | TheraPure GMP |
|---|---|---|---|
| Vaccinia Capping Enzyme | Modifying enzymes | TheraPure Vaccinia Capping Enzyme, 100 U/µL | - |
| Vaccinia mRNA Cap 2'-O-Methyltransferase | Modifying enzymes | TheraPure 2'-O-Methyltransferase (125 U/μL) | - |
| S-Adenosyl-L-Methionine (SAM) | Co-factors | TheraPure S-Adenosyl-L-Methionine (32 mM) | TheraPure GMP S-Adenosyl-L-methionine, 32 mM |
| Poly A polymerase | Poly(A) Polymerase (cloned) 2 U/μL | - | - |
After IVT, the DNA template can be removed using DNAse enzyme. Proteinase K might be used for protein removal.
| Product | Standard | TheraPure | TheraPure GMP |
|---|---|---|---|
| DNAse I | DNase I, RNase-free, HC (50 U/μL) | TheraPure modifying enzymes | TheraPure GMP DNase I, RNase-free, 50 U/μL |
| Turbo DNAse | TURBO DNase (2 U/μL) | - | - |
| Proteinase K | Proteinase K, recombinant, PCR grade | - | - |
Quality grades of products differ in variety of attributes, manufacturing processes, quality control documentation package available. See Table 1 for main attributes.
| Product | Standard | TheraPure | TheraPure GMP |
|---|---|---|---|
| AOF (animal origin free) and beta-lactam free process and materials | No | Yes | Yes |
| AOF and beta-lactam free facility | No | No | Yes |
| Manufacture and documentation follow ICH Q7 guidelines | No | No | Yes |
| Validated product-specific manufacturing processes and analytical methods | No | No | Yes |
| Product-specific stability | No | No | Yes |
| Impurity testing, including melamine and nitrosamine | No | No | Yes |
| Quality documentation | CoA, CoO | CoA, CoO, TSE/BSE statement | CoA, CoO, TSE/BSE statement, impurity profiles |
| DMF (drug master file) | No | No | Yes |
| Recommended use | Research use only | Proof of concept, research, and pre-clinical development | Clinical and commercial manufacturing |
Technical expertise
50 years in enzyme and other reagent development have generated a substantial knowledge background that can be applied to further innovations.
Manufacturing
Our facilities are equipped to support high-scale product manufacturing. The animal origin-free and beta-lactam free facility operates under ISO 9001 and follows applicable ICH Q7 guidelines, ensuring the highest standards of quality and safety for TheraPure GMP products.
Quality
Implemented ISO9001 and/or ISO13485 quality management systems and extensive product testing ensures traceability, high product quality, and batch-to-batch consistency.
Supply chain
Different methods of transportation ensure smooth and safe product delivery directly to the customer or via a network of worldwide distribution centers.
* “TheraPure GMP” refers to the quality level of the raw, ancillary, or starting materials to be used for further manufacturing. TheraPure GMP products are manufactured in facilities with ISO 9001–certified quality management systems that operate in accordance with relevant good manufacturing practice (GMP) principles, as outlined in ICH Q7 or equivalent guidance documents or standards.