Gene Expression Assays
TaqMan Gene Expression Assays
  • Gold-standard specificity, sensitivity, and reproducibility
  • Don't waste precious time optimizing your SYBR primers—get results faster with proven TaqMan qPCR chemistry
  • Designed to detect virtually any gene transcript, with more than 2.6 million predesigned assays
  • Best-coverage assays available to detect the highest number of transcript variants possible
  • Available for 29 species and some microbial pathogens
  • Flexible formats: single tubes, 96-well plates, 384-well plates, 384-well array cards, and OpenArray plates
Assay search tool
TaqMan Assay formats and custom assays
We offer a custom assay design tool to detect any gene product not covered by our predesigned assay collection.
TaqMan Gene Expression Assays are available in easy-to-use 96-well plates and higher-throughput 384-well microfluidic cards. Order from our collection of gene signature panels or customize your own plates or cards. 384-well plates can also be configured with any pre-designed or custom assay using Specialty TaqMan Array Plates.
Applied Biosystems TaqMan OpenArray technology is an affordable real-time PCR–based solution for studying hundreds of samples per day, using the Applied Biosystems QuantStudio 12K Flex Real-Time PCR System with OpenArray Block. Each OpenArray plate can hold as many samples as 8 traditional 384-well plates.
Watch this short video to see how easy it is to find the right TaqMan Gene Expression Assay using the Assay Search Tool above.