Fast and simple sample preparation
The Ion OneTouch 2 System simplifies the workflows for the Ion GeneStudio S5 Series, Ion Proton, and Ion PGM systems by providing an automated solution for scalable and reproducible template preparation.
Request a quote
Specification sheet  |  Learn more about Ion Torrent NGS systems
  • Enables automated template preparation workflows into a single system
  • Provides scalable template preparation for all Ion semiconductor sequencing chips
  • Enables parallel processing of multiple samples per day through modular design
  • Supports multiplexed samples
  • Fits easily on any benchtop with its small instrument footprint
  • Supports up to 400 bases on the Ion GeneStudio S5 Series and Ion PGM System and up to 200 bases on the Ion Proton System*
Ion OneTouch 2 workflow
  • Load amplification plate 
  • Install reagent and collection tubes 
  • Add library to reaction filter
  • Prepare 8-well strip 
  • Recover enriched sample
Ion GeneStudio S5 Series, Ion Proton, or Ion PGM sequencer run
  • Load amplification plate 
  • Install reagent and collection tubes 
  • Add library to reaction filter
  • Prepare 8-well strip 
  • Recover enriched sample
Ion GeneStudio S5 Series, Ion Proton, or Ion PGM sequencer run
Save even more hands-on time with the Ion Chef System for automated template preparation and chip loading.
Learn more ›
Visistat reference component