Reliably detect and monitor a broad range of emissions and source gases
Emissions and source gas monitoring solutions from Thermo Fisher Scientific leverage proven technologies, robust instrument designs and innovative data delivery systems to help ensure safety and regulatory compliance. Choose from a wide range of instruments that reliably measure air quality, low and high levels of criteria pollutants, various gases, and toxins according to your needs.
Carbon Dioxide (CO₂)
Human activity such as burning fossil fuels (coal, natural gas, and oil), solid waste, trees, and other materials, as well as some chemical reactions can release large amounts of CO₂ into the air. As such, continuous monitoring of carbon dioxide has become critical to compliance with every increasing government regulation for industry.
The US Environmental Protection Agency (EPA) has taken regulatory actions to reduce greenhouse gas emissions.
Measuring carbon dioxide in emissions
Our CO₂ analyzers employ reliable Nondispersive Infrared (NDIR) technology for long instrument life and advanced optical filters for accuracy.
Thermo Scientific 410iQ Carbon Dioxide Gas Analyzer
Thermo Scientific 410iQ High Level Carbon Dioxide Gas Analyzer
Thermo Scientific OPGAL EyeCGas Optical Gas Imaging Camera
Carbon Monoxide (CO)
CO is a colorless, odorless gas that is naturally occurring but can cause sickness and death when present in large concentrations.
Carbon Monoxide (CO) is one of the six air pollutants controlled by EPA's National Ambient Air Quality Standards (NAAQS).
Detecting poisonous carbon monoxide 
Our CO analyzers use advanced gas filter correlation technology to detect and measure CO gas concentrations in ambient air.
Thermo Scientific 48iQ Carbon Monoxide Analyzer
Thermo Scientific 48iQTL Trace Level Carbon Monoxide Analyzer
Thermo Scientific Model 48i CO Analyzer
Thermo Scientific Model 48i-TLE Enhanced Trace Level CO Analyzer
Hydrocarbons (CₙH2ₙ+2)
Monitoring hydrocarbons is crucial for environmental protection, industrial safety, and regulatory compliance. Hydrocarbons can be found, among other places, throughout petrochemical plants, refineries, and chemical manufacturing. Hydrocarbons contribute to the formation of ground-level ozone and smog, which can harm respiratory health, thus making effective air quality monitoring essential.
Hydrocarbons can be effectively monitored, helping to protect human health and the environment
Thermo Scientific emissions monitoring and source gas monitoring automated systems provide real-time monitoring of hydrocarbon emissions from industrial stacks and processes, which is essential for regulatory compliance and helping to ensure that emissions remain within permitted limits.
Thermo Scientific Model 51i Total Hydrocarbon Analyzer (Low Temperature)
Thermo Scientific Model 51i Total Hydrocarbon Analyzer (High Temperature)
Thermo Scientific OPGAL EyeCGas Optical Gas Imaging Camera
Hydrogen Sulfide (H₂S)
Hydrogen sulfide is an extremely flammable and highly toxic gas.  It is dangerous even at low concentrations and it occurs naturally in sewers, manure pits, well water, oil and gas wells, but is also used or produced in many manufacturing and industrial activities. 
Hydrogen sulfide (H2S) poses a significant risk to worker safety in the oil and gas production, mining, paper production and wastewater treatment industries, among others. 
Reliable data capture 
Hydrogen sulfide and sulfur dioxide analyzers utilize pulsed fluorescence technology for precise detection and concentration measurements of H₂S and S0₂ in the air.
Thermo Scientific 450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer
Thermo Scientific Model 450 Hydrogen Sulfide - Sulfur Dioxide Analyzer
Mercury (Hg)
Help ensure complete regulatory compliance with unattended, true continuous monitoring of mercury in exhaust stacks from coal-fired boilers, waste incinerators, cement kilns and other industrial combustion sources.
Meets U.S. EPA 40 CFR Part 75 requirements with true continuous monitoring
Regulatory compliance peace of mind
Measure elemental, ionic, and total mercury in exhaust stacks from coal-fired boilers and waste incinerator. Meet or exceed performance specifications outlined in US EPA PS-12A and/or Part 75 provisions for continuous Mercury CEM systems in addition to the latest MATS and MACT rules.
Thermo Scientific Mercury Freedom System
Multi-gas and multi-gas calibrator
Thermo Scientific multi-gas calibrators are essential for calibrating gas detectors and analyzers to help ensure accurate measurements. Instrument output is used to calibrate instruments that perform zero, precision, Level 1 span checks, audits, and multi-point routines in accordance with US EPA requirements.
CE, TUV-SUD Safety, UKCA Compliance
Help ensure accurate measurements
Produce precise and known concentrations of gas mixtures and generate gas standards to calibrate gas monitoring instrumentation.
Thermo Scientific 146iQ Multi-Gas Calibrator
Thermo Scientific Model 146i Multi-Gas Calibrator
Thermo Scientific NDIR Model 60i Multi-Gas Analyzer
Thermo Scientific OPGAL EyeCGas Optical Gas Imaging Camera
Nitrogen (N₂) and Nitrous Oxide Species (NO, NO₂, NOₓ NOᵧ)
Various nitrogen and nitrous oxide compounds (NO, NO2, NOY, NOx ) are emitted during agricultural, land use, industrial activities, fossil fuels combustion, solid waste management and the treatment of wastewater.  As these gases can be harmful in various concentrations, careful and accurate monitoring is essential.
N2 accounts for ~7% of all US greenhouse gas emissions from human activities.
Predictive diagnostics  
Our analyzers utilize chemiluminescence technology to measure the amount of nitrogen, and nitrous oxides in the air from sub-ppb levels up to 1000ppb.
Thermo Scientific 42iQ NO-NO2-NOx Analyzer
Thermo Scientific 42iQTL Trace Level NO-NO2-NOx Analyzer
Thermo Scientific Model 42i-TL TRACE Level NOx Analyzer
Thermo Scientific Model 42i-Y NOY Analyzer
Thermo Scientific 42iQ Low Source NO-NO2-NOx Analyzer
Thermo Scientific Model 42i-LS Low Source NO-NO2-NOx Analyzer
Thermo Scientific Model 42i-HL High Level NO-NO2-NOx Analyzer
Selective catalytic reduction (SCR)
Selective noncatalytic reduction (SNCR) is a post-combustion emissions control technology for reducing NOx by injecting an ammonia-type reactant into the furnace at a properly determined location. We have the experience and technology available to help you comply with ammonia slip regulations, or to make measurements to optimize your process for De-NOx in the Selective Catalytic Reduction (SCR) or Selective noncatalytic reduction (SNCR) of coal and gas power plants.
Coal-fired and gas power plants are significant sources of NOx emissions
NO, NOₓ, and Nₜ modes of operation
Our highly reliable ammonia analyzers utilize chemiluminescence technology for accurate, continuous ammonia monitoring.
Thermo Scientific Model 17i Ammonia Analyzer
Sulfur Dioxide (SO₂)
Sulfur dioxide (SO₂) is produced in significant quantity through the burning of fossil fuels during the production of energy and other industrial processes, including industrial transportation.  Sulfur dioxide can harm the human respiratory system and contribute to environmental pollution.
SO₂ is the sulfur oxide of most concern to regulatory agencies.
Highly precise
Our sulfur dioxide analyzers measure up to 100ppm in ambient air using pulsed fluorescence technology.
Thermo Scientific 43iQ Sulfur Dioxide Analyzer
Thermo Scientific 43iQ Trace Level Sulfur Dioxide Analyzer
Thermo Scientific 43iQ High Level Sulfur Dioxide Analyzer
Thermo Scientific Model 450 Hydrogen Sulfide - Sulfur Dioxide Analyzer
Thermo Scientific Continuous Emissions Monitoring System for Total Reduced Sulfur
Zero air supply and calibration
Zero air, or air that has been purified by removing pollutants such as NO, NO₂, O3, SO₂, CO, and hydrocarbons, is used to test and calibrate spanning ambient air analyzers during routine maintenance.  Other analyzers can be calibrated using a multi-gas calibration device that performs zero, precision, Level 1 span checks, audits, and multi-point routines in accordance with US EPA requirements.
Helping to ensure optimal performance for your monitoring and analysis systems.
Personal device connectivity
We offer zero air supply and multi-gas calibrators to keep your systems running at peak performance.
Thermo Scientific 111iQ Zero Air Supply
Thermo Scientific 146iQ Multi-Gas Calibrator
Thermo Scientific 1160 Zero Air Supply
Thermo Scientific Model 1150 Dual Reactor
Thermo Scientific™ 111 Zero Air Supply
iQ Series platform
Smart ambient air monitoring 
Built-in predictive diagnostics and preventive maintenance schedules identify problems before they occur.
Thermo Scientific 410iQ Carbon Dioxide Gas Analyzer
Thermo Scientific 410iQ High Level Carbon Dioxide Gas Analyzer
Thermo Scientific 48iQ Carbon Monoxide Analyzer
Thermo Scientific 48iQTL Trace Level Carbon Monoxide Analyzer
Thermo Scientific 450iQ Hydrogen Sulfide and Sulfur Dioxide Analyzer
Thermo Scientific 42iQ NO-NO2-NOx Analyzer
Thermo Scientific 42iQTL Trace Level NO-NO2-NOx Analyzer
Thermo Scientific 49iQ Ozone Analyzer
Thermo Scientific 49iQ Ozone Primary Standard
Thermo Scientific 43iQ Sulfur Dioxide Analyzer
Thermo Scientific 43iQ Trace Level Sulfur Dioxide Analyzer
Thermo Scientific 43iQ High Level Sulfur Dioxide Analyzer
Thermo Scientific 146iQ Multi-Gas Calibrator
Service and product availability may vary by country and are subject to varying regulatory requirements. Please contact your local sales representative for availability.