E-Gel Power Snap Electrophoresis Systems, including Power Snap Plus and Power Snap Lite, are compact benchtop instruments that have integrated nucleic acid electrophoresis and gel imaging functionalities. The instrument has two main components: the instrument device base and the camera to capture gel images. The instrument works with precast E-Gel agarose gels, which are buffer-less gels containing SYBR safe DNA stains, and ladders. This eliminates the need to pour gels, prepare buffers, and handle toxic dyes such as ethidium bromide. These include specialized formats such as E-Gel CloneWell gels, which enable DNA purification directly from the gel and eliminate the need for gel extraction steps.
The components of the E-Gel Power Snap systems are designed to help save time and streamline the nucleic acid electrophoresis workflow. This system facilitates a simple three-step workflow: load, run, and analyze (Figure 1).
E-Gel Power Snap Plus System
The Invitrogen E-Gel Power Snap Plus Electrophoresis System combines the convenience of high-quality visualization in both high- and low- throughput Invitrogen E-Gel products.
Invitrogen E-Gel Power Snap Plus Electrophoresis System
Watch the video to see how precast agarose E-Gels and integrated imaging simplify DNA gel electrophoresis, from loading and running gels to real-time visualization and analysis using the E-Gel Power Snap Plus system.
Advantages of E-Gel electrophoresis device and imaging systems
The current E-Gel electrophoresis lineup, including Power Snap Lite and Power Snap Plus, offers streamlined, integrated workflows and flexible sample throughput, representing a modern upgrade over traditional gel analyzers and separate imaging systems for DNA/RNA electrophoresis.
  • Real-time visualization: Amber filter to monitor real-time sample migration
  • Sample safety: Blue-light transilluminator does not damage DNA samples
  • Intuitive interface: Easy operation with user-friendly touch screen and preset protocols
  • Flexible throughput: Gels with 11-, 22-, 48-, and 96-wells can be run (model-dependent)
  • High resolution camera for image capture, with image editing capabilities
  • Internal storage: Up to 3 months of experimental data can be stored in the device
  • Small footprint: 6x smaller than traditional gel documentation systems
E-Gel Power Snap Systems: A comparison
 E-Gel Power Snap Plus Electrophoresis SystemE-Gel Power Snap Lite Electrophoresis System
 E-Gel Power Snap Plus System comprising gel electrophoresis device and camera moduleE-Gel Power Snap Lite agarose gel electrophoresis device with and without camera attachment and a precast agarose gel cassette
Video overviewWatch nowWatch now
Virtual tourView nowView now
Capability overviewFlexible, high-resolution DNA screening; designed for dedicated DNA or RNA analysis workflows, supporting both low- and high-throughput nucleic acid imaging and analysis.For routine DNA and RNA electrophoresis; optimized for quick verification and supporting workflow checks; suitable for running 11- to 22-well precast agarose gels.
E-Gel agarose gel compatibility11-, 22-, 48-, and 96-wells*, CloneWell II and SizeSelect II gels with adaptor11- and 22-wells, CloneWell II and SizeSelect II gels
Camera resolution13 MP4 MP
Image storage64 GB32 GB
Cloud enabledYesNo
iBright Analysis Software compatibleYesNo
Base module dim. (L x W x H)Plus: 29.1 x 22.0 x 10.1 cmLite: 24.2 x 13.0 x 7.0 cm
Full system dim. (L x W x H)Plus: 29.1 x 22.0 x 32.9 cmLite: 26.0 x 13.0 x 20.0 cm
Printer compatibleYes, Sony UP-X898MD thermal printerNo
*Contains additional wells for marker lanes
E-Gel Power Snap Plus Electrophoresis System
Maintain productivity with extended warranties
Rapid Exchange (REX) one-year extended warranties are available (at an additional charge) for both the E-Gel Power Snap Plus device and camera. If your instrument needs repair, just call our support center and we will exchange your instrument for a factory-certified, refurbished replacement instrument within one business day from the time we determine a replacement is required. Replacement instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.
E-Gel Power Snap Plus System
Learn how to perform high throughput gel electrophoresis, monitor DNA migration, and capture gel images with the E-Gel Power Snap Plus System. Find out about the image editing and image storage options.
E-Gel Power Snap Plus How-to: Deconvolution Overview
Discover the features of the Power Snap Plus E-Gel system, including how to easily read results with on‑instrument deconvolution and analysis for Invitrogen E-Gel 96-well gels.
E-Gel Power Snap Plus How-to: Perform an E-Gel CloneWell II Agarose Gel Run
Learn how to simplify agarose gel electrophoresis and analysis using the Invitrogen E-Gel Power Snap Plus Electrophoresis System. In this video we demonstrate how to perform an E-Gel CloneWell II, 0.8% agarose gel run.
E-Gel Power Snap Lite Electrophoresis System
How to quickly purify DNA with E-Gel CloneWell II precast gels
Watch a demo of how to gel purify DNA using the E-Gel Power Snap Lite System and E-Gel CloneWell II gel. Load samples, run the gel, capture bands of interest in the recovery well, and pipette out purified DNA bands. No additional purification steps needed.
How to perform rapid electrophoresis using an E-Gel Power Snap Lite device and E-Gel EX Cassette
Watch a demo of agarose gel electrophoresis using an E-Gel Power Snap Lite System and E-Gel EX Cassette. The ease and simplicity of the process is excellent for research labs.
Image analysis
iBright Image Analysis Software analyzes data from E-Gel images captured by the integrated camera. The gel images and the results of data analysis can be stored on Thermo Fisher’s Connect Platform. iBright Image Analysis is included in the E-Gel Power Snap Plus Electrophoresis System.
Learn more about iBright Image Analysis Software
The E-Gel Power Snap Plus System firmware is periodically updated with fixes and new and enhanced features to offer a rich user experience.
Keep up-to-date on firmware upgrades
iGlow Gel Documentation System
Designed for fast, one-touch image capture for protein and nucleic acid gel imaging with integrated UV, blue, and white light illumination. The system is compatible with nucleic acid and protein gels, and various gel sizes.
Learn more
E-Gel precast gel electrophoresis systems
See all the benefits of the innovative, fast, and buffer-less system in one place.
Download brochure
Invitrogen E-Gel products to complete your gel electrophoresis system
For best results, the E-Gel Power Snap and Power Snap Plus System must be used in conjunction with other Invitrogen E-Gel line of products, which include:
Citations
Separation and analysis of DNA and RNA by agarose gel electrophoresis is a critical aspect of many biological and research areas. Access peer-reviewed journal articles showing how Thermo Fisher Scientific products, including the E-Gel Power Snap systems, are being used to support important research and innovations. Visit the citations hub to explore additional publications. 
Area(s) of study
Research focus
Reference
Disease target discovery & validationCancerSun Y, Revach OY, Anderson S et al. (2023) Targeting TBK1 to overcome resistance to cancer immunotherapy. Nature 615(7950):158–167. doi: 10.1038/s41586-023-05704-6 PMID: 36634707
Foundational research, Disease target discovery and validation
Cancer, ecology
Garrett NR, Watkins J, Francis CM et al. (2023) Out of thin air: surveying tropical bat roosts through air sampling of eDNAPeerJ 11:e14772. doi: 10.7717/peerj.14772 PMID: 37128209
Molecular methodologyMolecular diagnosticsSaavedra-Matiz CA, Isabelle JT, Biski CK et al. (2013) Cost-effective and scalable DNA extraction method from dried blood spotsClin Chem 59(7):1045–1051. doi: 10.1373/clinchem.2012.198945 PMID: 23509109
Disease target discovery and validationNeurosciencePeruzzotti-Jametti L, Willis CM, Krzak G et al. (2024) Mitochondrial complex I activity in microglia sustains neuroinflammationNature 628(8006):195–203. doi: 10.1038/s41586-024-07167-9 PMID: 38480879
Application notes and manuals
Resources
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.