Search
The Applied Biosystems Veriti Thermal Cycler set a new standard for innovation when this thermocycler launched in 2007. It was the first thermal cycler to employ an LCD touchscreen interface, multiple temperature zone block, and web-enabled remote-control software. Building on these innovations, the VeritiPro Thermal Cycler upgrade features the latest block technology and connectivity options. These advancements provide higher ramp rates, quieter runs, and more intuitive VeriFlex temperature optimization setup while continuing to offer the same high-end performance.
The VeritiPro Thermal Cycler comes with two options:
Gradient temperature control is a feature of thermal cyclers to help primer annealing in PCR. However, thermal cyclers equipped with “better-than-gradient” technology are alternatives that help improve temperature control for primer annealing. These metal blocks often come with three or more segmented blocks, each with separate heating and cooling elements. VeritiPro Thermal Cycler is equipped with “better-than-gradient” technology, namely Veriflex Blocks.
We've made Veriti Thermal Cycler even better—Meet VeritiPro Thermal Cycler. Watch this video to view features and capabilities of our gradient thermocycler, VeritiPro Thermal Cycler.
Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
Plastics compatibility chart for Applied Biosystems instruments
PCR and cDNA Synthesis Support Center
Tips and troubleshooting help
School of Molecular Biology, PCR Education
Technical content for new and experienced molecular biologists
Thermal Cycler Software and Firmware Files
Download the latest updates for your instrument
Interested in private label thermal cyclers?
Submit an inquiry
We provide a full range of service options for all Applied Biosystems thermal cyclers.
View service plans
Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
View temperature service options
Download MDTV flyer
For Research Use Only. Not for use in diagnostic procedures.