Search
To help ensure efficient and accurate work across different cell lines, it is essential to adhere to fundamental processes in seeding, subculturing, and maintenance. These steps are crucial for the successful growth and proliferation of all cells. Procedures for maintaining cells can vary based on the cell line you are working with because each cell line often requires a unique growth environment and specific culture conditions.
However, foundational processes such as establishing an appropriate seeding density, monitoring growth patterns, and adhering to the appropriate subculturing protocols are elements that all cells need to grow and proliferate successfully.The information and protocols on this page will help you develop a baseline understanding of the majority of hands-on culturing procedures.
Cell seeding is a fundamental process in the cell culture workflow that involves placing cells into a culture vessel or onto a surface to allow them to attach, grow, and proliferate. For adherent cells, seeding cells means plating the cells into a culture vessel with pre-warmed medium and incubating them under suitable conditions. In suspension cells, seeding involves transferring the calculated number of cells directly into the culture medium. This process starts with preparing a single-cell suspension from an active culture, a newly thawed vial, or a newly established cell line. This is followed by counting the cells to determine the appropriate seeding density.
In adherent cultures, seeding density is the number of cells plated per surface area unit, typically cm2 for most common dishes, plates, and flasks. In suspension cultures, seeding density refers to the number of cells added per unit volume of the culture medium in a vessel. Seeding cells at the optimal density is crucial because it determines the initial cell concentration, which ultimately affects cell growth, nutrient availability, and space for proliferation. A proper cell seeding density provides adequate resources and room for cells to grow, leading to healthy and uniform cultures. Incorrect seeding density can result in overcrowding or sparse growth, negatively impacting experimental outcomes and cell viability. To achieve a consistent growth rate and yield appropriate for your cell type, always follow the instructions included with your product.
Successfully seeding cells for culture requires several essential materials to produce an environment conducive to cell proliferation. This involves choosing the tools and equipment you’ll employ during the experiment, but more importantly, the vessel and complete media formulation for the cell. Media composition directly influences proliferation and can be the root cause of experimental failures. Seeding density also influences confluence, so tracking cell reproduction with hemocytometers or automated cell counters is essential for checking baseline seeding densities and subsequent growth through the life of your culture.
You will need:
The protocol below describes how to seed cells for the adherent or suspension culture of mammalian cells. Before you prepare your cells for the cell seeding protocol, it is recommended to ready your materials from the list above and complete the following calculations to limit the time cells may be exposed to contamination.
Subculturing, also referred to as splitting or passaging cells, is the removal of medium and transfer of cells from a previous culture into fresh growth medium, a procedure that enables the further propagation of the cell line or cell strain.
The difference between seeding and passaging cells is that seeding is the placement of cells in a vessel for the purpose of starting a new culture, while subculturing is meant to split up those cells to maintain optimal environments for each set of cells. Passaging gives the cells room to proliferate and presents nutrient stability for cells to remain in log phase growth. In short, seeding is diluting a concentrated cell suspension for new cultures, while subculturing, passaging, and splitting all refer to dividing up an active culture to encourage log phase growth.
Passaging cells requires diligent maintenance and adherence to aseptic techniques. While passaging, carefully record your results and monitor morphology and cell viability for signs of contamination or abnormal behavior.
You can find direction for subculturing adherent or suspension cells in the following protocols:
The criteria for subculturing is similar in adherent and suspension cultures. Both culture types should be passaged when they are in the log phase, before they reach confluence. Normal cells stop growing when they reach confluence (contact inhibition), and it takes them longer to recover when reseeded. Transformed cells can continue proliferating even after they reach confluence, but they usually deteriorate after about two doublings. If not passaged prior to confluency, suspension cells can clump together and the medium will appear turbid when the culture flask is swirled.
You should also subculture your cells if you observe a rapid drop in pH (>0.1–0.2 pH units) with an increase in cell concentration. A drop in the pH of the growth medium usually indicates a buildup of lactic acid, which is a by-product of cellular metabolism. Lactic acid can be toxic to the cells, and the decreased pH can be sub-optimal for cell growth. The rate of change of pH is generally dependent on the cell concentration in those cultures—a high cell concentration exhausts medium faster than cells at lower concentrations.
Monitoring and subculturing cells on a precise schedule allows you to closely monitor the health of your cells and helps ensure your results are reproducible. Vary the seeding density of your cultures until you achieve consistent growth rate and yield appropriate for your cell type from a given seeding density. Deviations from the growth patterns usually indicate that the culture is unhealthy (e.g., deterioration, contamination) or a component of your culture system is not functioning properly (e.g., temperature is not optimal, culture medium too old).
You can passage cells for as long as the cell line is maintaining the expected properties, though it is important to track the number of times the cells have been split. Repeated subculture can result in changes to the cell’s key characteristics and genomic stability and potentially affect your experimental outcome. Be cautious of continuous cell lines that have been passaged repeatedly, and keep thorough notes.
In addition, we strongly recommend that you keep a detailed cell culture log, listing the feeding and subculture schedules, types of media used, the dissociation procedure followed, split ratios, morphological observations, seeding concentrations, yields, and any antibiotic use.
It is best to perform experiments and other non-routine procedures (e.g., changing type of media) according to your pre-determined subculture schedule. If your experimental schedule does not fit the routine subculture schedule, make sure that you do not passage your cells while they are still in the lag period or when they have reached confluency and ceased growing.
Maintaining a regular culture care schedule is crucial to the viability of your cells throughout the duration of your experiment. This is because the growth of cells in culture follows a standard pattern. A lag after seeding is followed by a period of exponential growth, called the log phase. When cells in adherent cultures use all the nutrients in the growth media or cover the entire substrate with no space left to expand, they enter the stationary phase (also known as the plateau phase). Similarly, cells in suspension cultures will enter the stationary phase when they exceed the medium's capacity to support further growth. In this phase, cell proliferation significantly slows down or stops entirely.
Cells should be passaged, or subcultured, when they cover the plate, or the cell density exceeds the capacity of the suspension medium. This will keep cells at an optimal density for continued growth and stimulate further proliferation. Maintaining log phase growth will maximize the number of healthy cells for your experiment.
These useful numbers for cell culture can help you determine a starting seeding density and cells at confluency count for various sizes of cell culture dishes, plates, and flasks.
The first phase of growth after the culture is seeded is the lag phase, which is a period of slow growth when the cells are adapting to the culture environment and preparing for fast growth.
The lag phase is followed by the log phase (i.e., “logarithmic” phase), a period where the cells proliferate exponentially and consume the nutrients in the growth medium. The log phase is usually a crucial time to perform subculturing procedures and is the ideal time to freeze your unused cell stock to maintain excellent cell health for future experiments.
The stationary phase is characterized by a slowing of mitosis within the cell culture. Confluency is at a maximum as the mitotic index declines, indicating that cell proliferation has reduced or stopped entirely. During this phase, cells may also undergo metabolic changes and increased stress due to nutrient depletion of the media and accumulation of waste products such as lactic acid, a byproduct of cell metabolism.
There are several common problems that you may encounter when culturing cells. Problems in primary cultures may have different causes than the same problem in established cell lines. The table below addresses some of the common problems, along with their possible causes and suggested solutions.
| Problem | Possible Cause | Suggested Solution |
|---|---|---|
| Rapid pH shift in medium | Incorrect carbon dioxide (CO2) tension | Increase or decrease percentage of CO2 in the incubator based on concentration of sodium bicarbonate in medium. For sodium bicarbonate concentrations of 2.0 to 3.7 g/L, use CO2 amounts of 5% to 10%, respectively. Switch to CO2-independent medium. |
| Overly tight caps on tissue culture flasks | Loosen caps one-quarter turn. | |
| Insufficient bicarbonate buffering | Add HEPES buffer to a final concentration of 10 to 25 mM. | |
| Incorrect salts in medium | Use an Earle’s salts-based medium in a CO2 environment and a Hanks’ salts-based medium in atmospheric conditions. | |
| Bacterial, yeast, or fungal contamination | Discard culture and medium. Try to decontaminate culture. See Using Antibiotics and Antimycotics | |
| Precipitate in medium, no change in pH | Residual phosphate left over from detergent washing, which may precipitate powdered medium components | Rinse glassware in deionized, distilled water several times, then sterilize. |
| Frozen medium | Warm medium to 37°C and swirl to dissolve. If precipitate remains, discard medium. | |
| Precipitate in medium, change in pH | Bacterial or fungal contamination | Discard medium. Try to decontaminate culture. See Using Antibiotics and Antimycotics |
| Cells not adhering to culture vessel | Overly trypsinized cells | Trypsinize for a shorter time or use less trypsin. See Cell Dissociation Protocol |
| Mycoplasma contamination | Segregate culture and test for mycoplasma infection. Clean hood and incubator. If culture is contaminated, discard. | |
| No attachment factors in medium | For serum-free formulations, be sure they contain attachment factors. | |
| Decreased or slow growth of culture | Change in medium or serum | Compare media formulations for differences in glucose, amino acids, and other components Compare the old lot of serum with the new lot in a growth experiment. Increase initial cell inoculum Adapt cells sequentially to new medium. |
| Growth medium is not correct | Use pre-warmed growth medium as recommended by the supplier. | |
| Serum in the growth medium is of poor quality | Use serum from a different lot. | |
| Cells have been passaged too many times | Use healthy, low passage-number cells. | |
| Cells were allowed to grow beyond confluency | Passage mammalian cells when they are in the log-phase before they reach confluence. | |
| Culture is contaminated with mycoplasma | Discard cells, media, and reagents. Obtain new stock of cells and use them with fresh media and reagents. |
A companion handbook that introduces the fundamentals of cell culture. Download a copy to keep as a frequent reference guide.
Watch the steps to prevent contamination of your cell culture and a demonstration of best-practice sterile techniques.
Test your cell culture knowledge and refresh on best practices in this virtual cell culture laboratory.
Grow and maintain a variety of mammalian cell lines with Gibco Cell Culture Media.
Nunc Cell Culture Plastics support every step of your research with a broad range of culture vessels.
Gibco FBS offers consistent growth for your cells through essential nutrients and growth factors.
Access cell culture and transfection educational resources to enable excellent experiment planning and execution.
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.