Search
Search

Síntese gênica personalizada GeneArt Invitrogen |
GeneArt Custom Gene Synthesis is a reliable and cost-effective method for obtaining custom DNA constructs with 100% sequence accuracy. To maximize expression of synthetic genes, we offer optimization with our patented GeneOptimizer algorithm which utilizes a unique multifactorial approach that goes beyond codon optimization.
Order clones now View GeneArt Dashboard guide
Order through the Classic GeneArt Gene Synthesis Portal
Download the sequence submission form for email ordering
If the gene synthesis vector selection tool doesn’t have the vector you’re looking for, or if you need a synthesized gene delivered in your unique vector; we have you covered.
Please note: For the best pricing and most accurate turnaround time, remember to optimize your sequences to reduce complexity.
| Range (bp) | Production time (business days) | |
|---|---|---|
| Value gene synthesis | Prime gene synthesis | |
| 100–199 | N/A | 7+ |
| 200–1,199 | 7+ | 7+ |
| 1,200–3,000 | 9+ | 9+ |
| 3,001–5,000 | 17+ | 17+ |
| 5,001–9,000 | N/A | 22+ |
| 9,001–12,000 | N/A | 27+ |
* Valid for gene synthesis (non-complex, GC content 20-80%, repetitions, secondary structures). For complex sequence synthesis, these longer production times will be communicated prior to production.
Solicite mais informações sobre os serviços GeneArt
Geneartsupport@thermofisher.com
América do Norte e Ásia: 1-800-955-6288
Europa: +49 (0)941 942 76-100
For Research Use Only. Not for use in diagnostic procedures.

