Search
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems thermal cyclers with the flexible configuration and control features that fit how you work today. Interchangeable block formats allow you to maximize your throughput or run independent experiments concurrently. And now you can access your ProFlex PCR System with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods and monitor runs from any smartphone or desktop computer.
Engineered to deliver exceptional performance, compatibility, and productivity for PCR workflows of any applications.
The ProFlex PCR System has five different blocks that can be changed with the flip of a switch, including the first-of-its-kind 3 x 32-well block. This block allows up to three experiments to be run simultaneously, completely independently of each other and without interference. Dual 96-well and dual 384-well blocks are available for your high-throughput PCR sequencing needs, and a dual flat block is available to support Applied Biosystems OpenArray plate technology for genotyping analysis on the Applied Biosystems QuantStudio 12K Flex Real-Time PCR System.
ProFlex PCR System with the 3 x 32-well block for running up to three independent experiments simultaneously.
Dual 384-well block for higher-throughput sequencing using ProFlex PCR Systems.
Dual flat block on the ProFlex PCR System supports OpenArray plate technology for genotyping analysis on the QuantStudio 12K Flex Real-Time PCR System.
96-well block for standard PCR applications using the ProFlex thermal cycler.
Dual 96-well block for the ProFlex thermal cycler for high-throughput sequencing.
Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
PCR and cDNA Synthesis Support Center
Tips and troubleshooting help.
School of Molecular Biology, PCR Education
Technical content for new and experienced molecular biologists.
Thermal Cycler Software and Firmware Files
Download the latest updates for your instrument.
Interested in private label thermal cyclers?
Submit an inquiry
We provide a full range of service options for all Applied Biosystems thermal cyclers.
View service plans
Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
View temperature service options
Download MDTV flyer
For Research Use Only. Not for use in diagnostic procedures.