Protein dialysis is used to remove small molecules such as salts and detergents from solutions, typically by diffusion through a semi-permeable membrane to improve stability of the protein or enable buffer compatibility with downstream applications. This process allows the protein to be captured while unwanted small molecules pass through the membrane into a surrounding buffer. Find high-quality dialysis products designed to help improve protein purification workflows. Dialysis products include tubing, cups, cassettes, flasks, and plates. The purpose of dialysis in protein purification is to remove small contaminants, which can interfere with protein function and downstream analyses. Explore our selection to find specific dialysis solutions for your research needs.
Dialysis products selection guide
 slide-a-lyzer dialysis cassettes of various sizes
Dialysis cassettes
slide-a-lyzer dialysis flask
Dialysis flask
slide-a-lyzer mini dialysis devices in various sizes
Mini dialysis cups
microdialysis plate and tube stripMicrodialysis platessnakeskin tube in plastic wrapping
Dialysis tubing
External float required?NoYesYes (0.1 mL size only)NoNo
Method of sample addition/removalPipettePipette, Directly pouringPipettePipettePipette
Volume range1–125 mL150–250 mL10 μL–2 mL10–100 μL10–100 mL
Number of devices with different volume capacities4–51313
Molecular weight cutoffs (MWCOs)2K, 3.5K, 7K (original only), 10K, 20K2K, 3.5K, 10K, 20K2K (0.1 mL size only), 3.5K, 10K, 20K3.5K, 10K3.5K, 7K, 10K
Automation compatible?NoNoNoYesNo
Integrated buffer reservoir?NoNoYes, except 0.1 mL sizeYesNo
Gamma-irradiated options?
  • No, E-beam irradiated available
    (G3 device)
  • Yes (original device)
NoNoNoNo
Molecular weight cutoff (MWCO) color coded?YesYesYesNoYes
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.