Search
Death receptors signaling and modulation are highly complex processes that are a part of apoptosis, or programmed cell death. Certain cells have unique sensors, termed Death Receptors (DRs), on their surface which detect the presence of extracellular death signals and, in response, rapidly ignite the cell's intrinsic apoptosis machinery. The Death Receptors belong to the TNF (Tumor Necrosis Factor) gene superfamily and generally will have several functions other than initiating apoptosis. Eight members of the Death Receptor family have been characterized so far:
Death receptor proteins are distinguished by a cytoplasmic region of approximately 80 residues termed the Death Domain (DD). When these receptors are triggered by corresponding ligands, a number of molecules are recruited to the Death Domain and a subsequent signaling cascade is activated. Death ligands also interact with Decoy Receptors (DcRs) that do not possess DDs and so cannot form signaling complexes. Decoy receptors are members of the TNFR superfamily that are capable of competing with signaling receptors for ligand binding, thereby inhibiting their function. TRAILR3 (also known as DcR1) and TRAILR4 (also known as DcR2) compete with DR4 and DR5 for binding of APO2L/TRAIL. DcR3 competes with Fas for binding of FasL and with DR3 for binding of TL1A.
Fas, a member of the TNFR family, typifies the classical view of Death Receptor function. The Fas receptor, upon binding to the Fas Ligand, trimerizes and induces apoptosis through a cytoplasmic Death Domain that interacts with signaling adaptors like FAF-1 (Fas-Associated Factor-1), FADD (Fas-Associated Death Domain), Daxx, FAP1 (Fas-Associated Protein-Tyrosine Phosphatase-1), Flash, and RIP (Receptor-Interacting Protein). FADD carries a Death Effector Domain (DED), and by homologous interaction, it recruits the DED containing Procaspase-8 protein which is in an inactive state. Procaspase-8 is proteolytically activated by Caspase-8. FLIP (FLICE-Inhibitory Protein) inhibits activation of Procaspase-8 at the DISC by blocking its processing. FLIPL also facilitates the cleavage of Procaspase-8 at the DISC by forming FLIPL-Procaspase-8 heterodimers. FADD also helps in the activation of Caspase-10. Caspase-8 can cleave the BH3-only protein BID (BH3 Interacting Domain Death Agonist), and the resulting tBID (Truncated BID) can inactivate Bcl2 (B-Cell CLL/Lymphoma-2) in the mitochondrial membrane. This allows the escape of CytoC (Cytochrome-C), which clusters with APAF1 (Apoptotic Protease Activating Factor-1) and Caspase-9 in the presence of dATP to activate Caspase-9. SMAC (Second Mitochondria-Derived Activator of Caspase)/DIABLO and HTRA2 (High Temperature Requirement Protein-A2) are also released from the mitochondria and inactivate IAPs (Inhibitors of Apoptosis), which further inhibits Caspase-3. Active Caspase-9 can cleave and activate Procaspase-3, leading to the breakdown of several cytoskeletal and nuclear proteins (structural, signaling proteins or kinases) like GDID4 (GDP-Dissociation Inhibitor-D4), PARP (Poly ADP-Ribose Polymerase), Alpha-Fodrin, GAS2 (Growth Arrest Specific-2), and Lamin-A, thus inducing apoptosis, and degradation of the ICAD (Inhibitor of Caspase-Activated DNase). Besides Fas, TRAILR1 and TRAILR2 also lead to apoptosis by formation of DISCs. TRAILR1 and TRAILR2 are activated by binding to the ligand TRAIL. DISCs also consist of oligomerized, probably trimerized, receptors, the DD-containing adaptor molecule FADD, two isoforms of Procaspase-8 (Procaspase-8/a and Procaspase-8/b), Procaspase-10, and the cellular FLIPL/S. Signaling downstream of TRAILR1/R2 receptors is similar to Fas signaling (3-4).
TNFR1 signaling differs from Fas receptor or TRAILR1/R2-induced apoptosis. TNFR1 is an integral membrane protein with its receptor domain exposed at the surface of the cell binding the complementary death activator, which transmits a signal to the cytoplasm that leads to activation of downstream products. TNFR1, after binding to its ligand TNF-alpha, recruits TRADD (TNFR-Associated Death Domain) as a platform adaptor, which then assembles alternative signaling complexes through secondary adaptors. One type of complex is a DISC that involves FADD and Caspase-8 (and probably Caspase-10) and triggers apoptosis in a manner similar to the other Death Receptors. Another complex involves RIP, TRADD, TRAF1/2 (TNFR-Associated Factor), and probably other, as-yet-unidentified molecules. It is proposed to trigger the NF-KappaB signaling pathway through recruitment of the IKK (I-KappaB Kinase) complex and activate JNK1 (c-Jun Kinase) through a TRAF2 and NIK (NF-KappaB-Inducing Kinase) dependent mechanism. ASK1 (Apoptosis signal-regulating Kinase-1), a MAP Kinase, is also required for TNF-mediated JNK activation. TNFR1 is also able to mediate apoptosis through the recruitment of an adapter molecule called RAIDD (RIP-associated ICH-1/CED-3 homologous protein with a death domain). RAIDD associates with RIP through interactions between death domains and can recruit Caspase-2 through an interaction with a motif, similar to the death effector domain, known as CARD (caspase recruitment domain). Recruitment of Caspase-2 leads to induction of apoptosis. Another kinase believed to be involved in TNF-alpha induced apoptosis is DAPK (Death-Associated Protein Kinase). DAPK is a calcium/calmodulin-regulated serine/threonine kinase that carries ankyrin repeats, a death domain, and is localized to the cytoskeleton (5-6).
The DR3 and DR6 signaling pathways are less well characterized. The ligand for DR3 is TWEAK (TNF-Related Weak Inducer of Apoptosis)/TL1A while the ligand for DR6 remains unknown. RIP and TRADD are recruited to the receptor complex, and DR3 and DR6 promote activation of NF-KappaB that leads to the expression of survival genes. Another death receptor that plays an important role in the nervous system is p75(NTR). p75(NTR) is a member of the TNFR (Tumor Necrosis Factor) receptor superfamily, having no tyrosine kinase domain. When NGF binds selectively to p75(NTR), it causes the activation of the pro-apoptotic JNK cascade. MEKKs (MAP/ERK Kinase Kinases) and SEK (SAPK/ERK Kinase) are the upstream regulators of JNK. JNKs, in turn, upregulate p53 and the pro-apoptotic member of the Bcl2 family: BAX (Bcl2 Associated-X Protein) in a sequential manner, which causes apoptosis of neuronal cells. Our current understanding of Death Receptors signaling and modulation has opened up possibilities for the design of new therapeutic strategies for targeting death receptor pathways. This may allow the treatment of a number of diseases potentially associated with defects in DR signaling, such as multiple sclerosis and Alzheimer's disease. In tumors that retain some responsiveness to conventional therapy, death receptor engagement, in combination with chemotherapy or irradiation, might lead to synergistic apoptosis activation, and reduce the probability that tumor cells resistant to either type of agent will emerge. In tumors that have lost p53 function, death receptor targeting might help to circumvent resistance to chemotherapy and radiotherapy (7-9).
Access a targeted collection of scientific application notes, methods, and cell signaling charts.
Apoptosis Information—Find application notes, webinars and other useful educational resources to help you navigate the complex world of apoptosis research.
BioProbes Journal of Cell Biology Application—Stay up-to-date with highlights of the latest breakthroughs and get information about new technologies and products.
For Research Use Only. Not for use in diagnostic procedures.