Search
Generation of reactive oxygen species (ROS) is inevitable for aerobic organisms and, in healthy cells, occurs at a controlled rate. Under conditions of oxidative stress, ROS production is dramatically increased, resulting in subsequent alteration of membrane lipids, proteins, and nucleic acids. Oxidative damage of these biomolecules is associated with aging as well as a variety of pathological events, including atherosclerosis, carcinogenesis, ischemia reperfusion injury, and neurodegenerative disorders.
CellROX Reagents are fluorogenic probes for measuring generalized oxidative stress in cells. The dyes are nonfluorescent in a reduced state and fluoresce bright green, orange, or deep red upon oxidation. Assays are simple and reliable with a sensitive and robust readout, and the reagents can be applied to cells in complete growth medium.
DAF-FM and DAF-FM diacetate can be used to detect and quantify low concentrations of nitric oxide (NO). The reagents are essentially nonfluorescent until reacted with NO to form a fluorescent benzotriazole. DAF-FM fluorescence can be detected by any fluorescence microplate reader that can detect fluorescein.
Reduced glutathione, also known as GSH, is a major thiol bound to proteins. Protein thiols, including GSH, play an important role in determining the redox status of cells. Hydrogen peroxide reacts with nucleic acids, proteins, and lipids and can result in cell and tissue damage.
Amplex UltraRed reagent (in the EnzChek MPO Activity Assay Kit) provides rapid and sensitive determination of both chlorination and peroxidation activities of MPO in solution and in cell lysates. The mix-and-read, room temperature assay can be used for continuous detection of these activities, making it ideal for high-throughput strategies.
Fluorescence emission spectra of DAF-FM in solutions containing zero to 1.2 μM nitric oxide (NO) radical.
| CellROX Deep Red Reagent, for oxidative stress detection | CellROX Green Reagent, for oxidative stress detection | CM-H2DCFDA, general oxidative stress indicator | |
|---|---|---|---|
| Target | Reactive oxygen | Reactive oxygen | Reactive oxygen |
| Reporter | CellROX Deep Red | CellROX Green | CM-H2DCFDA |
| Ex/Em (nm) | 644/665 | 485/520 | 495/525 |
| Live cell | Yes | Yes | Yes |
| Lysate | Yes | Yes | Yes |
| Purified enzyme | Yes | Yes | Yes |
| Usage | Cell permeant for live-cell determinations | Cell permeant for live-cell determinations | Cell permeant for live-cell determinations |
| Components | Bulk reagent | Bulk reagent | Bulk reagent |
| Format | 5 x 50 µL | 5 x 50 µL | 20 x 50 µg |
| Protocol outline |
|
|
|
| Cat. No. | C10422 | C10444 | C6827 |
| DAF-FM (4-Amino-5-Methylamino-2′,7′-Difluorofluorescein) | DAF-FM Diacetate (4-Amino-5-Methylamino-2′,7′-Difluorofluorescein Diacetate) | ThiolTracker Violet (Glutathione Detection Reagent) | |
|---|---|---|---|
| Target | Nitric oxide | Nitric oxide | Glutathione |
| Reporter | DAF-FM | DAF-FM | ThiolTracker Violet |
| Ex/Em (nm) | 495/515 | 495/515 | 405/526 |
| Live cell | No | Yes | Yes |
| Lysate | Yes | Yes | Yes |
| Purified enzyme | Yes | Yes | Yes |
| Usage | Lower detection limit ~3 nM of NO | Lower detection limit ~3 nM of NO | Cell permeant for live-cell determinations |
| Components | Bulk reagent | Bulk reagent | Bulk reagent |
| Format | 1 mg | 1 mg | 500 assays using 100 µL reaction volume |
| Protocol outline |
|
| |
| Cat. No. | D23841 | D23842 | T10096 |
| Amplex Red Hydrogen Peroxide/Peroxidase Assay Kit (500 assays) | APF (hydroxyl radical, hypochlorite or peroxynitrite sensor) | HPF (hydroxyl radical and peroxynitrite sensor) | MitoSOX Red Mitochondrial Superoxide Indicator, for live-cell imaging | Dihydroethidium (hydroethidine) | |
|---|---|---|---|---|---|
| Target | Hydrogen peroxide/Peroxidase | Use singly for general ROS detection; in tandem can be used to detect hypochlorous acid (HOCl) | Targeted to mitochondria | Blue fluorescent dye becomes red when oxidized | |
| Reporter | Amplex Red reagent | ARF | HPF | MitoSOX Red | Dihydroethidium (DHE) |
| Ex/Em (nm) | 571/585 | 490/515 | 510/580 | 518/606 | |
| Live cell | No | Yes | Yes | Yes | |
| Lysate | Yes | No | No | No | |
| Purified enzyme | Yes | No | NO | No | |
| Usage | Lower detection limit 1 x 10–5 U/ml HRP | Both reagents become fluorescent on reaction with ROS–APF exhibits higher response than HPF to hypochlorite anions | Detects increases in cellular superoxide production | Exhibits blue-fluorescence in the cytosol until oxidized, where it intercalates within the cell's DNA | |
| Components | Includes assay buffers and controls | Bulk reagent | Bulk reagent | Bulk reagent | Bulk reagent |
| Format | 500 assays using 100 µL reaction volume | 470 µL | 470 µL | 10 x 50 µg | 10 x 1 mg |
| Protocol outline |
|
|
|
|
|
| Cat. No. | A22188 | A36003 | H36004 | M36008 | D11347 |
| EnzChek Myeloperoxidase (MPO) Activity Assay Kit | |
|---|---|
| Target | Continuously detects chlorination and peroxidation activity of myeloperoxidase (MPO) |
| Reporter | Amplex UltraRed |
| Ex/Em (nm) | 571/585 |
| Live cell | No |
| Lysate | Yes |
| Purified enzyme | Yes |
| Usage | Measure 1.5 to 200 ng/mL MPO |
| Components | Includes assay buffers and controls |
| Format | 200 assays using 100 µL reaction volume |
| Protocol outline |
|
| Cat. No. | E33856 |
| CellROX Deep Red Reagent, for oxidative stress detection | CellROX Green Reagent, for oxidative stress detection | CM-H2DCFDA, general oxidative stress indicator | |
|---|---|---|---|
| Target | Reactive oxygen | Reactive oxygen | Reactive oxygen |
| Reporter | CellROX Deep Red | CellROX Green | CM-H2DCFDA |
| Ex/Em (nm) | 644/665 | 485/520 | 495/525 |
| Live cell | Yes | Yes | Yes |
| Lysate | Yes | Yes | Yes |
| Purified enzyme | Yes | Yes | Yes |
| Usage | Cell permeant for live-cell determinations | Cell permeant for live-cell determinations | Cell permeant for live-cell determinations |
| Components | Bulk reagent | Bulk reagent | Bulk reagent |
| Format | 5 x 50 µL | 5 x 50 µL | 20 x 50 µg |
| Protocol outline |
|
|
|
| Cat. No. | C10422 | C10444 | C6827 |
| DAF-FM (4-Amino-5-Methylamino-2′,7′-Difluorofluorescein) | DAF-FM Diacetate (4-Amino-5-Methylamino-2′,7′-Difluorofluorescein Diacetate) | ThiolTracker Violet (Glutathione Detection Reagent) | |
|---|---|---|---|
| Target | Nitric oxide | Nitric oxide | Glutathione |
| Reporter | DAF-FM | DAF-FM | ThiolTracker Violet |
| Ex/Em (nm) | 495/515 | 495/515 | 405/526 |
| Live cell | No | Yes | Yes |
| Lysate | Yes | Yes | Yes |
| Purified enzyme | Yes | Yes | Yes |
| Usage | Lower detection limit ~3 nM of NO | Lower detection limit ~3 nM of NO | Cell permeant for live-cell determinations |
| Components | Bulk reagent | Bulk reagent | Bulk reagent |
| Format | 1 mg | 1 mg | 500 assays using 100 µL reaction volume |
| Protocol outline |
|
| |
| Cat. No. | D23841 | D23842 | T10096 |
| Amplex Red Hydrogen Peroxide/Peroxidase Assay Kit (500 assays) | APF (hydroxyl radical, hypochlorite or peroxynitrite sensor) | HPF (hydroxyl radical and peroxynitrite sensor) | MitoSOX Red Mitochondrial Superoxide Indicator, for live-cell imaging | Dihydroethidium (hydroethidine) | |
|---|---|---|---|---|---|
| Target | Hydrogen peroxide/Peroxidase | Use singly for general ROS detection; in tandem can be used to detect hypochlorous acid (HOCl) | Targeted to mitochondria | Blue fluorescent dye becomes red when oxidized | |
| Reporter | Amplex Red reagent | ARF | HPF | MitoSOX Red | Dihydroethidium (DHE) |
| Ex/Em (nm) | 571/585 | 490/515 | 510/580 | 518/606 | |
| Live cell | No | Yes | Yes | Yes | |
| Lysate | Yes | No | No | No | |
| Purified enzyme | Yes | No | NO | No | |
| Usage | Lower detection limit 1 x 10–5 U/ml HRP | Both reagents become fluorescent on reaction with ROS–APF exhibits higher response than HPF to hypochlorite anions | Detects increases in cellular superoxide production | Exhibits blue-fluorescence in the cytosol until oxidized, where it intercalates within the cell's DNA | |
| Components | Includes assay buffers and controls | Bulk reagent | Bulk reagent | Bulk reagent | Bulk reagent |
| Format | 500 assays using 100 µL reaction volume | 470 µL | 470 µL | 10 x 50 µg | 10 x 1 mg |
| Protocol outline |
|
|
|
|
|
| Cat. No. | A22188 | A36003 | H36004 | M36008 | D11347 |
| EnzChek Myeloperoxidase (MPO) Activity Assay Kit | |
|---|---|
| Target | Continuously detects chlorination and peroxidation activity of myeloperoxidase (MPO) |
| Reporter | Amplex UltraRed |
| Ex/Em (nm) | 571/585 |
| Live cell | No |
| Lysate | Yes |
| Purified enzyme | Yes |
| Usage | Measure 1.5 to 200 ng/mL MPO |
| Components | Includes assay buffers and controls |
| Format | 200 assays using 100 µL reaction volume |
| Protocol outline |
|
| Cat. No. | E33856 |
High-sensitivity fluorescence detection for 6-1,536 samples can be quickly performed on the Varioskan ALF or Varioskan LUX Multimode Microplate Reader using Invitrogen reagents to enable optimal detection. Take advantage of automatic dynamic range selection to get optimal gain settings for each individual well and automation capabilities for even higher throughput.
For Research Use Only. Not for use in diagnostic procedures.