Search
Mouse cell isolation |
Gentle tube-based isolation of mouse cells from directly from whole blood, spleen, lymph node or thymus. For any downstream assay, including flow cytometry.
If you can't find a ready-to-use product for your specific cell type or cell subset, then select from our range of products for use with your own antibody:
Streptavidin Dynabeads | Secondary coated Dynabeads | Surface activated Dynabeads
| Cell type/ Subset | Product | Starting sample | Isolation technique | Main benefits / Applications |
|---|---|---|---|---|
Dendritic Mouse Cells | ||||
| All subsets | Dynabeads Mouse DC Enrichment Kit | Single cell suspensions | Negative Isolation | Reduce sorting time by typically 60-90% compared to sorting only. Reduce the need of fluorescent antibody for labeling of cells for your sorting by a factor of 6-14. This kit retains all DC subpopulations. |
Our Streptavidin Dynabeads, Secondary coated Dynabeads and Surface activated Dynabeads can be combined with your own antibody to isolate any cell type. Choose your preferred product in the below table.
| Product | Starting sample | Main benefit / Applications |
|---|---|---|
Streptavidin Dynabeads | ||
| Dynabeads FlowComp Flexi | Single cell suspensions | Combine with your own antibody of choice to isolate any cell type from any species. This kit provides everything you need for labeling of antibodies and subsequent cell isolation. Beads are removed after isolation to obtain pure and viable cells suitable for any downstream application. |
| CELLection Biotin Binder | Single cell suspensions | Use with any biotinylated antibody to positively isolate cells from any species. Remove beads after isolation to obtain pure and viable cells for use in any application. Read more about cell isolation using biotinylated antibodies. |
| Dynabeads Biotin Binder | Single cell suspensions | Isolate any cell from any species. This kit binds all biotinylated antibodies and is our most flexible secondary product for cell isolation or depletion. Read more about cell isolation using biotinylated antibodies. |
Secondary Coated Dynabeads | ||
| Dynabeads Sheep anti-Rat IgG | Single cell suspensions | Isolate any cell type from all species except rat. For use with your primary monoclonal rat antibody of choice- Ideal for positive isolation, negative isolation or depletion of cells. |
| Dynabeads Sheep anti-Rabbit IgG | Single cell suspensions | Isolate any cell type from any species except rabbit. For use with your primary rabbit antibody of choice. This product can be used for eukaryotic cells. |
Surface Activated Dynabeads | ||
| Dynabeads M-450 Epoxy | Single cell suspensions | Hydrophobic naked beads with glycidyl ether (epoxy) reactive groups. Ideal for coupling of antibodies for cell applications. Coupling reactions can be performed at neutral pH, with 24 hour incubation at room temperature. |
| Dynabeads M-450 Tosylactivated | Single cell suspensions | Hydrophobic naked beads with p-toluenesulphonyl (tosyl) reactive groups. For coupling of antibodies for cell applications. Coupling reactions can be performed at neutral pH although high pH and incubation at 37°C will promote covalent binding. |
Click here for recommended sample preparation procedures.
For Research Use Only. Not for use in diagnostic procedures.