1508251251223
Applied Biosystems TaqMan Pri-miRNA Assays allow researchers to quantitate primary miRNA transcripts with specificity and high sensitivity. Together with a standard reverse transcription kit and a qPCR master mix, these TaqMan Pri-miRNA Assays will enable researchers the opportunity to study miRNA biogenesis.
  • Highly specific―quantitates primary miRNA transcripts and not mature miRNAs
  • Fast, simple, and scalable―two-step quantitative RT-PCR assay provides high quality results in less than three hours
  • Designed using the same verified pipeline as TaqMan Gene Expression Assays
  • Primers and probe are located near the stem loop of interest
Already know your target of interest? Search for a pre-designed TaqMan pre-miRNA Assays using the tool below.
Assay search tool - find & buy your single tube TaqMan Assays:
Need to design your own custom TaqMan Pri-miRNA assay?  Use our custom assay design tool.
Looking for a pre-designed TaqMan Pri-miRNA assay with a dye other than FAM?  Please contact our Specialty TaqMan Assays and Oligos team.
Specifications
Species
TaqMan Pri-miRNA Assays are available for a wide range of species.
Assay components
TaqMan Pri-miRNA Assays are delivered in a single tube containing the specific pre-formulated TaqMan Assay (TaqMan MGB probe labeled with FAM, and forward and reverse primers).
Assay suffix nomenclature
_pri indicates a primary miRNA assay and not a mature miRNA assay.
Available sizes
Our TaqMan Pri-miRNA Assays are available in three sizes:
Cat. No. 4427012: Small (360 reactions, 20X, made to order)
Cat. No. 4427013: Medium (750 reactions, 20X, made to order)
Cat. No. 4427014: Large (2900 reactions, 20X, made to order)
Shipping and storage
Shipped at ambient temperature.
Store at –15ºC to –25ºC.
White Paper: Ambient Temperature Shipping Reduces Environmental Impact and Retains Quality and Stability
Related categories, applications, and data analysis tools
Product categories and applications
Applied Biosystems qPCR analysis modules