Search
Having trouble resolving your compounds using your C18 column? The new Thermo Scientific Accucore Biphenyl LC column is your solution for:
Now you can stop struggling with unresolved peaks, long run times, and poor lot-to-lot consistency. Obtain unique selectivity for aromatic and moderately polar analytes using Accucore Biphenyl LC Columns. These columns have optimized packing, offering a rugged platform for a variety of matrices.
Find out more!
| Catalog | Description | Diameter (metric) | Length (metric) | Particle size | |
|---|---|---|---|---|---|
| 17826-052130 | Accucore Biphenyl Analytical Column 2.1x50mm | 2.1mm | 50mm | 2.6µm | Order |
| 17826-102130 | Accucore Biphenyl Analytical Column 2.1x100mm | 2.1mm | 100mm | 2.6µm | Order |
| 17826-012105 | Accucore Defender Guard Cartridges 4/pk | 2.1mm | 10mm | 2.6µm | Order |
Generally, C18 columns offer the ability to resolve a wide variety of analytes without issue. However, there are times when alternative chemistry is required for the separation of aromatic and moderately polar analytes. A biphenyl chemistry offers complementary selectivity to a C18 column, and the ability to resolve isomeric compounds, such as drugs of abuse and steroids by taking advantage of π-π interactions between the analyte and the stationary phase. This can help pull critical peaks out of matrix interferences.
Our solid core column is designed based around voice of customer, addressing critical needs for customers in clinical research, forensic toxicology, pharmaceutical, and environmental labs. We have demonstrated outstanding column ruggedness and lot-to-lot reproducibility on this phase, optimizing critical parameters such as retention time stability, pressure, and peak asymmetry. Data quality should not be a concern.
We performed a ballistic gradient for over 3000 injections on an acidic, neutral, and basic compound. The data quality remained steady.
Further flexing its strength, the Accucore Biphenyl can separate a mix of steroid compounds in under 4 minutes, saving run time on instrumentation, and helping to improve sample throughput.
Generally, C18 columns offer the ability to resolve a wide variety of analytes without issue. However, there are times when alternative chemistry is required for the separation of aromatic and moderately polar analytes. A biphenyl chemistry offers complementary selectivity to a C18 column, and the ability to resolve isomeric compounds, such as drugs of abuse and steroids by taking advantage of π-π interactions between the analyte and the stationary phase. This can help pull critical peaks out of matrix interferences.
Our solid core column is designed based around voice of customer, addressing critical needs for customers in clinical research, forensic toxicology, pharmaceutical, and environmental labs. We have demonstrated outstanding column ruggedness and lot-to-lot reproducibility on this phase, optimizing critical parameters such as retention time stability, pressure, and peak asymmetry. Data quality should not be a concern.
We performed a ballistic gradient for over 3000 injections on an acidic, neutral, and basic compound. The data quality remained steady.
Further flexing its strength, the Accucore Biphenyl can separate a mix of steroid compounds in under 4 minutes, saving run time on instrumentation, and helping to improve sample throughput.
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.