Search
Want a C18 column that's right for your application? Thermo Scientific C18 columns are available with a range of characteristics to provide exceptional performance. These columns are broadly used in a wide variety of applications including pharmaceuticals, steroids, fatty acids, phthalates, environmental and more. They are the most retentive of the alkyl-bonded phases.
Thermo Scientific Column | Recommended Use |
|---|---|
| Accucore Vanquish C18+ UHPLC Columns, 1.5µm | Meet high throughput demands in your analytical laboratory. This newest innovation in UHPLC columns delivers better, more and easier separations at reduced cost. |
| Acclaim 120 C18 Columns | This high resolution silica column can improve the separation efficiency in your reversed phase applications. |
| Accucore aQ C18 Polar Endcapped LC Columns | Separate polar compounds with improved speed and resolution at low backpressures using the column’s rugged 2.6µm solid-core particles. |
| Accucore C18 LC Columns | Get optimal retention of a broad range of nonpolar analytes across multiple applications with a column that has high bonded phase coverage. |
| Accucore RP-MS LC Columns | Optimized for MS detection, these clumns are an excellent combination of speed and quality of separation. |
| Accucore XL C18 LC Columns | Optimize conventional HPLC methodsof nonpolar analytes. The rugged 4µm solid-core particles ensure very high efficiencies within standard HPLC pressure limits. |
| Hypersil GOLD aQ C18 Polar Endcapped LC Columns | Retain and resolve polar analytes. These columns are ideal for use with highly aqueous mobile phases. |
| Hypersil GOLD C18 Selectivity LC Columns | Achieve exceptional peak shape and resolution for your HPLC and LC-MS applications with these endcapped, ultrapure, silica-based columns. |
| Hypersil BDS C18 Columns | A robust, general-purpose column to use in a wide range of applications. |
| Hypersil ODS C18 Columns | A general purpose C18 phase to use for a broad range of applications. |
| Hypersil ODS-2 C18 Columns | Use this well-tested, dependable L1 as an alternative to older column brands commonly referenced in validated methods. |
| Syncronis aQ C18 Polar Endcapped LC Columns | Ensure consistent, predictable liquid chromatography (LC) separations of polar analytes. |
| Syncronis C18 Columns | Maximize reproducibility and laboratory productivity. |
Dafydd Milton introduces new Accucore HPLC column technology that enables high-speed, high-resolution separations with backpressures lower than those associated with UHPLC.
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.
Discover live and on-demand webinars that enable you to improve productivity, maintain compliance, and learn about the constantly evolving scientific landscape.