Search Thermo Fisher Scientific
Sample type | Saliva |
Sample input volume | 200 µL–2 mL |
Yield | 100ng/µL saliva on average (yield is dependent on each individual donor) |
Elution volume | 50 µL |
Total processing time | 25 min total time (5 min hands-on time) |
Reactions | 100 rxn, 500 rxn |
Shipping condition | Room temperature |
Cat. No. | A39059, A39060 |
Use this kit with KingFisher automation to speed up your processing of saliva samples for genetic testing. Recover DNA that is compatible with Next-Generation Sequencing (NGS) and array platforms, from 96 samples, in 25 minutes.
Why purchase multiple purification kits to isolate DNA present in your saliva sample types? Instead use our MagMAX Saliva gDNA Isolation Kit, fully scalable for the isolation of genomic DNA (gDNA) from fresh and stabilized saliva. With magnetic bead-based technology at its core, the result is high yields of purified gDNA. Post-sample preparation, downstream molecular biology applications include qPCR, microarray analysis, and next-generation sequencing. All components and reagents are available for purchase separately.
Extract gDNA from 200 µL–2 mL of stabilized or fresh saliva properly collected using the the MagMAX Saliva gDNA Isolation Kit. The workflow is compatible with saliva collection devices like the Oragene® from Genotek, the GeneFiX™ from IsoHelix, and the SalivaGard® from Biomatrica.
Simultaneously lyse cells, denature proteins, and capture gDNA on high-capacity magnetic binding beads with an innovative lysis/binding solution in the MagMAX Saliva gDNA Isolation Kit. Remove proteins, detergents, and other impurities from the bound nucleic acid with the Saliva gDNA Wash I Solution. Released the extracted gDNA from the beads using a low-TE elution solution.
Combine multiple time-consuming sample processing steps in one single step with the MagMAX Saliva gDNA Isolation Kit. Magnetic bead-based technology reduces the laborious tasks often associated with other methods of nucleic acid extraction. Process a wide range of sample volume inputs concurrently without sample normalization or reagent volume adjustments. Simply match your saliva input volumes to the standard input workflows (50 µL to 500 µL, up to 1 mL, and up to 2 mL) and follow the directions for semi-automated sample preparation using the KingFisher Duo Prime or KingFisher Flex purification systems.
The MagMAX Saliva gDNA Isolation Kit is optimized for moderate-throughput workflows using KingFisher Duo Prime while KingFisher Flex is used for higher-throughput sample processing. Process up to 500 µL of saliva in high-throughput capacity SBS 96-well deep-well microtiter plates with KingFisher Flex system, or smaller sample batches of 12 using the Duo Prime system. Dealing with larger sample input volumes for >500 µL and 1–2 mL? Use the higher capacity 24-well deep-well SBS plates for 24 samples on the Flex or 6 samples on the Duo Prime system. BindIt software scripts for both KingFisher instrument Flex and Duo Prime can be downloaded and installed on the instruments.
The MagMAX Saliva gDNA Isolation kit has a total processing time of 25 minutes, including 5 minutes hands-on time compared to other supplier kits which typically take anywhere from 30+ minutes to 50 minutes for processing.
Figure 1. Fast processing time compared to competitor kits.
Figure 4. Compatibility with saliva collection devices. 500 µL of stabilized saliva collected using the Oragene™ Discover, GeneFix™, or SalivaGard™ saliva collection devices was used as sample input for the MagMAX Saliva gDNA Isolation Kit. Total yield and concentration were analyzed using the Qubit Broad Range Assay or the RNAseP Verification Assay to quantitate genomic copies of the human RNaseP gene.
Figure 5. Performance compared to Competitor Q saliva gDNA isolation kit. A MagMAX Saliva gDNA Isolation Kit and Supplier Q recommended saliva gDNA isolation kit were used to isolate gDNA from 500 µL of saliva from six different donors. Total gDNA yield was determined using the Qubit dsDNA Broad Range Assay. Purity was determined by calculating the A260/A280 ratio using a Nanodrop spectrophotometer.
Figure 6. Performance compared to competitor saliva gDNA isolation kits. A MagMAX Saliva gDNA Isolation Kit and Supplier P and Supplier N recommended saliva gDNA isolation kits were used to isolate gDNA from 500 µL of saliva from six different donors. Total gDNA yield was determined using the Qubit dsDNA Broad Range Assay. TaqMan Assays for human actin B (ACTB) and GAPDH genes were used to assess the ability of the gDNA to amplify in qPCR reactions. The MagMAX Saliva gDNA Isolation Kit provided the lowest CT values for GAPDH across all six donors, indicating the presence of more amplifiable gDNA in samples prepared with the MagMAX kit. Purity was determined by calculating the A260/A280 ratio using a Nanodrop spectrophotometer.
Figure 6. Number of preps per kit. The MagMAX Saliva gDNA isolation kit will scale in proportion to the volume of saliva to be processed. The table shows the number of 500 mL preparations of saliva that can be processed with each kit format or stand-alone reagents.
Protocol Type | KingFisher Duo | KingFisher Flex |
---|---|---|
Saliva 2 mL | KingFisher Duo Protocol - MagMAX Saliva - 2 ml (.bdz) | KingFisher Flex Protocol - MagMAX Saliva - 2 mL (.bdz) |
Saliva 500 µL | KingFisher Duo Protocol - MagMAX Saliva - 500 µL (.bdz) | KingFisher Flex Protocol - MagMAX Saliva - 500 µL (.bdz) |
Sample type | Saliva |
Sample input volume | 200 µL–2 mL |
Yield | 100ng/µL saliva on average (yield is dependent on each individual donor) |
Elution volume | 50 µL |
Total processing time | 25 min total time (5 min hands-on time) |
Reactions | 100 rxn, 500 rxn |
Shipping condition | Room temperature |
Cat. No. | A39059, A39060 |
Use this kit with KingFisher automation to speed up your processing of saliva samples for genetic testing. Recover DNA that is compatible with Next-Generation Sequencing (NGS) and array platforms, from 96 samples, in 25 minutes.
Why purchase multiple purification kits to isolate DNA present in your saliva sample types? Instead use our MagMAX Saliva gDNA Isolation Kit, fully scalable for the isolation of genomic DNA (gDNA) from fresh and stabilized saliva. With magnetic bead-based technology at its core, the result is high yields of purified gDNA. Post-sample preparation, downstream molecular biology applications include qPCR, microarray analysis, and next-generation sequencing. All components and reagents are available for purchase separately.
Extract gDNA from 200 µL–2 mL of stabilized or fresh saliva properly collected using the the MagMAX Saliva gDNA Isolation Kit. The workflow is compatible with saliva collection devices like the Oragene® from Genotek, the GeneFiX™ from IsoHelix, and the SalivaGard® from Biomatrica.
Simultaneously lyse cells, denature proteins, and capture gDNA on high-capacity magnetic binding beads with an innovative lysis/binding solution in the MagMAX Saliva gDNA Isolation Kit. Remove proteins, detergents, and other impurities from the bound nucleic acid with the Saliva gDNA Wash I Solution. Released the extracted gDNA from the beads using a low-TE elution solution.
Combine multiple time-consuming sample processing steps in one single step with the MagMAX Saliva gDNA Isolation Kit. Magnetic bead-based technology reduces the laborious tasks often associated with other methods of nucleic acid extraction. Process a wide range of sample volume inputs concurrently without sample normalization or reagent volume adjustments. Simply match your saliva input volumes to the standard input workflows (50 µL to 500 µL, up to 1 mL, and up to 2 mL) and follow the directions for semi-automated sample preparation using the KingFisher Duo Prime or KingFisher Flex purification systems.
The MagMAX Saliva gDNA Isolation Kit is optimized for moderate-throughput workflows using KingFisher Duo Prime while KingFisher Flex is used for higher-throughput sample processing. Process up to 500 µL of saliva in high-throughput capacity SBS 96-well deep-well microtiter plates with KingFisher Flex system, or smaller sample batches of 12 using the Duo Prime system. Dealing with larger sample input volumes for >500 µL and 1–2 mL? Use the higher capacity 24-well deep-well SBS plates for 24 samples on the Flex or 6 samples on the Duo Prime system. BindIt software scripts for both KingFisher instrument Flex and Duo Prime can be downloaded and installed on the instruments.
The MagMAX Saliva gDNA Isolation kit has a total processing time of 25 minutes, including 5 minutes hands-on time compared to other supplier kits which typically take anywhere from 30+ minutes to 50 minutes for processing.
Figure 1. Fast processing time compared to competitor kits.
Figure 4. Compatibility with saliva collection devices. 500 µL of stabilized saliva collected using the Oragene™ Discover, GeneFix™, or SalivaGard™ saliva collection devices was used as sample input for the MagMAX Saliva gDNA Isolation Kit. Total yield and concentration were analyzed using the Qubit Broad Range Assay or the RNAseP Verification Assay to quantitate genomic copies of the human RNaseP gene.
Figure 5. Performance compared to Competitor Q saliva gDNA isolation kit. A MagMAX Saliva gDNA Isolation Kit and Supplier Q recommended saliva gDNA isolation kit were used to isolate gDNA from 500 µL of saliva from six different donors. Total gDNA yield was determined using the Qubit dsDNA Broad Range Assay. Purity was determined by calculating the A260/A280 ratio using a Nanodrop spectrophotometer.
Figure 6. Performance compared to competitor saliva gDNA isolation kits. A MagMAX Saliva gDNA Isolation Kit and Supplier P and Supplier N recommended saliva gDNA isolation kits were used to isolate gDNA from 500 µL of saliva from six different donors. Total gDNA yield was determined using the Qubit dsDNA Broad Range Assay. TaqMan Assays for human actin B (ACTB) and GAPDH genes were used to assess the ability of the gDNA to amplify in qPCR reactions. The MagMAX Saliva gDNA Isolation Kit provided the lowest CT values for GAPDH across all six donors, indicating the presence of more amplifiable gDNA in samples prepared with the MagMAX kit. Purity was determined by calculating the A260/A280 ratio using a Nanodrop spectrophotometer.
Figure 6. Number of preps per kit. The MagMAX Saliva gDNA isolation kit will scale in proportion to the volume of saliva to be processed. The table shows the number of 500 mL preparations of saliva that can be processed with each kit format or stand-alone reagents.
Protocol Type | KingFisher Duo | KingFisher Flex |
---|---|---|
Saliva 2 mL | KingFisher Duo Protocol - MagMAX Saliva - 2 ml (.bdz) | KingFisher Flex Protocol - MagMAX Saliva - 2 mL (.bdz) |
Saliva 500 µL | KingFisher Duo Protocol - MagMAX Saliva - 500 µL (.bdz) | KingFisher Flex Protocol - MagMAX Saliva - 500 µL (.bdz) |
Just like all Applied Biosystems MagMAX kits, the gDNA saliva kits are optimized for KingFisher instruments.
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.