Thermo Scientific Slide-A-Lyzer G2 Dialysis Cassettes provide convenience, flexibility, and performance for sample dialysis. These second-generation (G2) Slide-A-Lyzer Dialysis Cassettes are free-standing, self-floating, and pipette-loadable.
Sample loading and removal are easily accomplished using a serological pipette or hypodermic needle (optional) attached to a syringe (Figure 1). The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis. Slide-A-Lyzer G2 Dialysis Cassettes are available in 5 volume capacities and in 2K, 3.5K, 7K, 10K, and 20K molecular weight cut offs (MWCOs).
Choose the right Slide-A-Lyzer G2 cassette for your experiments
![]() |
![]() |
![]() |
![]() |
![]() |
|
---|---|---|---|---|---|
Volume range | |||||
Molecular weight cutoff | 0.25–0.5 mL | 1–3 mL | 5–15 mL | 10–30 mL | 25–70 mL |
2K | Catalog number 87717 |
Catalog number 87718 |
Catalog number 87719 |
Catalog number 87720 |
Catalog number 87721 |
3.5K | Catalog number 87722 |
Catalog number 87723 |
Catalog number 87724 |
Catalog number 87725 |
Catalog number 87726 |
7K | Catalog number 87727 |
Catalog number 87728 |
Not available | Not available | Not available |
10K | Catalog number 87729 |
Catalog number 87730 |
Catalog number 87731 |
Catalog number 87732 |
Catalog number 87733 |
10K (gamma irradiated) |
Catalog number 88250 |
Catalog number 88251 |
Catalog number 88252 |
Catalog number 88253 |
Catalog number 88254 |
20K | Catalog number 87734 |
Catalog number 87735 |
Catalog number 87736 |
Catalog number 87737 |
Catalog number 87738 |
Learn more about Slide-A-Lyzer G2 Dialysis Cassettes
For Research Use Only. Not for use in diagnostic procedures.