Search
Applied Biosystems TaqMan Pre-miRNA Assays allow researchers to quantitate precursor miRNAs with specificity and high sensitivity. When used with a standard reverse transcription kit and a qPCR master mix, these assays help researchers in the study miRNA biogenesis.
Already know your target of interest? Download a list of all pre-designed TaqMan pre-miRNA assays.
Want to design your own custom TaqMan pre-miRNA assay? Use our gene expression custom assay design tool.
Species | TaqMan Pre-miRNA Assays are available for a wide range of species. |
Assay components | TaqMan Pre-miRNA Assays are delivered in a single tube containing the specific pre-formulated TaqMan assay (TaqMan MGB probe, forward and reverse primers). |
Assay suffix nomenclature | _s1: An assay whose primers and probes are designed within a single exon. Such assays, by definition, detect genomic DNA. |
Available sizes | Our TaqMan Pre-miRNA Assays are categorized under Non-Coding RNA Assays, as well as Gene Expression Assays resulting in various catalog numbers. Please search for your precursor miRNA using our gene expression assay search tool to locate the appropriate catalog number. TaqMan Non-Coding RNA Assay TaqMan Gene Expression Assay |
Available assays | Complete list of TaqMan pre-miRNA Assays |
Shipping and storage | Shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.