Search
This resource is designed to provide researchers with insights and practical guidance on leveraging high content imaging techniques to analyze neuronal cultures grown in both two-dimensional (2D) and three-dimensional (3D) environments. By using optimized media and advanced imaging technologies, you can achieve precise and detailed visualization of neuronal structures and functions, facilitating a deeper understanding of neural development, connectivity, and responses to various treatments. Explore detailed protocols, recommendations, and best practices to help enhance your imaging and analysis workflows.
Neuronal cultures are laboratory-based systems where neurons are grown and maintained outside of their natural environment to study their behavior, function, and interactions in a controlled setting. There are various types of cell culture, including primary culture (derived directly from tissues), secondary culture (subcultured from primary cells), and cell lines (immortalized cells that can be cultured indefinitely). Neuronal cells are specialized cells within the nervous system, including neurons, which transmit nerve impulses, and glial cells, which provide support and protection for neurons. To prepare neuronal cell cultures, neurons are typically isolated from embryonic or neonatal brain tissue, dissociated into single cells, and then plated onto culture dishes coated with substrates like poly-D-lysine or laminin to promote cell adhesion. These cultures are maintained in a specialized growth medium that supports neuronal survival and differentiation, enabling researchers to investigate neural development, synaptic function, and neurodegenerative diseases in a controlled environment.
The Thermo Scientific CellInsight CX7 LZR HCA Platform is a 7-laser high-content imaging instrument that reduces scanning times and improves axialresolution in 2D and 3D cultures. These features provide the speed and sensitivity necessary to analyze neuronal culture systems for more physiologicallyrelevant drug discovery assays, and allow tracking and quantification of the dynamic changes in neurites and synapses underlying synaptogenesis orsynaptic damage. Here we discuss use of the new-generation Gibco B-27 Plus Neuronal Culture System to generate 2D and 3D neuronal cultures withenhanced growth. These neuronal cultures can then be stained with a variety of fluorescent markers and probes for high-throughput imaging so thatneuronal growth, length, and toxicity can be quantified using HCA, allowing data to be obtained from hundreds of neuronal samples over a short period of time.
Cryopreserved rat cortical or hippocampal neurons were cultured in the B-27 system (Neurobasal Medium with B-27 Supplement) or B-27 Plus system (Neurobasal Plus Medium with B-27 Plus Supplement) for 3–4 weeks. The cells were fixed with 4% formaldehyde for 15 min, permeabilized with 0.5% Triton X-100 detergent for 10 min, and then immunostained with HuC/HuD and MAP2 primary antibodies followed by Alexa Fluor 555 goat anti-mouse and Alexa Fluor 488 goat anti-rabbit secondary antibodies, respectively. The samples were imaged on a CellInsight CX7 LZR HCA Platform.
Cryopreserved rat cortical neurons were grown for 2 days and then treated with cadmium chloride or paclitaxel for 24 hr. The cells were then fixed and stained with HuC/HuD and MAP2 primary antibodies followed by Alexa Fluor 488 goat anti-mouse and Alexa Fluor 647 goat anti-rabbit secondary antibodies, respectively. The samples were imaged on a CellInsight CX7 LZR HCA Platform.
Neural stem cells (NSCs) were grown on Nunclon Sphera U-bottom plates (1.3 x 104 cells/well) to form neurospheres. The cells were differentiated in the B-27 system or B-27 Plus system, each with or without CultureOne Supplement. N-2 Supplement, growth factors BDNF, GDNF, and CNTF (all 10 μg/mL), and 10 μM ROCK inhibitor (Y-27632) were also added to the cultures. The neurons were differentiated for 2 weeks and stained with 1 μM Tubulin Tracker Deep Red for 1 hr. The spheroids were then washed with Live Cell Imaging Solution and imaged on the CellInsight CX7 LZR HCA Platform.
The neuronal cells were analyzed on a CellInsight CX7 LZR HCA Platform using the neuronal profiling bioapplication at 10x magnification. A total of 200 cells were imaged per well, and neuronal cell bodies and neurite outgrowth were measured using their respective parameters. The nuclei were segmented with DAPI, and neuronal cell bodies were identified with HuC/HuD-positive staining. Neurite length was measured with MAP2 staining. For 3D neurosphere imaging, confocal mode was used, where each image represents a maximum intensity projection (MIP) of thirty Z optical slices of 10 μm each.
2D neuronal cultures immunostained with MAP2 and HuC/HuD antibodies to label dendrites and neuronal cell bodies were imaged on the CellInsight CX7 LZR HCA Platform and analyzed with Thermo Scientific HCS Studio 2.0 Cell Analysis Software to obtain measurements of neurons per field and neurite length. Both rat cortical and hippocampal neurons had increased growth, as indicated by neurons per field, when grown in the B-27 Plus Neuronal Culture System compared to the original B-27 system (Figure 1).
Neuronal toxicity can also be quantitated by imaging and analysis on the CellInsight CX7 LZR HCA Platform and HCS Studio 2.0 Cell Analysis Software as demonstrated by treating rat cortical neurons with 10 mM cadmium chloride or 10 μM paclitaxel. Both treatments significantly decreased the total neurite length per field and percentage of neurons positive for HuC/HuD, demonstrating the neuronal toxicity of these compounds (Figure 2).
Figure 1. The B-27 Plus Neuronal Culture System enables superior growth of rodent primary cortical and hippocampal neurons, compared to the original B-27 system. Cryopreserved neurons were cultured for 3–4 weeks using the first-generation B-27 system (Neurobasal Medium with B-27 Supplement) or B-27 Plus system (Neurobasal Plus Medium with B-27 Plus Supplement). Neurons were fixed and immunostained for the neuronal dendritic marker MAP2 (green) and the neuronal cell body marker HuC/HuD (red). Fluorescence imaging and quantitation of neurons per field were performed on the CellInsight CX7 LZR HCA Platform. Both cultures have more neurons per field when grown in the B-27 Plus system than in the original B-27 system.
Figure 2. Monitoring neuronal toxicity with the CellInsight CX7 LZR HCA Platform. Rat cortical neurons were cultured for 2 days in a 96-well plate and treated with 10 mM cadmium chloride or 10 μM paclitaxel for 24 hr. Neurons were then fixed and immunostained for the neuronal dendritic marker MAP2 (red) and neuronal cell body marker HuC/HuD (green). Fluorescence imaging and quantitation were performed on the CellInsight CX7 LZR HCA Platform in confocal mode. Segmentation of the neuronal processes allows quantitation of total neurite length per field and percentage of neurons positive for HuC/HuD. Treatment with cadmium or paclitaxel significantly decreased the total neurite length per field and percentage of HuC/HuD-positive neurons, indicating that both treatments were toxic.
Neurons were also grown in 3D culture to form neurospheroids that were stained with Tubulin Tracker Deep Red to label neuronal processes in live cells without the need for fixation or immunostaining. As demonstrated by staining with Tubulin Tracker Deep Red and imaging on the CellInsight CX7 LZR HCA Platform, the 3D neurospheroids showed enhanced growth when grown in the B-27 Plus system with CultureOne Supplement, compared to neurospheroids grown in the original B-27 system, or without the supplement (Figure 3).
Figure 3. The B-27 Plus system with CultureOne Supplement enhances growth of neurospheroids in 3D culture. Neurospheroids were grown in the B-27 system or B-27 Plus system, each with or without CultureOne Supplement, on Nunclon Sphera U-bottom plates and stained with Tubulin Tracker Deep Red. Imaging of the stained neurospheroids on the CellInsight CX7 LZR HCA Platform shows enhanced neurite growth of the 3D neuronal cultures when grown in the B-27 Plus system with CultureOne Supplement.
High-content imaging and analysis with the CellInsight CX7 LZR HCA Platform and HCS Studio 2.0 Cell Analysis Software enable fast, high-resolution imaging of both 2D and 3D neuronal cell cultures along with automated quantitation and complex analysis of several parameters of neuronal growth and health, including identification of neuronal cell bodies, neurons per field, and neurite length. The B-27 Plus Neuronal Culture System allows superior growth of 2D and 3D neuronal cultures over longer periods of time than the original B-27 culture system. Tubulin Tracker Deep Red, a docetaxel-based fluorescent reagent for live-cell labeling of microtubules, provides neuronal labeling throughout live 3D neurospheroids without the need for fixation or immunostaining. These optimized neuronal culture systems and fluorescent staining reagents combined with high-resolution automated imaging and analysis allow for investigation of neuronal growth and toxicity of hundreds of samples over a relatively short period of time, helping to enable new discoveries in neuronal function and neuropharmacology. These discoveries may potentially be applied to understanding of the nervous system and treatment of neurological disorders.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Cell Culture Selection Tools
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support?
Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.