WPR2150_Gene-Expression-AGUC
Comprehensive solutions for gene expression profiling, verification, and research screening
Gene expression profiling simultaneously compares the expression levels of multiple genes between two or more samples. This analysis can help scientists identify the molecular basis of phenotypic differences and select gene expression targets for in-depth study. Gene expression profiling provides valuable insight into the role of differential gene expression in normal biological and disease processes.
NEW! Technical Note demonstrates High concordance of data between Gene Expression platforms, allowing the ability to switch without having to repeat experiments
Download now
Roberts
NEW! Scientist Spotlights in Gene Expression Analysis
Learn from key opinion leaders in the field of genomics as they discuss their research interests. Discover how our cross-technology solutions can help you in your gene expression analysis needs.
View Scientist Spotlights ›
Explore our gene expression solutions and technologies
Gene expression analysis by real-time PCR
Real-time PCR is the gold-standard technique for verification of differential gene expression profiles. Our comprehensive offering of TaqMan Gene Expression Assays and flexible formats enables fast and cost-effective results.
Gene expression profiling by sequencing
Transcriptome sequencing by next-generation sequencing enables the discovery of differentially expressed genes using a hypothesis-neutral approach.         
Gene expression profiling by microarray
Microarrays are designed with up-to-date content from public sequence data sources, and enable fast, detailed transcriptome-wide expression profiling, even from challenging and precious samples with no bioinformatics resources required.
Gene expression profiling by hybridization-based assay
Multiplex hybridization-based assays utilize a branched DNA technology for signal amplification for the direct quantitation of gene expression transcripts. These assays are designed to quantitatively measure RNA directly from cell or tissue homogenates without RNA purification or enzymatic manipulation.
Gene expression quantification by digital PCR
Digital PCR enables highly precise measurements and is able to resolve gene expression level changes of two-fold or less. Digital PCR can also be used to determine the absolute quantification of a transcript without the need for a reference gene.