Invitrogen GeneArt services are a suite of solutions for producing synthetic genes and DNA constructs for a wide range of applications, from basic research to advanced therapeutic development. These services include gene synthesis, cloning services, and protein expression in mammalian systems, enabling researchers to design and create specific genetic sequences tailored to their experimental needs. GeneArt services aid in accelerating research timelines and improving the reliability of genetic constructs.
The GeneArt Services Dashboard, our proprietary online ordering platform, integrated with Invitrogen GeneOptimizer technology, offers optimized gene design and enhanced protein expression.
Explore the GeneArt Services selection guide
GeneArt DNA optimization
The GeneArt GeneOptimizer algorithm goes beyond basic codon optimization to help improve transcription, mRNA stability, and translation to enable better protein yields.
Explore the industry-leading GeneOptimizer algorithm
GeneArt suite of services
Save up to 25% on your in-house cloning needs
Clone more. Save more.
GeneArt Services Dashboard for online ordering
GeneArt services: Selection guide
For all service options, no physical template is required. Take advantage of our industry-leading sequence optimization tool for improved expression and sequence design.
 GeneArt Strings DNA FragmentsGeneArt Cloned Gene SynthesisGeneArt Protein Expression Service
What we doSynthesis of linear dsDNA fragments up to 3 kbSynthesis of DNA of interest, cloned into standard vector or expression vector of choice (ready to use)Expression, purification, and characterization of proteins from mammalian cells, starting from only the electronic sequence
Advantages
  • Synthesis of large sequences
  • Synthesis of complex sequences (e.g., high/low GC content)
  • Downstream services available
Hands-on timeMediumLowNone
Deliverables200 ng linear dsDNA pool, dried
5 µg plasmid DNA clone, dried
Protein of interest, expression vector used for transfection, and data from standard QC and any optional analysis selected
Production time
  • 3–5 business days
  • As fast as 5 business days
  • Acceleration options available
  • Plasmid purification service starting at 3 additional business days
LengthUp to 3 kb
Up to 100 kb
N/A
Quality controlBulk sequencing100% sequence-verified
Documented analytical data options include:
  • Analytical SEC (purity)
  • UV/vis (yield)
  • SDS-PAGE or CE-SDS
  • BLI titer determination
  • nanoDSF
  • DLS
  • SEC MALS
Hero banner white gradient overlay
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments