Search
DNA Loading Buffers | Gel Loading Buffers |
We offer a variety of gel loading buffers for easy loading and tracking of DNA samples during gel electrophoresis. Invitrogen BlueJuice Gel Loading Buffer is designed for use with both agarose and polyacrylamide gels whereas TrackIt DNA loading buffers are specifically for agarose gels.
BlueJuice Gel Loading Buffer is an all-purpose DNA loading buffer and is compatible with both agarose gels (including E-Gel precast agarose gels) and native polyacrylamide gels. BlueJuice loading buffer is supplied at 10X concentration.
Invitrogen TrackIt DNA loading buffers contain two tracking dyes, one that runs behind the sample and one that runs ahead of the sample. The tracking dyes serve as visual markers for DNA migration during agarose gel electrophoresis and indicate when maximum resolution of the DNA fragments has been achieved. Additionally, visualization of DNA bands will not be obscured by the tracking dyes because they run outside the migration pattern of most DNA samples. TrackIt loading buffers are designed for loading and tracking DNA samples in agarose gels, including E-Gel precast agarose gels, and are supplied at 6X concentration.
![]() TrackIt Cyan/Orange Loading Buffer |
| Order here |
![]() TrackIt Cyan/Yellow Loading Buffer |
| Order here |
Invitrogen E-Gel Sample Loading Buffer is formulated for maximum performance on E-Gel precast gels and is supplied as a ready to-use 1X solution. This buffer contains xylene cyanol FF and tartrazine dyes.
Access learning resources—a knowledge base for all things nucleic acid electrophoresis.
View our webinars to learn more about molecular biology topics.
E-Gel Precast Gel and Ladder Selection Tool for Invitrogen Power Snap devices.
Find answers to your questions on nucleic acid gel electrophoresis using acrylamide or agarose gels, including E-gels.
For Research Use Only. Not for use in diagnostic procedures.