Search
Growth and attachment factors, natural and recombinant, are used in conjunction with other essential nutrients and hormones to maximize biological stimulation. The optimal conditions and concentrations of growth factors for cell types must be empirically determined. The 5 main types of growth factors used in cell culture are:
The following table provides a summary of conditions for growth and attachment factor supplementation reported to give maximal stimulation for various cell lines. It can be used to establish initial concentrations prior to optimization; for the best results please refer to your product’s growth factor specifications.
| Cell line | Cell type | Recommended growth factor or attachment factor | Growth factor concentration range (ng/mL) | Medium | Serum supplementation |
|---|---|---|---|---|---|
| – | Primary hepatocyte | EGF | 10 | DMEM | 10% FBS |
| – | Primary human fibroblast | EGF | 1 x 103 | MEM Earle's | 10% FBS |
| A431 | Epidermal carcinoma | EGF | 2.5 x 104 | DMEM | 10% FBS |
| A549 [4] | Lung carcinoma | EGF | 50 | Ham’s F-12K or DMEM | 10% FBS |
| BALB/c 3T3 | Mouse fibroblast | EGF | 50 | DMEM/F-12 | – |
| BHK-21 | Baby hamster kidney | FGF-2 (bFGF)/ hs FGF-2 | 3 | DMEM/F-12 | 5% Calf |
| C6 [5,6] | Glial cells | BDNF | – | DMEM, high glucose | 2.5% FBS |
| C8-D1A | Astrocyte | BDNF | – | DMEM | 10% FBS |
| Daudi [7] | Burkitt's Lymphoma | GM-CSF | – | RPMI 1640 | 10% FBS |
| FDC-P1 [8,9] | Mouse bone marrow | GM-CSF | – | DMEM | 10% FBS |
| GH3 | Rat neuroblastoma | FGF-2 (bFGF)/ hs FGF-2 | 1 | F-12 | – |
| H9 (ESC) | Embryonic stem cells | FGF-2 (bFGF)/ hs FGF-2 | 4 | Gibco StemFlex Medium and supplement | KnockOut Serum Replacement |
| HAEC [10,11] | Aortic endothelial cells | VEGF | – | Human Large Vessel Endothelial Cell Basal Medium | Large Vessel Endothelial Supplement |
| HCAEC [12] | Human coronary artery endothelial cells | VEGF | – | Human Large Vessel Endothelial Cell Basal Medium | Large Vessel Endothelial Supplement |
| HeLa | Human cervical carcinoma | EGF | 2.5 x 104 | DMEM | 10% FBS |
| HF | Human foreskin fibroblast | EGF | 2 | DMEM | 10% FBS |
| HL-60 [13,14] | Leukemia | GM-CSF | 0.01–100 ng/mL | IMDM | 20% FBS |
| HPAEC [10,15,16] | Pulmonary artery endothelial cells | VEGF | – | Human Large Vessel Endothelial Cell Basal Medium | 2% FBS |
| HUVEC [7] | Human umbilical vein endothelial cells | VEGF | – | Ham’s F-12K | 10% FBS |
| iPSC [17] | Induced pluripotent stem cells | FGF-2 (bFGF)/ hs FGF-2 | – | Gibco Essential 8 Medium | RevitaCell Supplement (100X) |
| MCF 7 | Human mammary carcinoma | EGF | 100 | DMEM | 2% FBS |
| MKN-7 | Human adenocarcinoma | EGF | 50 | DMEM | 8% FBS |
| Neuro-2A | Neuroblastoma | BDNF | – | DMEM | 10% FBS |
| NRK-49F | Rat fibroblast | EGF | 50 | DMEM/F-12 | – |
| SH-SY5Y [5] | Neuroblastoma | BDNF | – | DMEM | 10% FBS |
| TF-1 [5] | Erythroleukemia | GM-CSF | 2 | RPMI 1640 | 10% FBS |
| THP-1 [18,19] | Monocytic leukemia | GM-CSF | – | RPMI 1640 | 10% FBS |
TM-4 | Mouse testes epithelium | EGF | 3 | – | |
| U-87 MG | Glioblastoma | BDNF | – | DMEM | 10% FBS |
| U-937 [18,19] | Monocytic leukemia | GM-CSF | – | RPMI 1640 | 10% FBS |
| WB | Rat hepatic epithelium | EGF | 10 | 10% FBS |
Don’t see your cell line? Our Cell Culture Select Tool features over 150 common cell types for a variety of applications. Simply enter in your chosen cell line and see results for the recommended medium, supplements, and cryopreservation environment—then, when you’re ready, add them to your cart for simplified shopping.
The following list of materials is generic and should be used as a guide to collect the necessary materials to complete your experiment. For a complete list of materials needed, please refer to the product-specific instructions for your cell line.
The following protocol does not specify the appropriate concentration of working medium, serum, reconstitution buffer, or types of growth factors needed to mix a complete growth medium. For specific instructions related to media preparation for your chosen cell lines, download the Product Sheet and Certificate of Analysis (CoA) to obtain the necessary information before beginning your experiment. Failure to do so will result in incorrect concentration or possible damage to the products. Always work in aseptic conditions to avoid contamination.
Grow and maintain a variety of mammalian cell lines with Gibco Cell Culture Media.
Gibco FBS supports consistent growth for your cells through essential nutrients and growth factors.
Access cell culture and transfection educational resources to enable excellent experiment planning and execution.
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Contact our expert team for technical and application support of Laboratory Products.