Search
(See a list of the products featured in this article.)
Parkinson’s disease (PD) is a debilitating, chronic, and progressive neurological disease that affects around 10 million people worldwide. Parkinson’s disease is accompanied by loss of motor function and, without a curative treatment, care remains palliative. Early diagnosis of Parkinson’s disease and similar neurodegenerative diseases is difficult because of their common symptoms and pathophysiology and the lack of understanding of their distinguishing molecular characteristics. Thermo Fisher Scientific is committed to developing research tools for neuroscientists investigating the molecular mechanisms of Parkinson’s disease and other neurodegenerative diseases using in vitro, ex vivo, and in vivo experimental systems.
In general the etiology of Parkinson’s disease and many other neurodegenerative disorders, including Alzheimer’s disease (AD), is poorly understood. The loss of motor function associated with Parkinson’s disease has led researchers to investigate the degeneration of dopaminergic neurons in the substantia nigra [1,2]. Key molecular mechanisms implicated in this neurodegeneration can be broadly classified into categories that include mitochondrial dysfunction, genetic mutations, and defects in protein clearance pathways that lead to protein aggregation, formation of neuronal inclusion bodies, and ultimately neuronal deterioration [2-5].
Historically, diagnosis has relied upon the identification of protein aggregates (e.g., Lewy bodies, in the case of Parkinson’s disease) that occur in the later stages of the disease. Neuronal inclusion body formation, defined by the presence of these insoluble protein aggregates, is responsible for the breakdown of neuronal microtubules, a causative event of neuronal diseases [1-5]. Two proteins commonly identified in neurodegenerative diseases—also referred to as proteinopathies due to protein misfolding—are tau and synuclein [5]. The proteinopathy of Parkinson’s disease is primarily attributed to alpha-synuclein, the main structural component of Lewy bodies in nerve cells, which are a defining characteristic of not only Parkinson’s disease but also Lewy body dementia (LBD) and other disorders. Although tau aggregation and the formation of amyloid plaques have been more extensively described in Alzheimer's disease, tau has also been linked with alpha-synuclein as an interdependent partner in the progression of Parkinson’s disease [2-6].
Although the protein–protein interactions that influence formation of inclusion bodies and plaques have been studied at the molecular level, the underlying causes of the degenerative pathophysiology are still unknown. Obviously, there is a need for more appropriate models and reagents to elucidate the principal mechanisms and indicators for the onset and progression of Parkinson’s disease. In the first half of this article, we give an overview of some key proteins involved in Parkinson’s disease and other neurodegenerative diseases, and the Invitrogen primary antibodies available to study these targets (Figures 2–5, Tables 1–3). In the second half, we highlight recently developed fluorescent tubulin-selective probes that can be used in live neurons to further reveal the relationships between neuronal protein aggregation and microtubules, as well as the possible roles of this association in neuronal disease.
Several proteins have emerged as key players in signaling pathways associated with various stages of Parkinson’s disease, including early onset, and latestage progression. Cellular defense mechanisms inherent to neurons play a critical role in preventing protein aggregation and maintaining cellular homeostasis. Molecular chaperones, also known as heat shock proteins (HSPs), have gained attention in
Parkinson’s disease research for their protective nature against misfolded proteins, preventing their aggregation.
HSPs are constitutively expressed or inducibly regulated in a number of different cell types, and disruption in their function can quickly lead to the accumulation of misfolded proteins and eventually cell death. These proteins, therefore, are being investigated as a means both to understand Parkinson’s disease disease biology and to study their potential role in disease treatment. The functions of several HSPs (including HSP27, HSP40, HSP60, HSP70, and HSP90) that are localized to axons or synapses and associated with alpha-synuclein are found to be compromised in Parkinson’s disease [7-9]. Figure 1 shows the validation* data for representative Invitrogen HSP antibodies, and Table 1 provides a list of antibodies against several members of the HSP family.
Figure 1. Assessment of heat shock protein (HSP) antibody specificity.(A) Western blot analysis of HSP40 antibody using RNAi. (B) Western blot analysis across multiple cell lines and (C) immunofluorescence analysis in HeLa cells using HSP60 antibody (clone 2E1/53). For the immunofluorescence analysis, HeLa cells were labeled with HSP60 antibody (green) and counterstained with rhodamine phalloidin (red) and DAPI (blue). (D) Western blot analysis of HSP70 (clone 2A4) and (E)HSP90 (clone 3B6) antibodies across multiple cell lines. (F) Western blot analysis of phospho-HSP27 (Ser85) antibody in cells subjected to anisomycin or UV, both known to activate HSPs. For the western blot analyses, primary antibodies were probed with Invitrogen Goat Anti–Rabbit IgG Superclonal Recombinant Secondary Antibody, HRP or Invitrogen Goat Anti–Mouse IgG Superclonal Recombinant Secondary Antibody, HRP, detected using the Thermo Scientific Pierce ECL Western Blotting Substrate, and imaged using the Invitrogen iBright FL1000 Imaging System.
| Target protein | Cat. No. |
|---|---|
| HSP40 (polyclonal) | PA5-17382 |
| HSP60 (clone 4B9/89 or clone 2E1/53) | MA3-012,MA3-013 |
| HSP60 (polyclonal) | PA5-34760 |
| HSP70 (clone 4G4) | MA3-009 |
| HSP70 (clone 2A4) | MA3-008 |
| HSP90 alpha (clone 3B6) | MA3-010 |
| HSP90 beta (clone H9010) | 37-9400 |
| Phospho-HSP27 (Ser15, polyclonal) | PA1-018 |
| Phospho-HSP27 (Ser85, polyclonal) | PA1-005 |
The accumulation of alpha-synuclein, one of the main structural components of Lewy bodies, is a hallmark of the latter stages of Parkinson’s disease [10]. Although its specific role in the neuronal degeneration process is not fully understood, studies have suggested that it is involved in aggregation of misfolded protein that eventually converges upon dopaminergic neurons in the form of Lewy bodies [6,10,11]. Figure 2 demonstrates validation data for the Invitrogen alpha-synuclein antibody, which was tested using different parameters to ensure its specificity.
A growing number of genes associated with Parkinson’s disease are related to mitochondrial function and oxidative stress, emphasizing the role of mitochondria in neuronal degeneration and disease progression [12]. Similarly, impairment of lysosome-mediated protein degradation via autophagy or endocytosis directly impacts the ability of neurons to clear alpha-synuclein aggregates in Lewy bodies [13,14].
Several proteins involved in normal mitochondrial or lysosomal functioning, such as parkin, LIMP2, and PINK1 [15], are found to be mutated in Parkinson’s disease. As a result, affected cells are subjected to increased oxidative stress and eventually cell death. Additionally, mutations in proteins such as DNAJC13 (required for receptor-mediated endocytosis) and optineurin (important for maintenance of the Golgi complex and membrane trafficking) have been identified as contributing to Parkinson’s disease pathology [16,17]. In Figure 3, we showcase representative validation data for antibodies against several of these targets that can be used across applications, including immunofluorescence and western blotting. Table 2 provides a list of antibodies against a variety of important Parkinson’s disease targets.
Figure 2. Assessment of alpha-synuclein antibody specificity.(A) Immunofluorescence analysis in mouse brain tissue using alpha-synuclein polyclonal antibody (red) and a neurofilament-L monoclonal antibody (green); cells were counterstained with DAPI (blue). Western blot analysis of alpha-synuclein antibody using (B) CRISPR knockout (KO) of alpha-synuclein in HeLa cells and (C) relative expression of alpha-synuclein in brain and liver. Western blot detection was performed as described in Figure 1.
Figure 3. Assessment of antibody specificity. Western blot analysis of (A)parkin antibody (clone 21H24L9) and (B)DJ-1 antibody to show specificity using CRISPR knockout (KO) and RNAi, respectively. (C) Immunofluorescence analysis in HepG2 cells labeled with LIMP2 antibody (clone 22H6L14) (green) and Invitrogen LysoTracker Red DND-99 (red); cells were counterstained with DAPI (blue). (D) Immunofluorescence analysis of DNAJC13 antibody (clone 11H13L5) (green) in A431 cells using RNAi; cells were counterstained with rhodamine phalloidin (red) and DAPI (blue). (E) Immunofluorescence analysis of optineurin in mouse retina using an isotype control (green) or optineurin recombinant polyclonal antibody (green); cells were counterstained with DAPI (blue). Western blot analysis of the optineurin antibody in (F) liver and eye lysates and (G) optineurin siRNA–silenced NIH/3T3 cells. Western blot detection was performed as described in Figure 1.
| Target protein | Protein function | Cat. No. |
|---|---|---|
| Ataxin3 | Protein clearance | 711823 |
| DENND5A | Vesicle-mediated transport and RAB-activating guanine nucleotide exchange factor (GEF) | 702789 |
| DJ-1 (Park7) | Molecular chaperone | PA5-13404 |
| DNAJC13 (RME-8) | Membrane trafficking | 702773 |
| DNML1 | Clathrin-mediated endocytosis | 702782 |
| LIMP2 | Lysosomal membrane protein | 702770 |
| Mitofusin-2 (MFN2) | Mitochondrial maintenance | 711803 |
| Optineurin | Membrane trafficking | 711879 |
| Alpha-synuclein | Regulation of neurotransmitter release | PA5-17239 |
| TMEM230 | Synaptic vesicle recycling | 702792 |
| VAMP1 | Vesicular transport | 702787 |
Tau, a member of the microtubule-associated protein family (MAPT), is associated with microtubule stabilization, membrane binding, and axonal transport [17-20]. Under normal physiological conditions, tau is soluble and unfolded; however, under pathological conditions in Parkinson’s or Alzheimer's disease, misfolding of tau leads to insoluble neurofibrillary tangles (NFTs)—a pathological effect called a tauopathy [4-6,21]. In such pathology, hyperphosphorylation of tau causes protein aggregation and accumulation, which in turn damages neurons and disrupts axonal transport [17,18,21]. Although alpha-synuclein dysregulation is a possible cause of Parkinson’s disease, it has recently been reported that both alpha-synuclein and tau are present in the same Lewy body aggregate in many postmortem brains with Parkinson’s disease and dementia [4-6]. Consequently, clinicians and researchers alike are focusing on the link between tau and alpha-synuclein in Parkinson’s disease at the molecular level [4-6,10]. Invitrogen primary antibodies against total tau as well as several important phosphorylated forms have been published and highly cited in publications. Figure 4 shows representative validation data for some of these antibodies, and Table 3 provides a list of antibodies against these important Parkinson’s disease targets.
Figure 4. Assessment of tau antibody specificity.(A) Western blot analysis of tau monoclonal antibody (clone TAU-5) across multiple cell lines. (B) Immunofluorescence analysis in human iPSC-derived forebrain organoids at day 40 using tau monoclonal antibody (clone TAU-5) (green) and phospho-tau (Ser396) antibody (red), and counterstained with DAPI (blue); data used with permission from Zhexing Wen, Emory University. (C) Western blot analysis of the specificity of phospho-tau (Ser199, Ser202) antibody binding in the presence of the specific phosphopeptide target (lane 2), the corresponding non-phosphorylated peptide (lane 3), and other phosphoserine-containing peptides (lanes 4–6); signal loss was only seen with the specific peptide. (D) Western blot analysis of phospho-tau (Ser262) antibody in mouse brain, rat brain, and mouse kidney lysates. For the western blot analyses, primary antibodies were probed with Invitrogen Goat Anti–Rabbit IgG Secondary Antibody, HRP or Invitrogen Goat Anti–Mouse IgG Secondary Antibody, HRP, detected using the Thermo Scientific Pierce ECL Western Blotting Substrate, and imaged using the Invitrogen iBright FL1000 Imaging System.
| Target protein | Cat. No. |
|---|---|
| Tau (clone HT7) | MN1000, MN1000B (biotin conjugate) |
| Tau (clone TAU-5) | AHB0042, MA5-12808, MA5-12805 (biotin conjugate) |
| Tau (clone T46) | 13-6400 |
| Phospho-tau (Ser199, Ser202) | 44-768G |
| Phospho-tau (Ser202, Thr205) (clone AT8) | MN1020, MN1020B (biotin conjugate) |
| Phospho-tau (Thr231) (clone AT180) | MN1040 |
| Phospho-tau (Thr212, Ser214) (clone AT100) | MN1060 |
Recently, tau aggregation and spreading into naive neurons has been shown to occur through tunneling nanotubes (TNTs) [8,18,20,21]. There are different types of TNTs that are thought to provide cell-to-cell communication, and one type of TNT is composed of actin and microtubules [22]. Microtubules are highly dynamic polymers of tubulin that are important for neuronal outgrowth, morphology, and migration, and there is growing evidence that their dysfunction is related to Parkinson’s disease [21,23,24]. The relationships between alpha-synuclein and tau, as well as between tau and the microtubule cytoskeleton, present an opportunity to better characterize the mechanisms involved in protein aggregate formation in Parkinson’s disease.
To visualize dynamic microtubules in live neuronal cultures, we offer the tubulin-selective fluorescent probes Invitrogen Tubulin Tracker Deep Red and Tubulin Tracker Green. These are cell-permeant fluorescent dyes that bind to polymerized tubulin within the microtubule. The Tubulin Tracker Green reagent (Ex/Em = 494/522 nm) has been used for the study of microtubule dynamics in Parkinson’s disease–related research [18]. Neuronal staining using the recently introduced Tubulin Tracker Deep Red reagent (Ex/Em = 652/669 nm) is shown in Figure 5.
Figure 5. Live rat cortical neurons labeled with Tubulin Tracker Deep Red.Gibco Primary Rat Cortex Neurons were prepared and grown using the Gibco B-27 Plus Neuronal Culture System and Gibco GlutaMAX Supplement for 14 days. Cells were then stained with Invitrogen Tubulin Tracker Deep Red (Ex/Em = 652/669 nm) and imaged in Invitrogen Live Cell Imaging Solution on the Invitrogen EVOS FL Auto 2 Imaging System with a 20x objective and an Invitrogen EVOS Cy®5 Light Cube.
To test and demonstrate the low cytotoxicity and phototoxicity of Tubulin Tracker Deep Red, mouse embryonic hippocampal neurons were labeled with the tubulin-selective fluorescent reagent and microtubule formation in neuronal cultures was monitored over the course of 72 hours (at 5-minute intervals) using extended time-lapse imaging (Figure 6). Normal levels of dynamic assembly, outgrowth, and retraction of neurites were observed throughout the entire time course, with no detachment from the growth substrate or indication of other cytotoxic phenotypes, even under aggressive imaging conditions. Given the relative sensitivity of primary neuronal cultures, these data clearly demonstrate the advantages of long-wavelength excitation in live cells, as well as the low cytotoxicity of Tubulin Tracker Deep Red in primary neuronal cultures.
Figure 6. Tubulin Tracker Deep Red for extended time-lapse imaging in primary neurons.Gibco Primary Mouse Hippocampal Neurons were cultured for 14 days in vitro using the Gibco B-27 Plus Neuronal Culture System and Gibco GlutaMAX Supplement and then imaged across 72 hr at 5 min intervals. Left panel shows no changes in neuronal dynamic assembly, even after 72 hr of staining in Invitrogen Tubulin Tracker Deep Red. Right panel shows a 4-fold higher magnification of the neurons to monitor potential changes in neuronal outgrowth with dye treatment; arrows denote outgrowth activity, with each color tracking an individual neuron over time. Tubulin Tracker Deep Red was added to the culture at the beginning of the experiment; no wash step was required throughout the study. The labeled neurons were imaged in a live-cell chamber on a confocal laser-scanning microscope.
The benefits of microtubule specificity and absence of cytotoxicity in live primary neuronal cultures prompted us to investigate the use of Tubulin Tracker Deep Red as a novel tool for pharmacological screening in neurons. Figure 7 shows a dose-response study in live rat cortical neurons. Upon treatment with cadmium chloride (CdCl2), we quantified the dose-dependent toxicity in primary neurons by measuring Tubulin Tracker Deep Red intensity branch points per neuron, and neurite area using image segmentation software found on the Thermo Scientific CellInsight CX5 High-Content Screening (HCS) Platform. The results mirrored the known cytotoxic profile of CdCl2 in neuronal cultures, providing proof-of-concept that Tubulin Tracker Deep Red can be utilized for a readout of these stereotypical neuronal traits to aid in the screening of new drugs aimed at a variety of neuronal degenerative diseases, including
Parkinson’s disease.
Figure 7. Pharmacological screening of neurite outgrowth with Tubulin Tracker Deep Red.Gibco Primary Rat Cortex Neurons were cultured for 14 days in vitro using the Gibco B-27 Plus Neuronal Culture System and Gibco GlutaMAX Supplement. Cells were then treated with different concentrations of cadmium chloride (CdCl2) for 18 hr and stained using Invitrogen Tubulin Tracker Deep Red. Neurite morphology was assessed using the Thermo Scientific CellInsight CX5 High-Content Screening (HCS) Platform.
3D models have emerged as effective systems for bridging traditional monolayer cell-based assays and in vivo animal experiments, and their adoption is increasing in many different research areas. Gibco Neurobasal Medium combined with Tubulin Tracker Deep Red was used to establish 3D neurospheres, a useful approach for neuronal disease modeling. To demonstrate the specificity of Tubulin Tracker Deep Red for microtubules in neural stem cell (NSC)-derived preparations, the total neuronal content of 3D neurospheres cultured using different growth conditions was assessed by measuring the overall intensity of labeling with Tubulin Tracker Deep Red (Figure 8). When comparing traditional Neurobasal Medium to Neurobasal Plus Medium in the presence and absence of Gibco CultureOne Supplement, clear differences in microtubule content across the four growth conditions were observed (data not shown). We found that cultures in Neurobasal Plus Medium were far better at promoting neuronal growth for applications in 3D neuronal models.
Figure 8. Tubulin Tracker Deep Red as a marker of neurosphere maturation. Neural stem cell (NSC)-derived neurospheres were cultured using Gibco Neurobasal Plus Medium with Gibco CultureOne Supplement in Thermo Scientific Nunclon Sphera 96 U-Well Microplates. The neurospheres were then stained with Invitrogen NucBlue Live ReadyProbes Reagent and Invitrogen Tubulin Tracker Deep Red to assess neurosphere maturation. Image was generated using the Invitrogen EVOS FL Auto 2 Imaging System with a 20x objective and an Invitrogen EVOS Cy®5 Light Cube.
Although the number of genes and proteins implicated in Parkinson’s disease continues to grow, a majority of them are proving to be a challenge to study given their low abundance and a lack of quality reagents to investigate their functions. Invitrogen antibodies are tested in relevant models and applications, and their specificity is confirmed using siRNA- and CRISPR-mediated gene silencing, relative expression, and cell treatment.
Tubulin Tracker reagents have been used to visualize microtubules in a wide variety of live cell types, and Tubulin Tracker Deep Red has been demonstrated to label 3D neurospheres. Tubulin Tracker Deep Red can be multiplexed with common blue, green, orange, and near-IR fluorescent reagents and enables the study of microtubule dynamics with extended time-lapse imaging, including pharmacological effects on microtubule networks in live neuronal cultures.
* The use or any variation of the word “validation” refers only to research use antibodies that were subject to functional testing to confirm that the antibody can be used with the research techniques indicated. It does not ensure that the product(s) was validated for clinical or diagnostic uses.
Download a printer-friendly version of this article.
Download now
For Research Use Only. Not for use in diagnostic procedures.