High quality, pure, intact RNA is critical to most gene expression experiments. The Thermo Fisher Scientific portfolio of RNA preparation products runs both broad and deep, to provide the right solution every time. Choose from silica-column or magnetic bead kits for total RNA extraction. Find the plastics, magnets, and purification instruments best suited to your preferred method. And be confident in your RNase-free workspace with RNase-free lab essentials for every step.    
RNase-free workspace
Bottle of RNaseZap solution for surface decontamination
Eliminating nuclease contamination, particularly RNase contamination, is a constant, tedious challenge. We offer several surface decontamination products that are proven effective at eliminating RNase contamination from lab surfaces.
Learn more ›
RNase-free lab essentials
RNA extraction
For over two decades, our scientists have developed innovative, robust RNA extraction products that make your job easier and your gene expression analysis more precise.
Total RNA extraction | microRNA & small RNA isolation | mRNA extraction | Sequence-specific RNA extraction | Purified RNA | Viral RNA from liquid and cell-free samples | Automated RNA extraction
Total RNA extraction
Skip to bottom
Gold standard for highly pure, intact RNA
High-quality RNA in less than 20 minutes
High-throughput purification of RNA and DNA
Complete, no purification system for qRT-PCR results
TRIzol Reagents
PureLink Kits
MagMAX Kits
Cells to Ct
Isolation method
High purity organic extraction (requires alcohol precipitation)
Fast, convenient silica column
Scalable, flexible format with magnetic beads
Chemical lysis (time to qRT-PCR results)
Prep time
~1 hr
< 20 minutes
~45 minutes
10 minutes
Compatible sample types
Most sample types, particularly more difficult to lyse
Cells and tissues
Tissues, cells, blood, serum/plasma, urine, plants
Cells
microRNA & small RNA isolation
Back to top
Skip to bottom
30 min isolation from most samples
Recover all RNAs as well as proteins
HTP isolation of total RNA enriched for small RNAs
Complete kit, sample-to-qPCR from cells
mirVana miRNA Isolation Kit (with or without phenol)
mirVana Paris
MagMAX mirVana Total RNA Isolation Kit
TaqMan microRNA Cells-to-CT Kit
RNA types isolated
Small & large RNA molecules
Small & large RNA molecules
Small & large RNA molecules
Small & large RNA molecules
Prep time
20 minutes
30 minutes
<1 hour
10 minutes
Compatible sample types
Most sample types
Most sample types
Serum/Plasma, whole blood, Fresh/frozen tissue, cells, urine
Cells
Isolation method
Highest purity and convenience; includes both organic extraction and silica column
Highest purity and convenience; includes both organic extraction and silica column
Scalable, flexible format with magnetic beads
Cells-to-CT direct lysis, no purification required
Rapid mRNA purified from total RNA
No purification needed, mRNA direct from crude samples
mRNA from micro-sized samples best for next-generation sequencing of mRNA
HTP mRNA directly or enrich from total RNA
Dynabeads\ mRNA Purification Kit
Dynabeads mRNA DIRECT
Dynabeads mRNA DIRECT Micro Kit
mRNA Catcher Plus
Prep time
15 minutes
15 minutes
15 minutes
1.5 hours
Direct isolation of mRNA from crude samples
No
Yes
Yes
Yes
mRNA enrichment from total RNA
Yes
No
No
Yes
Sequence-specific RNA extraction
Back to top
Skip to bottom
4 beads in one kit, identify best for your application
Best solution for viscous samples
High surface area for maximum enrichment
Dynabeads Streptavidin Trial Kit
Dynabeads M-270 Streptavidin
Dynabeads MyOne  Streptavidin C1
Bead size
1 µm x 2, 2.8 µm x 2
2.8 µm
1 µm
Biotinylated Ig (µg/mg beads)
up to 20
up to 10
up to 20
Binding capacity (free biotin)
6502,500 pmoles/mg beads
650–1,350 pmoles/mg beads
2,500 pmoles/mg beads
  • RNA available from a wide variety of human tissues and cell lines
  • Certified to contain small RNAs (miRNA, siRNA, and snRNA)
To learn more, review our product offerings ›
  • DNase-treated to remove contaminating DNA
  • Superior quality control standards
Viral RNA from liquid and cell-free samples
Back to top
Skip to bottom
Product
Format
Most suitable downstream application
Real-Time RT-PCR
Microarray analysis
Sequencing
Northern blot
Cloning
Total RNA
PureLink Viral RNA⁄DNA Mini Kit
Chaotropic salt-based lysis, silica filter
RNAqueous Kit
Chaotropic salt-based lysis, silica filter
MagMAX AI⁄ND Viral RNA Isolation Kit
Chaotropic salt-based lysis, magnetic particles
MagMAX-96 AI⁄ND Viral RNA Isolation Kit
Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible
DNA/RNA
Product
Format
Most suitable downstream application
Real-Time RT-PCR
Microarray analysis
Sequencing
Northern blot
Cloning
PureLink Pro 96 Viral RNA⁄DNA Purification Kit
Chaotropic salt-based lysis, silica filter, 96-well processing, automation compatible
MagMAX Total Nucleic Acid Isolation Kit
Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible
MagMAX-96 Viral RNA Isolation Kit
Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible
Dynabeads SILANE Viral NA
Chaotropic salt-based lysis, magnetic beads, automation compatible
iPrep PureLink Virus Kit
Chaotropic salt-based lysis, magnetic particles, automation compatible
MagMAX Viral RNA Isolation Kit
Chaotropic salt-based lysis, magnetic particles
MagMAX-96 Viral RNA Isolation Kit (5 x 96 rxns)
Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible
MicroRNA
Product
Format
Most suitable downstream application
Real-Time RT-PCR
Microarray analysis
Sequencing
Northern blot
Cloning
mirVana miRNA Isolation Kit
Chaotropic salt-based lysis, silica filter
mirVana miRNA Isolation Kit (without phenol - must be purchased separately)
Chaotropic salt-based lysis, silica filter
TRIzol LS Reagent
Lysis in TRIzol reagent, one-step reagent
mRNA
Product
Format
Most suitable downstream application
Real-Time RT-PCR
Microarray analysis
Sequencing
Northern blot
Cloning
Dynabeads mRNA DIRECT Kit
Chaotropic salt-based lysis, magnetic beads
Dynabeads mRNA DIRECT Micro Kit
Chaotropic salt-based lysis, magnetic beads
Automated RNA extraction
Back to top
KingFisher instruments automate RNA extraction by moving magnetic beads (not liquids), for clean extractions and consistent, repeatable results. Automation enables increased sample output in shorter time, with minimal hands-on effort.
KingFisher Duo Prime
KingFisher Duo Prime
KingFisher Flex
KingFisher Flex
KingFisher Presto
KingFisher Presto
Complact benchtop
Benchtop
Benchtop, integrates with robotic liquid handler
Low–Medium
6 or 12 samples per run
Medium–High
24 or 96 samples per run
Ultrahigh
24 or 96 samples per run
View demo
RNA stabilization
RNAlater-ICE Frozen Tissue Transition Solution
Style Sheet for Global Design System
Style Sheet for Komodo Tabs