The increasing complexity of semiconductor device structures, along with the shrinking of structural dimensions, means that designing next-generation devices is more challenging and time-consuming than ever before. This, coupled with the fact that the number of technology and design options available is increasing, means a lower probability that any particular design will be commercially successful. As a result, device manufacturers need reliable tools for pathfinding that reduce the number of viable options available and help them implement solutions faster.
The complexity of considering all the chip, package, and system integration options that exist has made implementation an intimidating task. As a result, continuing evolution of the most advanced pathfinding capability has become a requirement in efficient semiconductor device design. Adding to this level of complexity are structures with multifaceted 3D architectures. Isolating defects or resolving material interfaces in a focused-ion-beam (FIB) cut structural cross-section requires highly precise preparation and subsequent scanning electron microscopy (SEM) or scanning transmission electron microscopy (STEM) imaging. Precision FIB editing tools can also perform microsurgery and nanoprototyping of new circuit designs. Finally, due to limited floorspace and budgets, labs are pushing to have multiple analysis options in a single system to get the most comprehensive data in the shortest possible time.
Thermo Fisher Scientific provides a full suite of analytical instruments that enable advanced R&D on innovative logic, memory, power and display device technologies. We offer the most advanced capability to perform high-end atomic-level research and prototyping, using STEM and FIB microscopy.
Semiconductor pathfinding and device development workflow examples
Style Sheet for Komodo Tabs
Techniques
Metrología de TEM
Las rutinas de metrología TEM avanzadas y automatizadas ofrecen una precisión significativamente mayor que los métodos manuales. Esto permite a los usuarios generar grandes cantidades de datos estadísticamente relevantes, con una especificidad de nivel subangstrom, sin fallos del operador.
Más información ›
Adquisición de imágenes y análisis TEM de semiconductores
Los microscopios de electrones de transmisión de Thermo Fisher Scientific ofrecen imágenes y análisis de alta resolución de dispositivos semiconductores, lo que permite a los fabricantes calibrar conjuntos de herramientas, diagnosticar mecanismos de fallos y optimizar la producción rendimiento general del proceso.
Más información ›
Preparación de muestras de dispositivos semiconductores
Los sistemas DualBeam de Thermo Scientific proporcionan una preparación precisa de las muestras de TEM para el análisis a escala atómica de dispositivos semiconductores. La automatización y las tecnologías de aprendizaje automático avanzado producen muestras de alta calidad, en la ubicación correcta y con un bajo costo por muestra.
Más información ›
Adquisición de imágenes y análisis de semiconductores
Thermo Fisher Scientific ofrece microscopios electrónicos de barrido para todas las funciones de un laboratorio de semiconductores, desde tareas generales de adquisición de imágenes hasta técnicas avanzadas de análisis de fallos que requieren mediciones precisas de contraste de tensión.
Más información ›
Aislamiento de fallo óptico
Los diseños cada vez más complejos complican el aislamiento de fallos y defectos en la fabricación de semiconductores. Las técnicas de aislamiento óptico de fallos le permiten analizar el rendimiento de los dispositivos activos eléctricamente para localizar defectos críticos que causan fallos en el dispositivo.
Más información ›
Aislamiento de fallo térmico
La distribución desigual de la disipación de energía local puede causar aumentos de temperatura importantes y localizados, lo que provoca un fallo del dispositivo. Ofrecemos soluciones únicas para el aislamiento de fallos térmicos con termografía infrarroja de alta sensibilidad (LIT).
Más información ›
Edición de circuito
Las soluciones exclusivas y avanzadas de edición de circuitos y creación de prototipos, que combinan nuevos sistemas de suministro de gas con una amplia gama de productos químicos y tecnología de haz de iones focalizada, ofrecen un control y una precisión sin precedentes para el desarrollo de dispositivos semiconductores.
Más información ›
Nanosondeo
A medida que aumenta la complejidad del dispositivo, también lo hace el número de ubicaciones que tienen que ocultar los defectos. El nanosondeo proporciona la ubicación precisa de fallos eléctricos, lo que es fundamental para un flujo de trabajo de análisis de fallos de microscopía electrónica de transmisión eficaz.
Más información ›
Ablación por láser de semiconductores
La ablación por láser proporciona un fresado de alto rendimiento de dispositivos semiconductores para la adquisición de imágenes y análisis con microscopía electrónica, a la vez que conserva la integridad de las muestras. Acceda a datos en 3D de gran volumen y optimice las condiciones de fresado para adaptarse mejor a su tipo de muestra.
Más información ›
Preparación de muestras de APT
La tomografía de sonda atómica (APT) proporciona un análisis de composición de materiales en 3D con resolución atómica. La microscopía Focused ion beam (FIB) es una técnica esencial para la preparación de muestras de alta calidad, orientación y sitio específico para la caracterización de APT.
Más información ›
Reestructuración de dispositivo
La contracción del tamaño de las características, junto con los resultados de diseño y arquitectura avanzados provocan fallos cada vez más complicados para los semiconductores. La reestructuración sin daños de los dispositivos es una técnica crucial para la detección de errores y fallos eléctricos interiores.
Más información ›
Metrología de TEM
Las rutinas de metrología TEM avanzadas y automatizadas ofrecen una precisión significativamente mayor que los métodos manuales. Esto permite a los usuarios generar grandes cantidades de datos estadísticamente relevantes, con una especificidad de nivel subangstrom, sin fallos del operador.
Más información ›
Adquisición de imágenes y análisis TEM de semiconductores
Los microscopios de electrones de transmisión de Thermo Fisher Scientific ofrecen imágenes y análisis de alta resolución de dispositivos semiconductores, lo que permite a los fabricantes calibrar conjuntos de herramientas, diagnosticar mecanismos de fallos y optimizar la producción rendimiento general del proceso.
Más información ›
Preparación de muestras de dispositivos semiconductores
Los sistemas DualBeam de Thermo Scientific proporcionan una preparación precisa de las muestras de TEM para el análisis a escala atómica de dispositivos semiconductores. La automatización y las tecnologías de aprendizaje automático avanzado producen muestras de alta calidad, en la ubicación correcta y con un bajo costo por muestra.
Más información ›
Adquisición de imágenes y análisis de semiconductores
Thermo Fisher Scientific ofrece microscopios electrónicos de barrido para todas las funciones de un laboratorio de semiconductores, desde tareas generales de adquisición de imágenes hasta técnicas avanzadas de análisis de fallos que requieren mediciones precisas de contraste de tensión.
Más información ›
Aislamiento de fallo óptico
Los diseños cada vez más complejos complican el aislamiento de fallos y defectos en la fabricación de semiconductores. Las técnicas de aislamiento óptico de fallos le permiten analizar el rendimiento de los dispositivos activos eléctricamente para localizar defectos críticos que causan fallos en el dispositivo.
Más información ›
Aislamiento de fallo térmico
La distribución desigual de la disipación de energía local puede causar aumentos de temperatura importantes y localizados, lo que provoca un fallo del dispositivo. Ofrecemos soluciones únicas para el aislamiento de fallos térmicos con termografía infrarroja de alta sensibilidad (LIT).
Más información ›
Edición de circuito
Las soluciones exclusivas y avanzadas de edición de circuitos y creación de prototipos, que combinan nuevos sistemas de suministro de gas con una amplia gama de productos químicos y tecnología de haz de iones focalizada, ofrecen un control y una precisión sin precedentes para el desarrollo de dispositivos semiconductores.
Más información ›
Nanosondeo
A medida que aumenta la complejidad del dispositivo, también lo hace el número de ubicaciones que tienen que ocultar los defectos. El nanosondeo proporciona la ubicación precisa de fallos eléctricos, lo que es fundamental para un flujo de trabajo de análisis de fallos de microscopía electrónica de transmisión eficaz.
Más información ›
Ablación por láser de semiconductores
La ablación por láser proporciona un fresado de alto rendimiento de dispositivos semiconductores para la adquisición de imágenes y análisis con microscopía electrónica, a la vez que conserva la integridad de las muestras. Acceda a datos en 3D de gran volumen y optimice las condiciones de fresado para adaptarse mejor a su tipo de muestra.
Más información ›
Preparación de muestras de APT
La tomografía de sonda atómica (APT) proporciona un análisis de composición de materiales en 3D con resolución atómica. La microscopía Focused ion beam (FIB) es una técnica esencial para la preparación de muestras de alta calidad, orientación y sitio específico para la caracterización de APT.
Más información ›
Reestructuración de dispositivo
La contracción del tamaño de las características, junto con los resultados de diseño y arquitectura avanzados provocan fallos cada vez más complicados para los semiconductores. La reestructuración sin daños de los dispositivos es una técnica crucial para la detección de errores y fallos eléctricos interiores.
Más información ›
Samples
Materiales semiconductores y caracterización de dispositivos
A medida que los dispositivos semiconductores se reducen y se vuelven más complejos, se necesitan nuevos diseños y estructuras. Los flujos de trabajo de análisis en 3D de alta productividad pueden reducir el tiempo de desarrollo de dispositivos, maximizar el rendimiento y garantizar que los dispositivos satisfacen las necesidades futuras del sector.
Más información ›
Products
Hoja de estilo para tarjetas originales instrumentos
Helios 5 PFIB DualBeam
  • Preparación de muestras STEM y TEM sin galio
  • Subsuperficie multimodal e información 3D
  • Columna FIB con plasma xenón de 2,5 μA de última generación
Helios 5 DualBeam
  • Preparación de muestras de TEM ultrafina, de alta calidad y completamente automatizada
  • Subsuperficie de alto rendimiento, alta resolución y caracterización en 3D
  • Capacidades de rápida creación de prototipos a nanoescala
Metrios AX TEM
  • Opciones de automatización compatibles con calidad, uniformidad, metrología y OPEX reducido
  • Aprovecha el aprendizaje automático para excelentes funciones automáticas y reconocimiento de funciones
  • Flujos de trabajo para preparación de laminillas in-situ y ex-situ
Scios 2 DualBeam
  • Soporte completo de muestras magnéticas y no conductoras
  • Subsuperficie de alto rendimiento y caracterización en 3D
  • Facilidad de uso y capacidades de automatización avanzadas
ExSolve WTP DualBeam
  • Puede preparar laminillas específicas de sitios de 20 nm de grosor en obleas completas de hasta 300 mm de diámetro
  • Soluciona las necesidad de muestreo automatizado de alto rendimiento en nodos de tecnología avanzada
Verios 5 XHR SEM
  • SEM monocroma para resolución de subnanómetros sobre el rango de energía completo de 1 keV a 30 keV
  • Fácil acceso a la energía de recepción de haz, a un nivel tan bajo como 20 eV
  • Excelente estabilidad con fase piezoeléctrica como estándar
Quattro ESEM
  • Tecnología de obtención de imágenes por microscopía electrónica de barrido (SEM) con cañón de emisión de campo (FEG) de alta resolución con versatilidad absoluta y capacidad medioambiental única (ESEM)
  • Consulte la información completa de todas las muestras con obtención de imágenes SE y BSE en cada modo de funcionamiento
Prisma E SEM
  • SEM de nivel básico con excelente calidad de imagen
  • Rápida y sencilla navegación y carga de muestras para varias muestras
  • Compatible con una amplia gama de materiales gracias a los exclusivos modos de vacío
Apreo 2 SEM
  • SEM de alto rendimiento para resolución de subnanómetro o nanómetro completo
  • Detector de retrodispersión de T1 incorporado en la columna para contraste de materiales sensibles de velocidad TV
  • Excelente rendimiento en distancias de trabajo largas (10 mm)
VolumeScope 2 SEM
  • Datos 3D isotrópicos de grandes volúmenes
  • Alto contraste y resolución en los modos de vacío alto y bajo
  • Un solo movimiento entre el uso de SEM normal y la adquisición de imágenes de cara de bloques en serie
SEM de escritorio Phenom ProX
  • SEM de escritorio de alto rendimiento con detector EDS integrado
  • Resolución de <8 nm (SE) y <10 nm (BSE); aumento de hasta 150.000x
  • Detector SE opcional
Centrios CE
  • Excelente resolución de imagen/fresado
  • Control y precisión de fresado mejorados
  • Basado en la plataforma Thermo Scientific Helios DualBeam
Sistema ELITE
  • Completamente no destructivo
  • Identifica rápidamente el componente defectuoso en la tarjeta de montaje y obtener la disposición exacta
  • Localiza el defecto en x-y con la precisión del micrómetro, con una ubicación de profundidad exacta de 20 µm
nProber IV
  • Localice los fallos BEOL y del transistor
  • Nanosondeo térmico (de -40° C a 150 °C)
  • Funcionamiento semiautomático
Sistema Hyperion II
  • Sondeo de fuerza atómica
  • Localice los fallos del transistor
  • Corriente de pico integrada (CAFM)
Sistema Meridian S
  • Diagnóstico de fallos con tecnología de sonda activa
  • Estimulación láser estática (SLS / OBIRCH) y opciones de emisión de fotones
  • Admite la estimulación de dispositivos de microsonda y de tarjetas de sonda
Sistema Meridian WS-DP
  • Detección de emisiones de fotones de baja tensión, bajo nivel de ruido y alta sensibilidad con sistemas de cámara DBX o InGaAs de banda ancha
  • Microscopio de barrido láser de longitud de onda múltiple para análisis de cadena de barrido, mapeo de frecuencia, sondeo de transistores y aislamiento de fallos
Sistema Meridian 7
  • Aislamiento dinámico de fallos ópticos para el nodo de 10 nm e inferiores
  • Luz infrarroja y visible de alta resolución
  • Preparación de muestras de alto rendimiento de 5 μm disponible
Sistema Meridian IV
  • Detección de emisión de fotones DBX de longitud de onda ampliada de alta sensibilidad
  • Detección de emisión de fotones de InGaAs estándar
  • Microscopio de barrido láser con opciones de longitud de onda múltiple
AutoTEM 5
  • Preparación de muestras S/TEM in situ totalmente automatizada
  • Compatible con geometría invertida, plana y "top-down" (arriba-abajo)
  • Flujo de trabajo altamente configurable
  • Interfaz de usuario intuitiva, fácil de utilizar
Software Auto Slice and View 4.0
  • Cortes en secciones en serie automatizadas para DualBeam
  • Adquisición de datos multimodales (SEM, EDS, EBSD)
  • Capacidades de edición sobre la marcha
  • Ubicación del corte en los bordes
Software Maps
  • Adquiera imágenes de alta resolución en grandes áreas
  • Encuentre fácilmente áreas de interés
  • Automatice el proceso de adquisición de imágenes
  • Correlacione datos de diferentes fuentes
Software Avizo
  • Compatibilidad con varios datos/varias vistas, multicanal, series temporales, datos de gran tamaño
  • Registro automático avanzado multimodo 2D/3D
  • Algoritmos de reducción de artefactos
Software Ifast
  • Grabador de macros para crear recetas más rápido
  • Electroforesis para funcionamiento nocturno sin supervisión
  • Herramientas de alineación: Reconocimiento de imágenes y detección de bordes
Software Inspect 3D
  • Filtros y herramientas de tratamiento de imágenes para correlación cruzada
  • Seguimiento de funciones para alineación de imágenes
  • Técnica de reconstrucción algebraica para la comparación de proyección iterativa
Software NEXS
  • Sincroniza automáticamente la posición/aumento entre NEXS y el sistema de edición de circuitos para experiencia de usuario perfecta
  • Se conecta con la mayoría de herramientas de Thermo Scientific utilizadas para EFA, PFA y el espacio de edición de circuitos