Search
One key target, p38 MAPK, is activated in response to a variety of extracellular stimuli including osmotic shock, inflammatory cytokines, lipopolysaccharides (LPS), anisomycin, UV light, and growth factors. The activation of p38 MAPK is mediated by several upstream kinases including MAP kinase kinase 3 (MKK3), MAP kinase kinase 6 (MKK6) and MAP kinase kinase 4 (MKK4, also known as SEK1 and JNKK1). These kinases phosphorylate p38 MAPK at threonine 180 and tyrosine 182, resulting in p38 activation.
Western blot analysis of p38 MAPK [pTpY180/182] antibody peptide competition.
Extracts prepared from HEK 293 cells treated with UV irradiation were resolved on a 10% polyacrylamide gel and transferred to PVDF. Prior to incubation with p38 MAPK [pTpY180/182] antibody, the immobilized membranes were incubated with the (lane 1) peptide immunogen, (lane 2) a generic phosphothreonine-containing peptide, (lane 3) a generic phosphotyrosine-containing peptide, (lane 4) the non-phosphorylated peptide corresponding to the phosphopeptide, (lane 5) no peptide, (lane 6) the phosphopeptide derived from the corresponding region of JNK1/2, and (lane 7) the phosphopeptide derived from the corresponding region of ERK1/2. The data show that only the phosphopeptide corresponding to p38 MAPK [pTpY180/182] blocks the antibody signal, thereby demonstrating the specificity of the phosphospecific antibody.
|
Immunohistochemistry of human lung carcinoma tissue labeled with rabbit anti-ERK1/2 [pT185/pY187].
FFPE human lung carcinoma tissue was labeled with rabbit anti-ERK1/2 [pT185/pY187] (5 µg/ml). Tissues were pretreated with EDTA, then detected with
SuperPicTure™ Polymer DAB . Images were taken at 20x magnification. Note nuclear and cytoplasmic staining in tumor cells.
|
ELISA quantification of total p38 MAPK and phosphorylated p38 MAPK [pTpY180/182] in vitro.
Recombinant p38 MAPK was phosphorylated in vitro using MKK6 enzyme. Non-phosphorylated p38 MAPK was used as control. The phosphorylated and non-phosphorylated p38 MAPK were analyzed with p38 MAPK [pTpY180/182] ELISA and p38 MAPK (Total) ELISA. Both forms of the p38 MAK protein were detected using the total ELISA whereas only the phosphorylated p38 MAPK protein was detected with the phosphospecific ELISA.
|
| Target | ABfinity™ Monoclonal Antibodies | Recombinant Oligoclonal Antibodies | Other Primary Antibodies** | Antibody Pairs | ELISA | Proteins | Luminex® Assays |
|---|---|---|---|---|---|---|---|
| BDNF | PHC7074 | LHC7071 | |||||
| BMP2 | 710022 | ||||||
| c-Met | 700261 | CHO0285 | KHO0251 | ||||
| c-Met (Soluble) | CHO0315 | KHO2031 | |||||
| c-Met [pYpYpY1230/34/35] | 700139 | KHO0281 | |||||
| c-Raf [pS289/296/301] | A14039 (Rab) |
| Target | ABfinity™ Monoclonal Antibodies | Recombinant Oligoclonal Antibodies | Other Primary Antibodies** | Antibody Pairs | ELISA | Proteins | Luminex® Assays |
|---|---|---|---|---|---|---|---|
| CREB | 710149 | ||||||
| CREB [pS133] | 700129 | KHO0241 | |||||
| CREB Total | KHO0231 | ||||||
| Cytokine 30-plex (Human) | LHC6003 (Polystyrene) LHC6003M (Magnetic) | ||||||
| EGF | AHG0064 (Rab) | LHG0061 | |||||
| EGFR | AHR5072 (Ms) | KHR9061 | |||||
| EGFR [pY1068] | 701053 | KHR9081 | LHR9081 | ||||
| EGFR [pY1173] | KHR9071 | ||||||
| ERK1 | 138600 (Ms) | ||||||
| ERK1/2 | 44654G (Rab) 136200 (Ms) | KHO0081 | LHO0231 | ||||
| ERK1/2 [pTpY185/187] | 700012 | 44680G (Rab) | KHO0091 | LHO0241 | |||
| ERK1/2 [pTpY202/204] | 368800 (Rab) | ||||||
| ERK5/BMK1 | 44688G (Rab) | ||||||
| ERK5/BMK1 [pTpY218/220] | 44612G (Rab) | ||||||
| FAK | 701094 | 710119 | |||||
| FAK [pT925] | A14010 (Rab) | ||||||
| FGFbasic | KHG0021 KHG0022 | PHG0021 PHG0023 PHG0024 PHG0026 PHG0264 | LHG0021 | ||||
| **Rab=rabbit polyclonal IgG antibody. Ms=mouse monoclonal IgG antibody | |||||||
| Target | ABfinity™ Monoclonal Antibodies | Recombinant Oligoclonal Antibodies | Other Primary Antibodies** | Antibody Pairs | ELISA | Proteins | Luminex® Assays |
|---|---|---|---|---|---|---|---|
| IGF1-R | 710065 | LHO0491 | |||||
| IGF-1R [pYpY1135/1136] | LHO0501 | ||||||
| IL-1α | AHC0912 (Ms) | KAC1191 | PHC0011 PHC0015 PHC0017 | LHC0811 | |||
| IL-1ß | CHC1213 | KHC0011 KHC0012 KHC0014 | PHC0811 PHC0815 PHC0816 | LHC0011 |
| Target | ABfinity™ Monoclonal Antibodies | Recombinant Oligoclonal Antibodies | Other Primary Antibodies** | Antibody Pairs | ELISA | Proteins | Luminex® Assays |
|---|---|---|---|---|---|---|---|
| IL-2 | 701080 | RHCIL220 (Rt) AHC0722 (Ms) | CHC1243 | KHC0021 KAC1241 | PHC0021 PHC0023 PHC0026 PHC0027 | LHC0021 | |
| IL-6 | 710042 | AHC0562 (Ms) | CHC1263 | KAC1261 KHC0061 KHC0061C KHC0062 KHC0063 KHC0064 KHC0064C | PHC0061 PHC0063 PHC0064 PHC0065 PHC0066 | LHC0061 LHC0063 | |
| IR | LHR9111 | ||||||
| IR (B-subunit) | KHR9111 | ||||||
| IR [pY1158] | KHR9121 | ||||||
| IR [pY1328] | KHR9151 | ||||||
| IR [pY1334] | KHR9161 | ||||||
| IR [pY972] | KHR9141 | ||||||
| IR [pYpY1162/1163] | KHR9131 | LHR9131 | |||||
| IRAK1 | 700025 | ||||||
| IRAK4 | 700026 | ||||||
| IκBα [pS32] | KHO0221 | ||||||
| JIP1 (N-Term) | 345200 (Rab) | ||||||
| JIP1/2 (SH3) | 345300 (Rab) | ||||||
| JNK | 710075 | ||||||
| JNK1 | 701048 | 359800 (Ms) | |||||
| JNK1/2 | AHO1362 (Ms) | KHO0121 | LHO0071 | ||||
| JNK1/2 [pTpY183/185] | 44682G (Rab) | KHO0131 | |||||
| JNK2 | AHO1362 (Ms) | ||||||
| **Ms=mouse monoclonal IgG antibody; Rab=rabbit polyclonal IgG antibody. Rt=rat monoclonal IgG antibody | |||||||
| Target | ABfinity™ Monoclonal Antibodies | Recombinant Oligoclonal Antibodies | Other Primary Antibodies | Antibody Pairs | ELISA | Proteins | Luminex® Assays |
|---|---|---|---|---|---|---|---|
| MAPKAPK2 | AHO1242 (Ms) | ||||||
| MAPKAPK2 [pT334] | 44516G (Rab) | ||||||
| MEK1 [pS298] | 44460G (Rab) | ||||||
| MEK1 [pT286] | A14036 (Rab) |
| Target | ABfinity™ Monoclonal Antibodies | Recombinant Oligoclonal Antibodies | Other Primary Antibodies | Antibody Pairs | ELISA | Proteins | Luminex® Assays |
|---|---|---|---|---|---|---|---|
| MEK1 [pT292] | 44458G (Rab) | ||||||
| MEK1 [pT386] | 44462G (Rab) | ||||||
| MEK1/2 [pS222] | 44452 (Rab) | ||||||
| MEK1/2 [pSpS218/222]/222/226 | 44454G (Rab) | ||||||
| MEK2 [pT394] | 44466G (Rab) | ||||||
| MEK3 | 326900 (Ms) | ||||||
| MEK4 [pSpT257/261] | 44474 (Rab) | ||||||
| MEK7 [pSpT271/275] | 44478 (Rab) | ||||||
| MKK7 | 327000 (Ms) | ||||||
| NFκB | KHO0371 | ||||||
| p38 -Beta 2 MAPK | 338700 (Ms) | ||||||
| p38 MAPK | 331300 (Ms) | KHO0061 | LHO0031 | ||||
| p38 MAPK [pTpY180/182] | 701057 | 44684G (Rab) | CHO0075 | KHO0071 | |||
| p53 [pT81] | A13988 (Rab) | ||||||
| Pyk2 [pY402] | 700632 | 710087 | |||||
| RSK1 | AHO1472 (Ms) | ||||||
| RSK1 [pSpS221/227] | 44924G (Rab) | ||||||
| Shc [pY239/240] | A13980 (Rab) | ||||||
| STAT2 | 701105 | 710135 | |||||
| STAT3 [pY705] | KHO0481 | ||||||
| STAT5a | KHO0751 | ||||||
| STAT5a [pY694] | KHO0761 | ||||||
| STAT5b [pY699] | KHO5721 | ||||||
| STAT6 | 701110 | 710140 | |||||
| STAT6 [pY641] | 700247 | 44375M (Ms) | |||||
| TGFb1 | CHC1683 | PHG9202 PHG9203 PHG9204 PHG9211 PHG9214 | |||||
| TNFa | 710162 | CHC1753 | KHC3011 | PHC3011 PHC3013 PHC3015 PHC3016 | LHC3011 | ||
| **Rab=rabbit polyclonal IgG antibody. Ms=mouse monoclonal IgG antibody | |||||||
* This promotion is open to customers in the US and Canada. Discount will apply to orders received by Life Technologies no later than July 31, 2013, or until promotional supplies are depleted, whichever comes first. Customer can use the discount only once. Discount only applies to a maximum of 1 kit per customer. Discount applies to list price in effect at the time order is received by Life Technologies. Cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.