Search
This kit is used to assess the viability of bacterial populations as a function of the membrane integrity of the cell. Cells with a compromised membrane that are considered to be dead or dying will stain red, whereas cells with an intact membrane will stain green.
Micrococcus luteus and Bacillus cereus stained with the LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L7012). When incubated with the SYTO 9 stain and the propidium iodide nucleic acid stain provided in this kit, live bacteria with intact cell membranes fluoresce green and dead bacteria with compromised membranes fluoresce red.
| SYTO 9 | Propidium iodide | |
|---|---|---|
| Excitation/Emission | 480/500 nm | 490/635 nm |
| Standard filter set | FITC | Texas Red |
| Storage conditions | ≤20°C, protect from light | ≤20°C, protect from light |