Spend your time on research, not on instrument maintenance
Quickly and easily calibrate and verify the performance of your Applied Biosystems real-time PCR system using our instrument-specific kits and plates.
Applied Biosystems spectral calibration kits establish the pure dye spectra and multicomponent values (standards) for each reference dye. These standards enable the real-time PCR system to collect pure spectral information, which it uses algorithmically to help ensure accurate data analysis. Depending on which Applied Biosystems instrument you have, there are plate options for individual dye calibration or multi-dye plates that enable simultaneous calibration, which can greatly reduce the time and effort spent on maintenance.
Confirm the performance of your Applied Biosystems instrument with a TaqMan RNase P instrument verification plate. These plates are pre-loaded with real-time PCR reagents and the TaqMan Assay for the single-copy RNase P gene. Each well contains PCR master mix, RNase P primers, FAM dye-labeled probe, and a known concentration of human genomic DNA template.
Always consult your instrument user guide for the recommended calibration/verification schedule and instructions.
Calibration kits and verification plates by instrument model
QuantStudio 1 System, 96-well, 0.2 mL
QuantStudio 5 System, 384-well
QuantStudio 6 Pro System, 384-well
QuantStudio 7 Pro System, 384-well
QuantStudio 7 Pro System, TaqMan Array Card
QuantStudio 6 Flex System, 384-well, with QuantStudio Real-Time PCR Software version 1.6 or later
QuantStudio 6 Flex System, 96-well, 0.1 mL, with QuantStudio Real-Time PCR Software version 1.6 or later
QuantStudio 6 Flex System, 96-well, 0.2 mL, with QuantStudio Real-Time PCR Software version 1.6 or later
QuantStudio 7 Flex System, 384-well, with QuantStudio Real-Time PCR Software version 1.6 or later
QuantStudio 7 Flex System, 96-well, 0.1 mL, with QuantStudio Real-Time PCR Software version 1.6 or later
QuantStudio 7 Flex System, 96-well, 0.2 mL, with QuantStudio Real-Time PCR Software version 1.6 or later
QuantStudio 7 Flex System, TaqMan Array Card
QuantStudio 12K Flex System, 384-well, with QuantStudio Real-Time PCR Software v1.5 or later
QuantStudio 12K Flex System, 96-well, 0.1 mL, with QuantStudio Real-Time PCR Software v1.5 or later
QuantStudio 12K Flex System, 96-well, 0.2 mL, with QuantStudio Real-Time PCR Software v1.4 or earlier
QuantStudio 12K Flex System, 96-well, 0.2 mL, with QuantStudio Real-Time PCR Software v1.5 or later
QuantStudio 12K Flex System, OpenArray
QuantStudio 12 Flex System, TaqMan Array Card
7500 System, 96-well, 0.1 mL
7500 System, 96-well, 0.2 mL
StepOne System, 48-well, 0.1 mL
StepOnePlus System, 96-well, 0.1 mL
Catalog # Name Size Price Qty
4425618 Each
889.65
Online Exclusive
924.00 Save 34.35 (4%)
4371435 Each
989.65
Online Exclusive
1,020.00 Save 30.35 (3%)
4351979 Each
1,060.65
Online Exclusive
1,130.00 Save 69.35 (6%)
ViiA7 System, 384-well with QuantStudio Real-Time PCR Software version 1.6.1
ViiA7 System, 96-well, 0.1 mL with QuantStudio Real-Time PCR Software version 1.6.1
ViiA7 System, 96-well, 0.2 mL with QuantStudio Real-Time PCR Software version 1.6.1
ViiA7 System, TaqMan Array Card
7900HT System, 96-well, 0.1 mL
Catalog # Name Size Price Qty
4425618 Each
889.65
Online Exclusive
924.00 Save 34.35 (4%)
4351653 Each
942.00
4351979 Each
1,060.65
Online Exclusive
1,130.00 Save 69.35 (6%)
7900HT System, 96-well, 0.2 mL
Catalog # Name Size Price Qty
4328639 Each
942.00
4310982 Each
1,058.65
Online Exclusive
1,096.00 Save 37.35 (3%)
7900HT System, TaqMan Array Card
7300 System, 96-well, 0.2 mL
Catalog # Name Size Price Qty
4349182 Each
1,672.00
4350584 Each
1,039.65
Online Exclusive
1,134.00 Save 94.35 (8%)
Types of calibration
Our calibration kits and plates support several types of calibration.
Region of interest (ROI) calibration
ROI calibration maps the positions of the wells on the sample block of the instrument. The software uses the ROI calibration data to associate increases in fluorescence during a run with the wells on the plate or array card. The instrument uses a set of optical filters to distinguish the fluorescence emissions gathered during runs. The software generates a calibration image for each filter to account for minor differences in the optical path.
Successful ROI calibration
During the ROI calibration, the instrument software captures images of the ROI calibration plate or array card at each instrument filter. An ROI calibration passes if the collected image for each filter distinguishes all wells of the ROI plate or array card. Each well in the image must be distinct and visible at the same luminosity relative to the other wells in the image.
Background calibration
Background calibration captures a background image for each optical filter in the absence of sample and reagent and compares the fluorescence from each well to the average for the plate. The software uses that data during subsequent experiments to remove background fluorescence from the experimental data.
Successful background calibration
During the background calibration, the software captures a series of images of the background plate or array card using each instrument filter. The software compares the fluorescence from each well to the average for the plate or array card. A background calibration passes if the collected images for all filters are free of abnormal fluorescence.
Uniformity calibration
Uniformity calibration generates data that allows the software to compensate for the physical effects (non-uniformity of the optical system such as light source) of the instrument filters.
Successful Uniformity calibration
During the uniformity calibration, the software captures a series of images of the ROI plate or array card using each instrument filter. After the calibration, you can review the data. The Analysis Data plot displays the fluorescence data in all filters. A uniformity calibration passes if signals from each well follows a uniform trend.
Dye calibration
Dye calibration collects calibration data generated from the dye standards to characterize and distinguish the individual contribution of each dye in the total fluorescence collected by the instrument. After each run, the instrument software receives data in the form of a raw spectra signal for each reading. The software determines the contribution of the fluorescent dyes used in each reaction by comparing the raw spectra to the pure spectra calibration data. When you save an experiment after analysis, the instrument software stores the pure spectra with the collected fluorescence data for that experiment.
Successful Dye calibration
During the dye calibration, the software extracts the dye calibration data and compares the fluorescence signal generated by each well to the collective spectra for the entire calibration plate or array card. Dye spectra are generally acceptable if they peak within the same filter as their group but diverge slightly at other wavelengths.
Calibration helps enable effective multiplexing
To enable success when multiplexing (which helps save on both samples and costs), our custom dye calibration plates help ensure you realize the same level of sensitivity when running multiple targets in a single well as you would running a just one (singleplex). This gene quantification experiment demonstrates comparable results between reactions performed in individual tubes and in 4-plex reactions.
Comparison of singleplex and multiplex gene expression assays
For Research Use Only. Not for use in diagnostic procedures.