Introduction
Our Invitrogen™ Vybrant™ MTT Cell Proliferation Assay Kit provides a simple method for determination of cell number using standard microplate absorbance readers. Determination of cell growth rates is widely used in the testing of drug action and cytotoxic agents, and screening other biologically active compounds.
Several methods can be used for such determinations, but indirect approaches using fluorescent or chromogenic indicators provide the most rapid and large scale assays. Among such procedures, the MTT assay developed by Mossman 1 is still one of the most versatile and popular assays.
The MTT assay involves the conversion of the water soluble MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) to an insoluble formazan. 2-4 The formazan is then solubilized, and the concentration determined by optical density at 570 nm.
The result is a sensitive assay with excellent linearity up to approximately 10 6 cells per well.
Our MTT Cell Proliferation Assay Kit provides enough material to perform 1000 individual tests using standard 96-well microplates. Following the protocol described below, a complete assay requires an overnight incubation. However, with a slight modification, the whole procedure can be performed in five hours (not including cell preparation time). For additional information concerning the numerous variations and modifications of the MTT assay, consult the citations provided.
Materials
Kit Contents
  • MTT 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MW = 414, Component A), 10 vials, each containing 5 mg
  • SDS sodium dodecyl sulfate (MW = 288, Component B), 10 vials, each containing 1 gm
Storage and Handling
Upon receipt the kit should be stored at 4°C protected from light. Stored properly, the kit components should remain stable for 12 months.
Materials Required but Not Provided
phosphate-buffered saline (PBS), sterile HCl, 0.01 M solution dimethylsulfoxide (DMSO) optional
Experimental Protocol
Reagent Preparation
  1. Prepare a 12 mM MTT stock solution by adding 1 mL of sterile PBS to one 5 mg vial of MTT (Component A). Mix by vortexing or sonication until dissolved. Occasionally there may be some particulate material that will not dissolve; this can be removed by filtration or centrifugation.  Each 5 mg vial of MTT provides sufficient reagent for 100 tests, using 10 µL of the stock solution per well. Once prepared, the MTT solution can be stored for four weeks at 4°C protected from light. 
  2. Add 10 mL of 0.01 M HCl to one tube containing 1 gm of SDS (Component B).  Mix the solution gently by inversion or sonication until the SDS dissolves. Once prepared, the solution should be used promptly. Each tube makes sufficient solution for 100 tests, using 100 µL per well.
Culturing Cells
The culture conditions used to grow the cells can affect the results and must be taken into consideration when analyzing the data. The age of the cultures, number of passages and details of the growth medium can all be important factors. Natural variation in the requirements and growth rates of different cell lines make it difficult to provide precise guidelines for preparing your cells. In general, cells seeded at densities between 5000-–10,000 cells per well should reach optimal population densities within 48-–72 hours. Note that the presence of phenol red in the final assay samples can seriously affect results. We strongly recommend that the cells be cultured in medium free of phenol red, if possible. Alternatively, the final incubation with the MTT can be performed after exchanging the cells into medium free of phenol red.
Labeling Cells
  1. For adherent cells, remove the medium and replace it with 100 µL of fresh culture medium. For non-adherent cells, centrifuge the microplate, pellet the cells, carefully remove as much medium as possible and replace it with 100 µL of fresh medium.
  2. Add 10 µL of the 12 mM MTT stock solution (prepared in step 1.1) to each well. Include a negative control of 10 µL of the MTT stock solution added to 100 µL of medium alone.
  3. Incubate at 37°C for 4 hours. At high cell densities (>100,000 cells per well) the incubation time can be shortened to 2 hours.
  4. Add 100 µL of the SDS-HCl solution (prepared in step 1.2) to each well and mix thoroughly using the pipette.
  5. Incubate the microplate at 37°C for 4– hours in a humidified chamber. Longer incubations will decrease the sensitivity of the assay.9
  6. Mix each sample again using a pipette and read absorbance at 570 nm.
Quick Protocol Option
To shorten the time of the assay it is possible to use DMSO (not provided) as a solubilizing agent to dissolve the formazan. 6
  1. After labeling the cells with MTT, as described above, remove all but 25 µL of medium from the wells. For non-adherent cells it may be necessary to first centrifuge the plates to sediment the cells.
  2. Add 50 µL of DMSO to each well and mix thoroughly with the pipette.
  3. Incubate at 37°C for 10 minutes.
  4. Mix each sample again and read absorbance at 540 nm not 570 nm, as above.
References
  1. J Immunol Methods 65, 55 (1983)
  2. J Neurochem 69, 581 (1997)
  3. Arch Biochem Biophys 303, 474 (1993)
  4. Cancer Res 51, 2515 (1991)
  5. J Immunol Methods 168, 253 (1994)
  6. Cancer Res 47, 943 (1987)
  7. Br J Cancer 56, 279 (1987)
  8. J Immunol Methods 93, 157 (1986)
  9. J Immunol Methods 130, 149 (1990)
MP13154       27-Mar-2002