Search
The HCS NuclearMask Blue Stain is used to measure DNA content and perform cell demarcation in both live and fixed cells on high-content imaging and analysis (HCS) platforms. The versatile HCS NuclearMask Stains survive standard formaldehyde-based fixation and detergent-based permeabilization methods. This protocol produces sufficient stain for one 96-well plate at a staining volume of 100 μL/well.
| HCS NuclearMask Blue | |
|---|---|
| Excitation/Emission (nm) | 350/461 |
| Standard filter set | DAPI |
| EVOS Light Cube | DAPI |
| Storage conditions | 2–6°C |
Cells stained with HCS NuclearMask Blue Stain and imaged with the Thermo Scientific CellInsight High-Content System.