Search Thermo Fisher Scientific
NucRed Live 647 ReadyProbes Reagent is a bright far-red cell-permeant nuclear stain for live or fixed cells.
| NucRed | |
|---|---|
| Excitation/Emission (nm) | 642/661 |
| Standard filter set | Cy®5 |
| EVOS Light Cube | Cy®5 |
| Storage conditions | Room temperature |
Nuclear staining in live hUVECs. hUVECs were grown overnight, stained with NucRed Live 647 ReadyProbes Reagent for 20 minutes at 37°C, and then imaged.