Search
Invitrogen Hoechst 33342 nucleic acid stain is a popular cell-permeant nuclear counterstain that emits blue fluorescence when bound to dsDNA. This dye is often used to distinguish condensed pycnotic nuclei in apoptotic cells and for cell cycle studies in combination with BrdU. It is also available as a solution (Cat. No. H3570).
The Stain-iT simulator can help reduce trial and error and wasted reagents.
| Hoechst 33342 | |
|---|---|
| Excitation/Emission (nm) | 350/461 |
| Standard filter set | DAPI |
| EVOS Light Cube | DAPI |
| Storage conditions | 2–6°C or ≤–20°C |