Search Thermo Fisher Scientific
NucBlue Fixed Cell ReadyProbes Reagent is a bright blue cell-impermeant nuclear stain for fixed cells. It is useful to distinguish the condensed nuclei of apoptotic cells. It is also suitable for staining nuclei in fixed-cell preparations and tissue sections.
First, fix and permeabilize cultured cells with a protocol appropriate for your sample.
| NucBlue | |
|---|---|
| Excitation/Emission (nm) | 360/460 |
| Standard filter set | DAPI |
| EVOS Light Cube | DAPI |
| Storage conditions | Room temperature |
Fixed U2OS cell staining. U2OS cells were fixed and permeabilized using the Image-iT Fixation/Permeabilization Kit. Cells were then with NucBlue Fixed Cell ReadyProbes Reagent for 15 minutes and imaged.